Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 4436914-4437513 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJA3
Entrez Gene ID
9093
Gene Name
DnaJ heat shock protein family (Hsp40) member A3
Gene Aliases
HCA57, TID1, hTID-1
UniGene
Hs.459779
Gene Chromosome Location
Chr.16: 4425805-4456775 on Build GRCh38
Transcripts
NM_001135110.2 NM_001286516.1 NM_005147.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4492984 c.407C>T p.S136F Chr.16: 4437463-4437463 on Build GRCh38
6213200 c.421G>A p.A141T Chr.16: 4437477-4437477 on Build GRCh38
5510781 c.429+2T>C p.? Chr.16: 4437487-4437487 on Build GRCh38
4060783 c.347T>C p.L116P Chr.16: 4437403-4437403 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs199508691 Chr.16: 4437381-4437381 on Build GRCh38
rs991076268 Chr.16: 4437136-4437136 on Build GRCh38
rs911575805 Chr.16: 4437069-4437069 on Build GRCh38
rs978531380 Chr.16: 4437008-4437008 on Build GRCh38
rs758333681 Chr.16: 4437490-4437490 on Build GRCh38
rs895160408 Chr.16: 4436964-4436964 on Build GRCh38
rs964349797 Chr.16: 4437089-4437089 on Build GRCh38
rs750244120 Chr.16: 4437352-4437352 on Build GRCh38
rs928105121 Chr.16: 4436951-4436951 on Build GRCh38
rs185706514 Chr.16: 4437230-4437230 on Build GRCh38
rs966514234 Chr.16: 4437110-4437110 on Build GRCh38
rs951813423 Chr.16: 4437343-4437343 on Build GRCh38
rs774758462 Chr.16: 4437415-4437415 on Build GRCh38
rs867689615 Chr.16: 4437276-4437276 on Build GRCh38
rs987806221 Chr.16: 4437273-4437273 on Build GRCh38
rs374605192 Chr.16: 4437084-4437084 on Build GRCh38
rs781160134 Chr.16: 4437364-4437364 on Build GRCh38
rs763751705 Chr.16: 4437426-4437426 on Build GRCh38
rs779752404 Chr.16: 4437493-4437493 on Build GRCh38
rs748178754 Chr.16: 4437374-4437374 on Build GRCh38
rs568565503 Chr.16: 4437167-4437167 on Build GRCh38
rs763446342 Chr.16: 4437394-4437394 on Build GRCh38
rs1038841577 Chr.16: 4437265-4437265 on Build GRCh38
rs1062706 Chr.16: 4437485-4437485 on Build GRCh38
rs1001777674 Chr.16: 4436943-4436943 on Build GRCh38
rs747243136 Chr.16: 4437359-4437359 on Build GRCh38
rs537540634 Chr.16: 4437168-4437168 on Build GRCh38
rs978226277 Chr.16: 4437114-4437114 on Build GRCh38
rs753408433 Chr.16: 4437441-4437441 on Build GRCh38
rs779884775 Chr.16: 4437356-4437356 on Build GRCh38
rs773642896 Chr.16: 4437389-4437389 on Build GRCh38
rs960835126 Chr.16: 4437128-4437128 on Build GRCh38
rs190038505 Chr.16: 4437377-4437377 on Build GRCh38
rs1008647863 Chr.16: 4437248-4437248 on Build GRCh38
rs564322560 Chr.16: 4437007-4437007 on Build GRCh38
rs554076982 Chr.16: 4437251-4437251 on Build GRCh38
rs895804979 Chr.16: 4436965-4436966 on Build GRCh38
rs1026173541 Chr.16: 4437011-4437011 on Build GRCh38
rs760325462 Chr.16: 4437216-4437216 on Build GRCh38
rs1008783892 Chr.16: 4437051-4437051 on Build GRCh38
rs1021984805 Chr.16: 4437305-4437305 on Build GRCh38
rs764832415 Chr.16: 4437483-4437483 on Build GRCh38
rs897275927 Chr.16: 4437006-4437006 on Build GRCh38
rs573894661 Chr.16: 4437270-4437270 on Build GRCh38
rs890264752 Chr.16: 4437024-4437024 on Build GRCh38
rs1051101108 Chr.16: 4437220-4437220 on Build GRCh38
rs991023927 Chr.16: 4437472-4437472 on Build GRCh38
rs533694094 Chr.16: 4437299-4437299 on Build GRCh38
rs72766601 Chr.16: 4437227-4437227 on Build GRCh38
rs928134748 Chr.16: 4437122-4437122 on Build GRCh38
rs989200952 Chr.16: 4437096-4437096 on Build GRCh38
rs1038861829 Chr.16: 4436993-4436993 on Build GRCh38
rs962017767 Chr.16: 4437303-4437303 on Build GRCh38
rs534120725 Chr.16: 4437233-4437233 on Build GRCh38
rs966755258 Chr.16: 4437323-4437323 on Build GRCh38
rs1010752993 Chr.16: 4436971-4436971 on Build GRCh38
rs768933569 Chr.16: 4437330-4437330 on Build GRCh38
rs987386285 Chr.16: 4437044-4437044 on Build GRCh38
rs546963527 Chr.16: 4437070-4437070 on Build GRCh38
rs112187255 Chr.16: 4437146-4437146 on Build GRCh38
rs867243529 Chr.16: 4437396-4437396 on Build GRCh38
rs918567035 Chr.16: 4437194-4437194 on Build GRCh38
rs1048626226 Chr.16: 4437134-4437134 on Build GRCh38
rs533723096 Chr.16: 4437025-4437025 on Build GRCh38
rs548677534 Chr.16: 4437154-4437154 on Build GRCh38
rs1028776536 Chr.16: 4437016-4437016 on Build GRCh38
rs937620956 Chr.16: 4437144-4437144 on Build GRCh38
rs1022370786 Chr.16: 4437106-4437106 on Build GRCh38
rs760287425 Chr.16: 4437421-4437421 on Build GRCh38
rs753594155 Chr.16: 4437271-4437271 on Build GRCh38
rs771261744 Chr.16: 4437409-4437409 on Build GRCh38
rs1017441998 Chr.16: 4437052-4437052 on Build GRCh38
rs761337674 Chr.16: 4437461-4437461 on Build GRCh38
rs900461183 Chr.16: 4437285-4437285 on Build GRCh38
rs1055360724 Chr.16: 4436958-4436958 on Build GRCh38
rs370449115 Chr.16: 4437362-4437362 on Build GRCh38
rs940340082 Chr.16: 4437081-4437081 on Build GRCh38
rs374734992 Chr.16: 4437253-4437253 on Build GRCh38
rs773839491 Chr.16: 4437159-4437159 on Build GRCh38
rs1020346643 Chr.16: 4436985-4436985 on Build GRCh38
rs939606480 Chr.16: 4436953-4436953 on Build GRCh38
rs566751714 Chr.16: 4437126-4437126 on Build GRCh38
rs150335605 Chr.16: 4437277-4437277 on Build GRCh38
rs941306663 Chr.16: 4436976-4436976 on Build GRCh38

View Full Product Details