Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 66853096-66853625 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA7
Entrez Gene ID
766
Gene Name
carbonic anhydrase 7
Gene Aliases
CAVII
UniGene
Hs.37014
Gene Chromosome Location
Chr.16: 66844379-66854149 on Build GRCh38
Transcripts
NM_001014435.1 NM_005182.2 XM_005256136.3 XM_011523311.1 XM_011523312.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
703923 c.731T>A p.V244E Chr.16: 66853434-66853434 on Build GRCh38
3387526 c.705G>A p.S235S Chr.16: 66853408-66853408 on Build GRCh38
5896274 c.706G>A p.E236K Chr.16: 66853409-66853409 on Build GRCh38
2993814 c.768C>T p.R256R Chr.16: 66853471-66853471 on Build GRCh38
6876010 c.761A>G p.K254R Chr.16: 66853464-66853464 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776317515 Chr.16: 66853514-66853514 on Build GRCh38
rs1030239192 Chr.16: 66853182-66853182 on Build GRCh38
rs758150913 Chr.16: 66853329-66853329 on Build GRCh38
rs781063892 Chr.16: 66853148-66853148 on Build GRCh38
rs949752743 Chr.16: 66853589-66853589 on Build GRCh38
rs564240124 Chr.16: 66853417-66853417 on Build GRCh38
rs749472816 Chr.16: 66853408-66853408 on Build GRCh38
rs1003083605 Chr.16: 66853122-66853122 on Build GRCh38
rs894161726 Chr.16: 66853189-66853189 on Build GRCh38
rs771342266 Chr.16: 66853124-66853124 on Build GRCh38
rs147331762 Chr.16: 66853438-66853438 on Build GRCh38
rs769567330 Chr.16: 66853286-66853286 on Build GRCh38
rs1031167470 Chr.16: 66853317-66853317 on Build GRCh38
rs768132566 Chr.16: 66853527-66853527 on Build GRCh38
rs774703311 Chr.16: 66853497-66853497 on Build GRCh38
rs191188852 Chr.16: 66853167-66853167 on Build GRCh38
rs370643955 Chr.16: 66853504-66853504 on Build GRCh38
rs1006729718 Chr.16: 66853158-66853158 on Build GRCh38
rs910390750 Chr.16: 66853271-66853271 on Build GRCh38
rs768856543 Chr.16: 66853492-66853492 on Build GRCh38
rs763806244 Chr.16: 66853520-66853520 on Build GRCh38
rs775123258 Chr.16: 66853518-66853518 on Build GRCh38
rs985950393 Chr.16: 66853237-66853237 on Build GRCh38
rs756882271 Chr.16: 66853522-66853522 on Build GRCh38
rs752345640 Chr.16: 66853327-66853327 on Build GRCh38
rs111546164 Chr.16: 66853230-66853230 on Build GRCh38
rs958136297 Chr.16: 66853347-66853347 on Build GRCh38
rs772935023 Chr.16: 66853415-66853415 on Build GRCh38
rs577594506 Chr.16: 66853126-66853126 on Build GRCh38
rs758063042 Chr.16: 66853453-66853453 on Build GRCh38
rs549003904 Chr.16: 66853489-66853489 on Build GRCh38
rs1034915501 Chr.16: 66853130-66853130 on Build GRCh38
rs781755614 Chr.16: 66853366-66853366 on Build GRCh38
rs773908448 Chr.16: 66853552-66853552 on Build GRCh38
rs1045398171 Chr.16: 66853602-66853602 on Build GRCh38
rs542432911 Chr.16: 66853323-66853323 on Build GRCh38
rs759410482 Chr.16: 66853427-66853427 on Build GRCh38
rs144059543 Chr.16: 66853414-66853414 on Build GRCh38
rs755263626 Chr.16: 66853356-66853356 on Build GRCh38
rs752257842 Chr.16: 66853448-66853448 on Build GRCh38
rs747410244 Chr.16: 66853349-66853349 on Build GRCh38
rs576104100 Chr.16: 66853135-66853141 on Build GRCh38
rs745904643 Chr.16: 66853373-66853373 on Build GRCh38
rs906812308 Chr.16: 66853604-66853604 on Build GRCh38
rs200227502 Chr.16: 66853402-66853402 on Build GRCh38
rs894653046 Chr.16: 66853474-66853474 on Build GRCh38
rs192545276 Chr.16: 66853354-66853354 on Build GRCh38
rs775569838 Chr.16: 66853407-66853407 on Build GRCh38
rs778214531 Chr.16: 66853332-66853332 on Build GRCh38
rs765276310 Chr.16: 66853517-66853517 on Build GRCh38
rs371594749 Chr.16: 66853491-66853491 on Build GRCh38
rs746931802 Chr.16: 66853216-66853216 on Build GRCh38
rs760578726 Chr.16: 66853416-66853416 on Build GRCh38
rs371139723 Chr.16: 66853516-66853516 on Build GRCh38
rs769909864 Chr.16: 66853401-66853401 on Build GRCh38
rs373390082 Chr.16: 66853503-66853503 on Build GRCh38
rs763896416 Chr.16: 66853461-66853461 on Build GRCh38
rs144835873 Chr.16: 66853480-66853480 on Build GRCh38
rs1047091957 Chr.16: 66853266-66853266 on Build GRCh38
rs183352681 Chr.16: 66853171-66853171 on Build GRCh38
rs909671572 Chr.16: 66853162-66853162 on Build GRCh38
rs376540986 Chr.16: 66853274-66853274 on Build GRCh38
rs187595808 Chr.16: 66853217-66853217 on Build GRCh38
rs751864019 Chr.16: 66853470-66853470 on Build GRCh38
rs954624671 Chr.16: 66853221-66853221 on Build GRCh38
rs1046095842 Chr.16: 66853403-66853403 on Build GRCh38
rs551744859 Chr.16: 66853125-66853125 on Build GRCh38
rs372983215 Chr.16: 66853490-66853490 on Build GRCh38
rs762837486 Chr.16: 66853423-66853423 on Build GRCh38
rs751273926 Chr.16: 66853521-66853521 on Build GRCh38
rs907448062 Chr.16: 66853118-66853118 on Build GRCh38
rs762617011 Chr.16: 66853519-66853519 on Build GRCh38
rs140875572 Chr.16: 66853471-66853471 on Build GRCh38
rs894587158 Chr.16: 66853144-66853144 on Build GRCh38
rs781527720 Chr.16: 66853472-66853472 on Build GRCh38
rs972003808 Chr.16: 66853176-66853176 on Build GRCh38

View Full Product Details