Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 83125288-83125868 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH13
Entrez Gene ID
1012
Gene Name
cadherin 13
Gene Aliases
CDHH, P105
UniGene
Hs.654386
Gene Chromosome Location
Chr.16: 82626794-83796610 on Build GRCh38
Transcripts
NM_001220488.1 NM_001220489.1 NM_001220490.1 NM_001220491.1 NM_001220492.1 NM_001257.4 XM_011522804.2 XM_017022848.1 XM_017022849.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3742050 c.387A>T p.R129S Chr.16: 83125405-83125405 on Build GRCh38
5851605 c.116C>T p.P39L Chr.16: 83125449-83125449 on Build GRCh38
5587470 c.152C>T p.P51L Chr.16: 83125485-83125485 on Build GRCh38
5035840 c.417C>A p.S139S Chr.16: 83125435-83125435 on Build GRCh38
974278 c.447G>T p.E149D Chr.16: 83125465-83125465 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs7196370 Chr.16: 83125745-83125745 on Build GRCh38
rs752885689 Chr.16: 83125802-83125802 on Build GRCh38
rs774549982 Chr.16: 83125439-83125439 on Build GRCh38
rs755508562 Chr.16: 83125358-83125361 on Build GRCh38
rs1033151440 Chr.16: 83125746-83125746 on Build GRCh38
rs111610505 Chr.16: 83125518-83125518 on Build GRCh38
rs182133328 Chr.16: 83125631-83125631 on Build GRCh38
rs781694251 Chr.16: 83125487-83125487 on Build GRCh38
rs569104883 Chr.16: 83125726-83125726 on Build GRCh38
rs566530071 Chr.16: 83125349-83125349 on Build GRCh38
rs781265131 Chr.16: 83125441-83125441 on Build GRCh38
rs941564330 Chr.16: 83125629-83125629 on Build GRCh38
rs1027732304 Chr.16: 83125767-83125767 on Build GRCh38
rs766253004 Chr.16: 83125371-83125371 on Build GRCh38
rs151001954 Chr.16: 83125622-83125622 on Build GRCh38
rs1021197315 Chr.16: 83125512-83125512 on Build GRCh38
rs769274496 Chr.16: 83125535-83125535 on Build GRCh38
rs537777339 Chr.16: 83125495-83125495 on Build GRCh38
rs369911404 Chr.16: 83125490-83125490 on Build GRCh38
rs964709989 Chr.16: 83125759-83125759 on Build GRCh38
rs950893743 Chr.16: 83125772-83125772 on Build GRCh38
rs983287969 Chr.16: 83125776-83125776 on Build GRCh38
rs776104734 Chr.16: 83125424-83125424 on Build GRCh38
rs183071248 Chr.16: 83125836-83125836 on Build GRCh38
rs74819019 Chr.16: 83125722-83125722 on Build GRCh38
rs113563430 Chr.16: 83125395-83125395 on Build GRCh38
rs771045815 Chr.16: 83125330-83125330 on Build GRCh38
rs926668176 Chr.16: 83125835-83125835 on Build GRCh38
rs773782669 Chr.16: 83125623-83125623 on Build GRCh38
rs1038845437 Chr.16: 83125657-83125657 on Build GRCh38
rs754379129 Chr.16: 83125381-83125381 on Build GRCh38
rs542827562 Chr.16: 83125592-83125592 on Build GRCh38
rs768077542 Chr.16: 83125630-83125630 on Build GRCh38
rs527452638 Chr.16: 83125847-83125847 on Build GRCh38
rs753660020 Chr.16: 83125485-83125485 on Build GRCh38
rs573755397 Chr.16: 83125591-83125591 on Build GRCh38
rs182303342 Chr.16: 83125317-83125317 on Build GRCh38
rs767772468 Chr.16: 83125446-83125446 on Build GRCh38
rs747741297 Chr.16: 83125400-83125400 on Build GRCh38
rs922815478 Chr.16: 83125584-83125584 on Build GRCh38
rs905295879 Chr.16: 83125667-83125667 on Build GRCh38
rs191314071 Chr.16: 83125800-83125800 on Build GRCh38
rs546582727 Chr.16: 83125663-83125663 on Build GRCh38
rs941510313 Chr.16: 83125821-83125821 on Build GRCh38
rs186595225 Chr.16: 83125524-83125524 on Build GRCh38
rs777518940 Chr.16: 83125510-83125510 on Build GRCh38
rs569194198 Chr.16: 83125491-83125491 on Build GRCh38
rs770305970 Chr.16: 83125417-83125417 on Build GRCh38
rs778035376 Chr.16: 83125556-83125556 on Build GRCh38
rs866882361 Chr.16: 83125811-83125811 on Build GRCh38
rs546427968 Chr.16: 83125322-83125322 on Build GRCh38
rs768563646 Chr.16: 83125435-83125435 on Build GRCh38
rs987450281 Chr.16: 83125525-83125525 on Build GRCh38
rs79739619 Chr.16: 83125550-83125550 on Build GRCh38
rs1056478784 Chr.16: 83125686-83125686 on Build GRCh38
rs532795605 Chr.16: 83125652-83125652 on Build GRCh38
rs565569747 Chr.16: 83125637-83125637 on Build GRCh38
rs781668418 Chr.16: 83125406-83125406 on Build GRCh38
rs1034660240 Chr.16: 83125432-83125432 on Build GRCh38
rs934252382 Chr.16: 83125586-83125586 on Build GRCh38
rs974282700 Chr.16: 83125828-83125828 on Build GRCh38
rs1044018143 Chr.16: 83125692-83125692 on Build GRCh38
rs866439312 Chr.16: 83125418-83125418 on Build GRCh38
rs780484532 Chr.16: 83125527-83125527 on Build GRCh38
rs140871269 Chr.16: 83125732-83125732 on Build GRCh38
rs375538547 Chr.16: 83125449-83125449 on Build GRCh38
rs562771271 Chr.16: 83125593-83125593 on Build GRCh38
rs963842377 Chr.16: 83125750-83125750 on Build GRCh38
rs1005593951 Chr.16: 83125401-83125401 on Build GRCh38
rs1009723633 Chr.16: 83125730-83125730 on Build GRCh38
rs1040346782 Chr.16: 83125321-83125321 on Build GRCh38
rs560364334 Chr.16: 83125777-83125777 on Build GRCh38
rs200634279 Chr.16: 83125549-83125549 on Build GRCh38
rs1047175860 Chr.16: 83125612-83125612 on Build GRCh38
rs749748090 Chr.16: 83125532-83125532 on Build GRCh38
rs767289896 Chr.16: 83125346-83125346 on Build GRCh38
rs372289566 Chr.16: 83125410-83125410 on Build GRCh38
rs950034264 Chr.16: 83125690-83125690 on Build GRCh38
rs766166879 Chr.16: 83125479-83125479 on Build GRCh38
rs762469698 Chr.16: 83125600-83125600 on Build GRCh38
rs563960757 Chr.16: 83125315-83125315 on Build GRCh38
rs760418496 Chr.16: 83125475-83125475 on Build GRCh38
rs756020883 Chr.16: 83125368-83125368 on Build GRCh38
rs73602301 Chr.16: 83125713-83125713 on Build GRCh38
rs1056989914 Chr.16: 83125841-83125841 on Build GRCh38
rs905551571 Chr.16: 83125694-83125694 on Build GRCh38
rs549074790 Chr.16: 83125721-83125721 on Build GRCh38
rs781281462 Chr.16: 83125843-83125843 on Build GRCh38
rs758189619 Chr.16: 83125402-83125402 on Build GRCh38
rs975450001 Chr.16: 83125756-83125756 on Build GRCh38
rs146022036 Chr.16: 83125760-83125760 on Build GRCh38
rs535657828 Chr.16: 83125377-83125377 on Build GRCh38
rs1020247834 Chr.16: 83125725-83125725 on Build GRCh38
rs142704660 Chr.16: 83125426-83125426 on Build GRCh38
rs1002588779 Chr.16: 83125683-83125683 on Build GRCh38
rs778498316 Chr.16: 83125383-83125383 on Build GRCh38
rs890689435 Chr.16: 83125740-83125740 on Build GRCh38
rs745727903 Chr.16: 83125468-83125468 on Build GRCh38
rs189820472 Chr.16: 83125544-83125544 on Build GRCh38
rs186805844 Chr.16: 83125714-83125714 on Build GRCh38

View Full Product Details