Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7442936-7443515 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF11
Entrez Gene ID
2256
Gene Name
fibroblast growth factor 11
Gene Aliases
FGF-11, FHF-3, FHF3
UniGene
Hs.655193
Gene Chromosome Location
Chr.17: 7438273-7444932 on Build GRCh38
Transcripts
NM_001303460.1 NM_004112.3 NR_130156.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3980293 c.652T>C p.S218P Chr.17: 7443120-7443120 on Build GRCh38
2978899 c.659C>T p.S220F Chr.17: 7443127-7443127 on Build GRCh38
7089161 c.645C>A p.P215P Chr.17: 7443113-7443113 on Build GRCh38
6663473 c.649G>A p.A217T Chr.17: 7443117-7443117 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1009964279 Chr.17: 7443282-7443282 on Build GRCh38
rs546338566 Chr.17: 7443490-7443490 on Build GRCh38
rs552919024 Chr.17: 7443172-7443172 on Build GRCh38
rs4151133 Chr.17: 7443420-7443420 on Build GRCh38
rs747355004 Chr.17: 7443152-7443152 on Build GRCh38
rs933222138 Chr.17: 7443423-7443423 on Build GRCh38
rs368064389 Chr.17: 7443188-7443188 on Build GRCh38
rs979603616 Chr.17: 7443305-7443305 on Build GRCh38
rs938006101 Chr.17: 7443117-7443117 on Build GRCh38
rs535510177 Chr.17: 7443293-7443293 on Build GRCh38
rs769137193 Chr.17: 7443044-7443044 on Build GRCh38
rs929319121 Chr.17: 7443367-7443367 on Build GRCh38
rs1038845797 Chr.17: 7443421-7443421 on Build GRCh38
rs968174040 Chr.17: 7443298-7443298 on Build GRCh38
rs373959662 Chr.17: 7443136-7443136 on Build GRCh38
rs774109701 Chr.17: 7443313-7443313 on Build GRCh38
rs991144281 Chr.17: 7443491-7443491 on Build GRCh38
rs762557980 Chr.17: 7443196-7443196 on Build GRCh38
rs924465563 Chr.17: 7443273-7443273 on Build GRCh38
rs889152599 Chr.17: 7442985-7442985 on Build GRCh38
rs992829387 Chr.17: 7443352-7443352 on Build GRCh38
rs1033892190 Chr.17: 7443312-7443312 on Build GRCh38
rs762788468 Chr.17: 7443131-7443132 on Build GRCh38
rs574237283 Chr.17: 7443231-7443231 on Build GRCh38
rs201355356 Chr.17: 7443113-7443113 on Build GRCh38
rs994011702 Chr.17: 7443365-7443365 on Build GRCh38
rs955775296 Chr.17: 7443006-7443006 on Build GRCh38
rs751302260 Chr.17: 7443104-7443104 on Build GRCh38
rs145080386 Chr.17: 7443160-7443160 on Build GRCh38
rs201225903 Chr.17: 7443194-7443194 on Build GRCh38
rs766309678 Chr.17: 7443138-7443138 on Build GRCh38
rs997214139 Chr.17: 7442996-7442996 on Build GRCh38
rs891442501 Chr.17: 7443488-7443488 on Build GRCh38
rs1021091115 Chr.17: 7443016-7443016 on Build GRCh38
rs1008052741 Chr.17: 7442986-7442986 on Build GRCh38
rs988972957 Chr.17: 7443007-7443007 on Build GRCh38
rs983540174 Chr.17: 7443370-7443370 on Build GRCh38
rs761936340 Chr.17: 7443058-7443061 on Build GRCh38
rs751477117 Chr.17: 7443139-7443139 on Build GRCh38
rs977260283 Chr.17: 7443266-7443266 on Build GRCh38
rs754761305 Chr.17: 7443110-7443110 on Build GRCh38
rs764881955 Chr.17: 7443071-7443071 on Build GRCh38
rs770535837 Chr.17: 7442988-7442988 on Build GRCh38
rs887617308 Chr.17: 7442963-7442963 on Build GRCh38
rs997843492 Chr.17: 7443230-7443230 on Build GRCh38
rs1033854680 Chr.17: 7443458-7443458 on Build GRCh38
rs900457103 Chr.17: 7443228-7443228 on Build GRCh38
rs183482868 Chr.17: 7443002-7443002 on Build GRCh38
rs201773281 Chr.17: 7443114-7443114 on Build GRCh38
rs772998713 Chr.17: 7443077-7443077 on Build GRCh38
rs761389123 Chr.17: 7443065-7443065 on Build GRCh38
rs758586117 Chr.17: 7443139-7443139 on Build GRCh38
rs1038118927 Chr.17: 7443325-7443325 on Build GRCh38
rs767357819 Chr.17: 7443364-7443364 on Build GRCh38
rs914293912 Chr.17: 7443247-7443247 on Build GRCh38
rs765535296 Chr.17: 7443121-7443121 on Build GRCh38
rs768851880 Chr.17: 7443052-7443052 on Build GRCh38
rs1006116232 Chr.17: 7442971-7442971 on Build GRCh38
rs140341581 Chr.17: 7443090-7443090 on Build GRCh38
rs746073027 Chr.17: 7443135-7443135 on Build GRCh38
rs1042778856 Chr.17: 7443493-7443493 on Build GRCh38
rs1026427978 Chr.17: 7443382-7443382 on Build GRCh38
rs775495908 Chr.17: 7443385-7443385 on Build GRCh38
rs189331761 Chr.17: 7443443-7443443 on Build GRCh38
rs763884213 Chr.17: 7443450-7443450 on Build GRCh38
rs772791951 Chr.17: 7443190-7443190 on Build GRCh38
rs146983043 Chr.17: 7443143-7443143 on Build GRCh38
rs552124436 Chr.17: 7443097-7443097 on Build GRCh38
rs759534478 Chr.17: 7443314-7443314 on Build GRCh38
rs891465942 Chr.17: 7443272-7443272 on Build GRCh38
rs535025891 Chr.17: 7443246-7443246 on Build GRCh38
rs899223739 Chr.17: 7443363-7443363 on Build GRCh38
rs779030552 Chr.17: 7443120-7443120 on Build GRCh38
rs3853818 Chr.17: 7442983-7442983 on Build GRCh38
rs762753911 Chr.17: 7443082-7443082 on Build GRCh38
rs372032542 Chr.17: 7443182-7443182 on Build GRCh38
rs947139113 Chr.17: 7443254-7443254 on Build GRCh38
rs199872318 Chr.17: 7443054-7443054 on Build GRCh38
rs933214752 Chr.17: 7443285-7443285 on Build GRCh38
rs772861113 Chr.17: 7443120-7443121 on Build GRCh38
rs532651729 Chr.17: 7443111-7443111 on Build GRCh38

View Full Product Details