Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44148136-44148715 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf53
Entrez Gene ID
78995
Gene Name
chromosome 17 open reading frame 53
Gene Aliases
-
UniGene
Hs.437059
Gene Chromosome Location
Chr.17: 44141902-44162477 on Build GRCh38
Transcripts
NM_001171251.2 NM_001321310.1 NM_001321311.1 NM_024032.4 XM_011525187.1 XM_011525188.1 XM_011525189.1 XM_011525190.1 XM_011525191.2 XM_011525192.1 XM_011525193.1 XM_011525194.1 XM_011525195.1 XM_017025031.1 XM_017025032.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3362167 c.736G>T p.G246C Chr.17: 44148539-44148539 on Build GRCh38
5689126 c.603G>T p.R201R Chr.17: 44148406-44148406 on Build GRCh38
1610314 c.366G>A p.V122V Chr.17: 44148169-44148169 on Build GRCh38
4492325 c.395C>T p.S132L Chr.17: 44148198-44148198 on Build GRCh38
1710356 c.818C>T p.S273F Chr.17: 44148621-44148621 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774790889 Chr.17: 44148270-44148270 on Build GRCh38
rs147358437 Chr.17: 44148689-44148689 on Build GRCh38
rs564465225 Chr.17: 44148602-44148602 on Build GRCh38
rs529091036 Chr.17: 44148165-44148165 on Build GRCh38
rs114501944 Chr.17: 44148511-44148511 on Build GRCh38
rs771078402 Chr.17: 44148403-44148403 on Build GRCh38
rs745451476 Chr.17: 44148369-44148369 on Build GRCh38
rs879213519 Chr.17: 44148672-44148672 on Build GRCh38
rs769188545 Chr.17: 44148185-44148185 on Build GRCh38
rs776797282 Chr.17: 44148404-44148404 on Build GRCh38
rs939252942 Chr.17: 44148632-44148632 on Build GRCh38
rs138840321 Chr.17: 44148214-44148214 on Build GRCh38
rs536677395 Chr.17: 44148207-44148207 on Build GRCh38
rs894857786 Chr.17: 44148351-44148351 on Build GRCh38
rs760895979 Chr.17: 44148274-44148274 on Build GRCh38
rs758305519 Chr.17: 44148330-44148330 on Build GRCh38
rs775844397 Chr.17: 44148527-44148527 on Build GRCh38
rs780541398 Chr.17: 44148528-44148528 on Build GRCh38
rs149621712 Chr.17: 44148504-44148504 on Build GRCh38
rs1022282255 Chr.17: 44148333-44148333 on Build GRCh38
rs116036794 Chr.17: 44148470-44148470 on Build GRCh38
rs568829325 Chr.17: 44148211-44148211 on Build GRCh38
rs372853672 Chr.17: 44148664-44148664 on Build GRCh38
rs760544676 Chr.17: 44148621-44148621 on Build GRCh38
rs143175589 Chr.17: 44148162-44148162 on Build GRCh38
rs775128864 Chr.17: 44148436-44148436 on Build GRCh38
rs758769585 Chr.17: 44148497-44148497 on Build GRCh38
rs142061559 Chr.17: 44148256-44148256 on Build GRCh38
rs144388335 Chr.17: 44148525-44148525 on Build GRCh38
rs765034844 Chr.17: 44148649-44148649 on Build GRCh38
rs373213273 Chr.17: 44148271-44148271 on Build GRCh38
rs756823653 Chr.17: 44148365-44148365 on Build GRCh38
rs749877800 Chr.17: 44148549-44148549 on Build GRCh38
rs759639441 Chr.17: 44148405-44148405 on Build GRCh38
rs920899420 Chr.17: 44148163-44148163 on Build GRCh38
rs759820033 Chr.17: 44148315-44148315 on Build GRCh38
rs762791043 Chr.17: 44148437-44148437 on Build GRCh38
rs754700735 Chr.17: 44148577-44148577 on Build GRCh38
rs1013346318 Chr.17: 44148542-44148542 on Build GRCh38
rs371609858 Chr.17: 44148284-44148284 on Build GRCh38
rs767528407 Chr.17: 44148548-44148548 on Build GRCh38
rs752571578 Chr.17: 44148322-44148322 on Build GRCh38
rs138262467 Chr.17: 44148477-44148477 on Build GRCh38
rs770099032 Chr.17: 44148413-44148413 on Build GRCh38
rs375932726 Chr.17: 44148587-44148587 on Build GRCh38
rs778245506 Chr.17: 44148581-44148581 on Build GRCh38
rs749407735 Chr.17: 44148686-44148686 on Build GRCh38
rs114189169 Chr.17: 44148616-44148616 on Build GRCh38
rs148808091 Chr.17: 44148540-44148540 on Build GRCh38
rs953829273 Chr.17: 44148422-44148422 on Build GRCh38
rs774151326 Chr.17: 44148545-44148545 on Build GRCh38
rs765085851 Chr.17: 44148232-44148232 on Build GRCh38
rs946467682 Chr.17: 44148454-44148454 on Build GRCh38
rs1048769296 Chr.17: 44148192-44148192 on Build GRCh38
rs227584 Chr.17: 44148179-44148179 on Build GRCh38
rs368399208 Chr.17: 44148337-44148337 on Build GRCh38
rs766317453 Chr.17: 44148280-44148281 on Build GRCh38
rs765865097 Chr.17: 44148623-44148623 on Build GRCh38
rs779105010 Chr.17: 44148381-44148381 on Build GRCh38
rs762836620 Chr.17: 44148224-44148224 on Build GRCh38
rs747890538 Chr.17: 44148691-44148691 on Build GRCh38
rs772683739 Chr.17: 44148388-44148388 on Build GRCh38
rs187179200 Chr.17: 44148638-44148638 on Build GRCh38
rs778492460 Chr.17: 44148401-44148401 on Build GRCh38
rs763162380 Chr.17: 44148535-44148535 on Build GRCh38
rs764010009 Chr.17: 44148443-44148443 on Build GRCh38
rs755812416 Chr.17: 44148375-44148375 on Build GRCh38
rs141126495 Chr.17: 44148661-44148661 on Build GRCh38
rs575989228 Chr.17: 44148500-44148500 on Build GRCh38
rs751963411 Chr.17: 44148650-44148650 on Build GRCh38
rs757710553 Chr.17: 44148656-44148656 on Build GRCh38
rs751428715 Chr.17: 44148450-44148450 on Build GRCh38
rs758356836 Chr.17: 44148234-44148234 on Build GRCh38
rs748023037 Chr.17: 44148617-44148617 on Build GRCh38
rs1036799598 Chr.17: 44148219-44148219 on Build GRCh38
rs755690572 Chr.17: 44148471-44148471 on Build GRCh38
rs564833131 Chr.17: 44148246-44148246 on Build GRCh38
rs753654970 Chr.17: 44148226-44148226 on Build GRCh38
rs771890893 Chr.17: 44148618-44148618 on Build GRCh38
rs1028447794 Chr.17: 44148523-44148524 on Build GRCh38
rs938806153 Chr.17: 44148501-44148501 on Build GRCh38
rs761696285 Chr.17: 44148546-44148546 on Build GRCh38
rs370325344 Chr.17: 44148462-44148462 on Build GRCh38
rs752522189 Chr.17: 44148233-44148233 on Build GRCh38
rs779804885 Chr.17: 44148475-44148475 on Build GRCh38
rs370317090 Chr.17: 44148202-44148202 on Build GRCh38
rs746565971 Chr.17: 44148240-44148240 on Build GRCh38
rs751052551 Chr.17: 44148342-44148342 on Build GRCh38
rs533644938 Chr.17: 44148272-44148272 on Build GRCh38
rs374024374 Chr.17: 44148463-44148463 on Build GRCh38
rs377372267 Chr.17: 44148228-44148228 on Build GRCh38
rs748585124 Chr.17: 44148384-44148384 on Build GRCh38
rs776313962 Chr.17: 44148626-44148626 on Build GRCh38
rs111477455 Chr.17: 44148533-44148533 on Build GRCh38
rs756598724 Chr.17: 44148685-44148685 on Build GRCh38
rs148863060 Chr.17: 44148646-44148646 on Build GRCh38
rs752013426 Chr.17: 44148583-44148583 on Build GRCh38
rs757687168 Chr.17: 44148585-44148585 on Build GRCh38
rs765812047 Chr.17: 44148563-44148563 on Build GRCh38
rs920953075 Chr.17: 44148420-44148420 on Build GRCh38
rs749460836 Chr.17: 44148613-44148613 on Build GRCh38
rs747470626 Chr.17: 44148503-44148503 on Build GRCh38
rs754200311 Chr.17: 44148254-44148254 on Build GRCh38
rs753483940 Chr.17: 44148564-44148564 on Build GRCh38
rs764894724 Chr.17: 44148319-44148319 on Build GRCh38
rs1031806922 Chr.17: 44148380-44148380 on Build GRCh38
rs749656674 Chr.17: 44148266-44148266 on Build GRCh38
rs781754508 Chr.17: 44148598-44148598 on Build GRCh38
rs1013193475 Chr.17: 44148301-44148301 on Build GRCh38
rs139526139 Chr.17: 44148693-44148693 on Build GRCh38
rs773135063 Chr.17: 44148620-44148620 on Build GRCh38
rs746013130 Chr.17: 44148512-44148512 on Build GRCh38
rs766747684 Chr.17: 44148217-44148217 on Build GRCh38
rs756148705 Chr.17: 44148612-44148612 on Build GRCh38
rs777229469 Chr.17: 44148236-44148236 on Build GRCh38
rs776653027 Chr.17: 44148311-44148311 on Build GRCh38
rs780756285 Chr.17: 44148366-44148366 on Build GRCh38
rs999449993 Chr.17: 44148354-44148354 on Build GRCh38
rs201296545 Chr.17: 44148304-44148304 on Build GRCh38

View Full Product Details