Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 46782388-46782897 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESD
Entrez Gene ID
2098
Gene Name
esterase D
Gene Aliases
FGH
UniGene
Hs.432491
Gene Chromosome Location
Chr.13: 46771256-46797681 on Build GRCh38
Transcripts
NM_001984.1 XM_005266278.2 XM_011534954.1 XM_017020414.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7300401 c.362A>G p.Y121C Chr.13: 46782686-46782686 on Build GRCh38
3793325 c.331G>A p.E111K Chr.13: 46782717-46782717 on Build GRCh38
1367095 c.292G>C p.D98H Chr.13: 46782756-46782756 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs750967710 Chr.13: 46782785-46782785 on Build GRCh38
rs752375776 Chr.13: 46782666-46782666 on Build GRCh38
rs1031676798 Chr.13: 46782473-46782473 on Build GRCh38
rs993040786 Chr.13: 46782523-46782523 on Build GRCh38
rs772177874 Chr.13: 46782704-46782704 on Build GRCh38
rs998713434 Chr.13: 46782574-46782574 on Build GRCh38
rs769734830 Chr.13: 46782695-46782695 on Build GRCh38
rs373644672 Chr.13: 46782676-46782676 on Build GRCh38
rs760808758 Chr.13: 46782724-46782724 on Build GRCh38
rs968681614 Chr.13: 46782450-46782450 on Build GRCh38
rs547871760 Chr.13: 46782519-46782519 on Build GRCh38
rs536649183 Chr.13: 46782596-46782596 on Build GRCh38
rs778065238 Chr.13: 46782843-46782843 on Build GRCh38
rs755948929 Chr.13: 46782808-46782808 on Build GRCh38
rs932845566 Chr.13: 46782756-46782756 on Build GRCh38
rs767515215 Chr.13: 46782798-46782798 on Build GRCh38
rs777141750 Chr.13: 46782744-46782744 on Build GRCh38
rs761936989 Chr.13: 46782637-46782637 on Build GRCh38
rs762573762 Chr.13: 46782774-46782774 on Build GRCh38
rs763659971 Chr.13: 46782776-46782776 on Build GRCh38
rs776609366 Chr.13: 46782538-46782538 on Build GRCh38
rs759852282 Chr.13: 46782750-46782750 on Build GRCh38
rs190773739 Chr.13: 46782422-46782422 on Build GRCh38
rs899463526 Chr.13: 46782427-46782427 on Build GRCh38
rs983119588 Chr.13: 46782638-46782638 on Build GRCh38
rs904785771 Chr.13: 46782777-46782777 on Build GRCh38
rs757896479 Chr.13: 46782671-46782671 on Build GRCh38
rs768457178 Chr.13: 46782530-46782530 on Build GRCh38
rs1024821652 Chr.13: 46782537-46782537 on Build GRCh38
rs553184353 Chr.13: 46782825-46782825 on Build GRCh38
rs35872816 Chr.13: 46782603-46782604 on Build GRCh38
rs757213416 Chr.13: 46782679-46782679 on Build GRCh38
rs780496230 Chr.13: 46782632-46782640 on Build GRCh38
rs371931425 Chr.13: 46782865-46782865 on Build GRCh38
rs748375942 Chr.13: 46782838-46782838 on Build GRCh38
rs181819737 Chr.13: 46782522-46782522 on Build GRCh38
rs765632914 Chr.13: 46782757-46782757 on Build GRCh38
rs773746308 Chr.13: 46782640-46782640 on Build GRCh38
rs752595190 Chr.13: 46782792-46782792 on Build GRCh38
rs374640883 Chr.13: 46782828-46782828 on Build GRCh38
rs574815169 Chr.13: 46782841-46782841 on Build GRCh38
rs772695871 Chr.13: 46782627-46782629 on Build GRCh38
rs745601294 Chr.13: 46782692-46782692 on Build GRCh38
rs768565458 Chr.13: 46782639-46782639 on Build GRCh38
rs536382450 Chr.13: 46782468-46782468 on Build GRCh38
rs138925242 Chr.13: 46782675-46782675 on Build GRCh38
rs973331665 Chr.13: 46782549-46782549 on Build GRCh38
rs781194315 Chr.13: 46782681-46782681 on Build GRCh38
rs868726203 Chr.13: 46782717-46782717 on Build GRCh38
rs750419665 Chr.13: 46782807-46782807 on Build GRCh38
rs769666240 Chr.13: 46782803-46782803 on Build GRCh38
rs534854590 Chr.13: 46782760-46782760 on Build GRCh38
rs565748501 Chr.13: 46782452-46782452 on Build GRCh38
rs894466855 Chr.13: 46782528-46782528 on Build GRCh38
rs761143179 Chr.13: 46782648-46782648 on Build GRCh38
rs201758721 Chr.13: 46782736-46782736 on Build GRCh38
rs957311039 Chr.13: 46782607-46782607 on Build GRCh38
rs754130803 Chr.13: 46782654-46782654 on Build GRCh38
rs779238877 Chr.13: 46782699-46782699 on Build GRCh38
rs776799278 Chr.13: 46782644-46782645 on Build GRCh38
rs986454905 Chr.13: 46782697-46782697 on Build GRCh38
rs747558987 Chr.13: 46782637-46782640 on Build GRCh38
rs995488170 Chr.13: 46782455-46782455 on Build GRCh38
rs773232227 Chr.13: 46782720-46782720 on Build GRCh38
rs35111834 Chr.13: 46782700-46782700 on Build GRCh38
rs761988558 Chr.13: 46782653-46782654 on Build GRCh38
rs764817848 Chr.13: 46782655-46782655 on Build GRCh38
rs975744012 Chr.13: 46782646-46782646 on Build GRCh38
rs766761628 Chr.13: 46782652-46782652 on Build GRCh38
rs928955469 Chr.13: 46782604-46782604 on Build GRCh38

View Full Product Details