Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 14625416-14625672 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRE3
Entrez Gene ID
84658
Gene Name
adhesion G protein-coupled receptor E3
Gene Aliases
EMR3
UniGene
Hs.658712
Gene Chromosome Location
Chr.19: 14600175-14674918 on Build GRCh38
Transcripts
NM_001289158.1 NM_001289159.1 NM_032571.4 XM_011528374.2 XM_017027383.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4473971 c.1886C>T p.S629F Chr.19: 14625526-14625526 on Build GRCh38
6897650 c.1822C>T p.Q608* Chr.19: 14625590-14625590 on Build GRCh38
4531980 c.1843G>A p.E615K Chr.19: 14625569-14625569 on Build GRCh38
6021801 c.1882C>A p.L628I Chr.19: 14625530-14625530 on Build GRCh38
6083671 c.1850T>A p.V617E Chr.19: 14625562-14625562 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs918684319 Chr.19: 14625482-14625482 on Build GRCh38
rs571110477 Chr.19: 14625594-14625594 on Build GRCh38
rs773360659 Chr.19: 14625625-14625625 on Build GRCh38
rs369248321 Chr.19: 14625620-14625620 on Build GRCh38
rs780625939 Chr.19: 14625460-14625460 on Build GRCh38
rs746648681 Chr.19: 14625488-14625488 on Build GRCh38
rs964037319 Chr.19: 14625599-14625599 on Build GRCh38
rs761828928 Chr.19: 14625623-14625623 on Build GRCh38
rs902287953 Chr.19: 14625617-14625617 on Build GRCh38
rs764381295 Chr.19: 14625448-14625448 on Build GRCh38
rs147270469 Chr.19: 14625564-14625564 on Build GRCh38
rs149148345 Chr.19: 14625563-14625563 on Build GRCh38
rs780371799 Chr.19: 14625474-14625474 on Build GRCh38
rs895309023 Chr.19: 14625621-14625621 on Build GRCh38
rs770900694 Chr.19: 14625494-14625494 on Build GRCh38
rs376729505 Chr.19: 14625495-14625495 on Build GRCh38
rs117374816 Chr.19: 14625514-14625514 on Build GRCh38
rs750966022 Chr.19: 14625468-14625468 on Build GRCh38
rs201958148 Chr.19: 14625569-14625569 on Build GRCh38
rs777245566 Chr.19: 14625641-14625641 on Build GRCh38
rs750804682 Chr.19: 14625547-14625547 on Build GRCh38
rs374894225 Chr.19: 14625581-14625581 on Build GRCh38
rs774602580 Chr.19: 14625531-14625531 on Build GRCh38
rs757002547 Chr.19: 14625577-14625577 on Build GRCh38
rs201506202 Chr.19: 14625455-14625455 on Build GRCh38
rs762634650 Chr.19: 14625522-14625522 on Build GRCh38
rs368065969 Chr.19: 14625540-14625540 on Build GRCh38
rs752218548 Chr.19: 14625496-14625496 on Build GRCh38
rs1016980972 Chr.19: 14625470-14625470 on Build GRCh38
rs780096170 Chr.19: 14625613-14625613 on Build GRCh38
rs139987545 Chr.19: 14625503-14625503 on Build GRCh38
rs761988614 Chr.19: 14625532-14625532 on Build GRCh38
rs775223026 Chr.19: 14625519-14625519 on Build GRCh38
rs757783411 Chr.19: 14625478-14625478 on Build GRCh38
rs757988080 Chr.19: 14625565-14625565 on Build GRCh38
rs756672370 Chr.19: 14625471-14625471 on Build GRCh38
rs200554519 Chr.19: 14625466-14625466 on Build GRCh38
rs761171373 Chr.19: 14625639-14625639 on Build GRCh38
rs777637528 Chr.19: 14625481-14625481 on Build GRCh38
rs777402584 Chr.19: 14625575-14625575 on Build GRCh38
rs780943563 Chr.19: 14625579-14625579 on Build GRCh38
rs199670503 Chr.19: 14625520-14625520 on Build GRCh38
rs34381727 Chr.19: 14625476-14625476 on Build GRCh38
rs1035172021 Chr.19: 14625450-14625450 on Build GRCh38
rs751299196 Chr.19: 14625576-14625576 on Build GRCh38
rs537124035 Chr.19: 14625518-14625518 on Build GRCh38
rs762221824 Chr.19: 14625464-14625464 on Build GRCh38
rs999684445 Chr.19: 14625562-14625562 on Build GRCh38
rs74560369 Chr.19: 14625482-14625482 on Build GRCh38
rs61729083 Chr.19: 14625504-14625504 on Build GRCh38
rs1012387428 Chr.19: 14625622-14625622 on Build GRCh38
rs749157123 Chr.19: 14625619-14625619 on Build GRCh38

View Full Product Details