Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41433476-41433747 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP5SL
Entrez Gene ID
55101
Gene Name
ATP5S like
Gene Aliases
-
UniGene
Hs.351099
Gene Chromosome Location
Chr.19: 41431318-41447822 on Build GRCh38
Transcripts
NM_001167867.1 NM_001167868.1 NM_001167869.1 NM_001167870.1 NM_001167871.1 NM_001320838.1 NM_001320839.1 NM_001320840.1 NM_001320841.1 NM_001320844.1 NM_018035.2 NR_030765.1 NR_135476.1 NR_135477.1 NR_135478.1 XM_011527065.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3227133 c.433C>T p.L145F Chr.19: 41433537-41433537 on Build GRCh38
5418292 c.418G>C p.G140R Chr.19: 41433552-41433552 on Build GRCh38
6607991 c.334G>A p.G112S Chr.19: 41433636-41433636 on Build GRCh38
6866251 c.382G>A p.A128T Chr.19: 41433588-41433588 on Build GRCh38
1179555 c.430C>A p.L144I Chr.19: 41433540-41433540 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782310524 Chr.19: 41433697-41433697 on Build GRCh38
rs782379341 Chr.19: 41433528-41433528 on Build GRCh38
rs576191896 Chr.19: 41433578-41433578 on Build GRCh38
rs782238718 Chr.19: 41433525-41433525 on Build GRCh38
rs1007204864 Chr.19: 41433512-41433512 on Build GRCh38
rs377249221 Chr.19: 41433721-41433721 on Build GRCh38
rs782141061 Chr.19: 41433705-41433705 on Build GRCh38
rs782479831 Chr.19: 41433715-41433715 on Build GRCh38
rs372920057 Chr.19: 41433710-41433710 on Build GRCh38
rs763590381 Chr.19: 41433650-41433650 on Build GRCh38
rs782066981 Chr.19: 41433577-41433577 on Build GRCh38
rs782757473 Chr.19: 41433689-41433689 on Build GRCh38
rs782044176 Chr.19: 41433670-41433670 on Build GRCh38
rs782002045 Chr.19: 41433560-41433560 on Build GRCh38
rs782688425 Chr.19: 41433529-41433529 on Build GRCh38
rs782522790 Chr.19: 41433725-41433725 on Build GRCh38
rs782325715 Chr.19: 41433681-41433681 on Build GRCh38
rs782361522 Chr.19: 41433643-41433643 on Build GRCh38
rs373886313 Chr.19: 41433501-41433501 on Build GRCh38
rs773742192 Chr.19: 41433682-41433682 on Build GRCh38
rs781805785 Chr.19: 41433506-41433506 on Build GRCh38
rs11539797 Chr.19: 41433592-41433592 on Build GRCh38
rs201700305 Chr.19: 41433632-41433632 on Build GRCh38
rs782679228 Chr.19: 41433692-41433693 on Build GRCh38
rs148919104 Chr.19: 41433638-41433638 on Build GRCh38
rs781788109 Chr.19: 41433576-41433576 on Build GRCh38
rs782499765 Chr.19: 41433688-41433688 on Build GRCh38
rs372385852 Chr.19: 41433537-41433537 on Build GRCh38
rs368134749 Chr.19: 41433536-41433536 on Build GRCh38
rs140377634 Chr.19: 41433644-41433644 on Build GRCh38
rs782094200 Chr.19: 41433688-41433688 on Build GRCh38
rs782488726 Chr.19: 41433599-41433599 on Build GRCh38
rs782262645 Chr.19: 41433649-41433649 on Build GRCh38
rs141059890 Chr.19: 41433555-41433555 on Build GRCh38
rs782289487 Chr.19: 41433591-41433591 on Build GRCh38
rs375627343 Chr.19: 41433640-41433640 on Build GRCh38
rs782147616 Chr.19: 41433570-41433570 on Build GRCh38
rs782475265 Chr.19: 41433513-41433513 on Build GRCh38
rs782592311 Chr.19: 41433611-41433611 on Build GRCh38
rs781964026 Chr.19: 41433545-41433545 on Build GRCh38
rs781994996 Chr.19: 41433701-41433701 on Build GRCh38
rs762545674 Chr.19: 41433572-41433572 on Build GRCh38
rs199899695 Chr.19: 41433723-41433723 on Build GRCh38
rs1015957565 Chr.19: 41433523-41433523 on Build GRCh38
rs529644883 Chr.19: 41433601-41433601 on Build GRCh38
rs782406286 Chr.19: 41433668-41433668 on Build GRCh38
rs782400350 Chr.19: 41433628-41433628 on Build GRCh38
rs782696045 Chr.19: 41433709-41433709 on Build GRCh38
rs145875700 Chr.19: 41433597-41433597 on Build GRCh38
rs782456027 Chr.19: 41433609-41433609 on Build GRCh38
rs782602159 Chr.19: 41433515-41433515 on Build GRCh38
rs782095277 Chr.19: 41433553-41433553 on Build GRCh38
rs978707978 Chr.19: 41433664-41433664 on Build GRCh38
rs370082149 Chr.19: 41433669-41433669 on Build GRCh38
rs934727861 Chr.19: 41433678-41433678 on Build GRCh38
rs781809092 Chr.19: 41433711-41433711 on Build GRCh38
rs781881409 Chr.19: 41433605-41433605 on Build GRCh38

View Full Product Details