Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 15243920-15244497 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB9
Entrez Gene ID
140462
Gene Name
ankyrin repeat and SOCS box containing 9
Gene Aliases
-
UniGene
Hs.19404
Gene Chromosome Location
Chr.X: 15243987-15270476 on Build GRCh38
Transcripts
NM_001031739.2 NM_001168530.1 NM_001168531.1 NM_024087.2 XM_005274446.1 XM_011545458.1 XM_011545460.1 XM_017029283.1 XM_017029284.1 XM_017029285.1 XM_017029286.1

SNP Detail

SNP ID Chromosome Location
rs761614725 Chr.X: 15244125-15244125 on Build GRCh38
rs767199688 Chr.X: 15244160-15244160 on Build GRCh38
rs1021108426 Chr.X: 15244147-15244147 on Build GRCh38
rs950328612 Chr.X: 15243965-15243965 on Build GRCh38
rs760570015 Chr.X: 15244203-15244203 on Build GRCh38
rs774025955 Chr.X: 15244108-15244108 on Build GRCh38
rs775689858 Chr.X: 15244003-15244003 on Build GRCh38
rs916616324 Chr.X: 15244263-15244263 on Build GRCh38
rs779348623 Chr.X: 15244272-15244272 on Build GRCh38
rs1054596468 Chr.X: 15244124-15244124 on Build GRCh38
rs890161077 Chr.X: 15244419-15244419 on Build GRCh38
rs14252 Chr.X: 15244310-15244310 on Build GRCh38
rs17216316 Chr.X: 15244180-15244180 on Build GRCh38
rs753295643 Chr.X: 15244293-15244293 on Build GRCh38
rs1010424901 Chr.X: 15244144-15244144 on Build GRCh38
rs374142219 Chr.X: 15244107-15244107 on Build GRCh38
rs778131623 Chr.X: 15244154-15244154 on Build GRCh38
rs950080746 Chr.X: 15243992-15243992 on Build GRCh38
rs936210950 Chr.X: 15244353-15244353 on Build GRCh38
rs906736795 Chr.X: 15243961-15243961 on Build GRCh38
rs924043324 Chr.X: 15244173-15244173 on Build GRCh38
rs1010810603 Chr.X: 15244406-15244406 on Build GRCh38
rs770377450 Chr.X: 15244399-15244399 on Build GRCh38
rs1047483144 Chr.X: 15244398-15244398 on Build GRCh38
rs761932422 Chr.X: 15244463-15244463 on Build GRCh38
rs964564268 Chr.X: 15244162-15244162 on Build GRCh38
rs1053697597 Chr.X: 15244230-15244230 on Build GRCh38
rs1048294639 Chr.X: 15243972-15243972 on Build GRCh38
rs906655122 Chr.X: 15244342-15244342 on Build GRCh38
rs774246796 Chr.X: 15244457-15244457 on Build GRCh38
rs201067436 Chr.X: 15244454-15244454 on Build GRCh38
rs17216302 Chr.X: 15244056-15244056 on Build GRCh38
rs9560 Chr.X: 15244250-15244250 on Build GRCh38
rs183497389 Chr.X: 15243988-15243988 on Build GRCh38
rs778561552 Chr.X: 15244322-15244322 on Build GRCh38
rs948296912 Chr.X: 15244267-15244267 on Build GRCh38
rs954518446 Chr.X: 15244149-15244149 on Build GRCh38
rs768887734 Chr.X: 15244091-15244091 on Build GRCh38
rs768778112 Chr.X: 15244431-15244431 on Build GRCh38
rs950587722 Chr.X: 15244242-15244242 on Build GRCh38
rs752349319 Chr.X: 15244000-15244000 on Build GRCh38
rs112232773 Chr.X: 15244409-15244409 on Build GRCh38
rs184687335 Chr.X: 15244423-15244423 on Build GRCh38
rs964966786 Chr.X: 15244075-15244075 on Build GRCh38
rs746428099 Chr.X: 15244354-15244354 on Build GRCh38
rs1008975624 Chr.X: 15244155-15244155 on Build GRCh38
rs1026071535 Chr.X: 15243963-15243965 on Build GRCh38
rs772248162 Chr.X: 15244473-15244473 on Build GRCh38

View Full Product Details