Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 41343046-41343608 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX3X
Entrez Gene ID
1654
Gene Name
DEAD-box helicase 3, X-linked
Gene Aliases
CAP-Rf, DBX, DDX14, DDX3, HLP2, MRX102
UniGene
Hs.728563
Gene Chromosome Location
Chr.X: 41333308-41364472 on Build GRCh38
Transcripts
NM_001193416.2 NM_001193417.2 NM_001356.4 NR_126093.1 XM_011543892.2 XM_017029313.1 XM_017029314.1

SNP Detail

SNP ID Chromosome Location
rs1631136 Chr.X: 41343459-41343459 on Build GRCh38
rs869312692 Chr.X: 41343291-41343291 on Build GRCh38
rs374618680 Chr.X: 41343524-41343524 on Build GRCh38
rs765195859 Chr.X: 41343110-41343110 on Build GRCh38
rs72460118 Chr.X: 41343562-41343563 on Build GRCh38
rs781025119 Chr.X: 41343456-41343456 on Build GRCh38
rs373063765 Chr.X: 41343196-41343196 on Build GRCh38
rs754497446 Chr.X: 41343569-41343570 on Build GRCh38
rs971167527 Chr.X: 41343111-41343111 on Build GRCh38
rs1032666321 Chr.X: 41343359-41343359 on Build GRCh38
rs920167322 Chr.X: 41343504-41343504 on Build GRCh38
rs773721753 Chr.X: 41343421-41343421 on Build GRCh38
rs6610547 Chr.X: 41343559-41343559 on Build GRCh38
rs755750366 Chr.X: 41343268-41343268 on Build GRCh38
rs750715499 Chr.X: 41343184-41343184 on Build GRCh38
rs753872224 Chr.X: 41343082-41343082 on Build GRCh38
rs1039007665 Chr.X: 41343123-41343123 on Build GRCh38
rs777274761 Chr.X: 41343272-41343272 on Build GRCh38
rs778286223 Chr.X: 41343383-41343383 on Build GRCh38
rs59846147 Chr.X: 41343476-41343476 on Build GRCh38
rs763097796 Chr.X: 41343185-41343185 on Build GRCh38
rs762956543 Chr.X: 41343199-41343199 on Build GRCh38
rs981340205 Chr.X: 41343583-41343584 on Build GRCh38
rs747425977 Chr.X: 41343400-41343400 on Build GRCh38
rs765444398 Chr.X: 41343181-41343181 on Build GRCh38
rs766133574 Chr.X: 41343190-41343190 on Build GRCh38
rs912714230 Chr.X: 41343519-41343519 on Build GRCh38
rs778066345 Chr.X: 41343377-41343379 on Build GRCh38
rs1049962341 Chr.X: 41343149-41343149 on Build GRCh38
rs1021070719 Chr.X: 41343208-41343208 on Build GRCh38
rs192995367 Chr.X: 41343088-41343088 on Build GRCh38
rs1050446380 Chr.X: 41343513-41343513 on Build GRCh38
rs886039719 Chr.X: 41343307-41343307 on Build GRCh38
rs915761966 Chr.X: 41343090-41343090 on Build GRCh38
rs1040940182 Chr.X: 41343432-41343432 on Build GRCh38
rs374232164 Chr.X: 41343388-41343388 on Build GRCh38
rs769179688 Chr.X: 41343166-41343166 on Build GRCh38
rs767609622 Chr.X: 41343204-41343204 on Build GRCh38
rs751416043 Chr.X: 41343365-41343365 on Build GRCh38
rs754913084 Chr.X: 41343197-41343197 on Build GRCh38
rs776673110 Chr.X: 41343167-41343167 on Build GRCh38
rs981774747 Chr.X: 41343126-41343126 on Build GRCh38
rs758794511 Chr.X: 41343366-41343371 on Build GRCh38
rs898099964 Chr.X: 41343452-41343452 on Build GRCh38
rs746419382 Chr.X: 41343512-41343512 on Build GRCh38
rs772645508 Chr.X: 41343514-41343514 on Build GRCh38
rs369874066 Chr.X: 41343381-41343381 on Build GRCh38
rs375996245 Chr.X: 41343258-41343258 on Build GRCh38
rs902634573 Chr.X: 41343541-41343541 on Build GRCh38
rs944212751 Chr.X: 41343520-41343520 on Build GRCh38
rs141846961 Chr.X: 41343145-41343145 on Build GRCh38
rs754612891 Chr.X: 41343407-41343407 on Build GRCh38
rs1631137 Chr.X: 41343460-41343460 on Build GRCh38
rs567580408 Chr.X: 41343135-41343135 on Build GRCh38
rs924587210 Chr.X: 41343150-41343150 on Build GRCh38
rs1000898735 Chr.X: 41343308-41343308 on Build GRCh38
rs959358916 Chr.X: 41343366-41343366 on Build GRCh38
rs941717938 Chr.X: 41343083-41343083 on Build GRCh38
rs200078632 Chr.X: 41343363-41343363 on Build GRCh38
rs907652791 Chr.X: 41343453-41343453 on Build GRCh38
rs1027332454 Chr.X: 41343479-41343479 on Build GRCh38
rs752955693 Chr.X: 41343365-41343369 on Build GRCh38
rs1024775881 Chr.X: 41343420-41343420 on Build GRCh38
rs796052237 Chr.X: 41343271-41343272 on Build GRCh38
rs983138452 Chr.X: 41343426-41343426 on Build GRCh38
rs780055981 Chr.X: 41343155-41343155 on Build GRCh38
rs753358534 Chr.X: 41343135-41343138 on Build GRCh38
rs1001272847 Chr.X: 41343542-41343542 on Build GRCh38
rs761925135 Chr.X: 41343176-41343176 on Build GRCh38
rs875989803 Chr.X: 41343249-41343249 on Build GRCh38

View Full Product Details