Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 111246315-111246903 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPN6
Entrez Gene ID
827
Gene Name
calpain 6
Gene Aliases
CANPX, CAPNX, CalpM, DJ914P14.1
UniGene
Hs.496593
Gene Chromosome Location
Chr.X: 111245099-111270546 on Build GRCh38
Transcripts
NM_014289.3

SNP Detail

SNP ID Chromosome Location
rs751042151 Chr.X: 111246571-111246571 on Build GRCh38
rs749934171 Chr.X: 111246726-111246726 on Build GRCh38
rs138687868 Chr.X: 111246723-111246723 on Build GRCh38
rs780070196 Chr.X: 111246824-111246824 on Build GRCh38
rs970567068 Chr.X: 111246513-111246513 on Build GRCh38
rs1012089002 Chr.X: 111246460-111246460 on Build GRCh38
rs749809071 Chr.X: 111246658-111246658 on Build GRCh38
rs748188014 Chr.X: 111246685-111246685 on Build GRCh38
rs762939222 Chr.X: 111246557-111246557 on Build GRCh38
rs751170869 Chr.X: 111246375-111246375 on Build GRCh38
rs193921059 Chr.X: 111246746-111246746 on Build GRCh38
rs762841737 Chr.X: 111246730-111246730 on Build GRCh38
rs772438919 Chr.X: 111246686-111246686 on Build GRCh38
rs368557380 Chr.X: 111246782-111246782 on Build GRCh38
rs765226420 Chr.X: 111246556-111246556 on Build GRCh38
rs771611775 Chr.X: 111246823-111246823 on Build GRCh38
rs761242600 Chr.X: 111246762-111246762 on Build GRCh38
rs779163358 Chr.X: 111246676-111246676 on Build GRCh38
rs1032345859 Chr.X: 111246431-111246431 on Build GRCh38
rs748244880 Chr.X: 111246789-111246789 on Build GRCh38
rs982738925 Chr.X: 111246478-111246478 on Build GRCh38
rs943917558 Chr.X: 111246521-111246521 on Build GRCh38
rs147112567 Chr.X: 111246694-111246694 on Build GRCh38
rs374298832 Chr.X: 111246597-111246597 on Build GRCh38
rs779944838 Chr.X: 111246547-111246547 on Build GRCh38
rs34550022 Chr.X: 111246643-111246643 on Build GRCh38
rs760947724 Chr.X: 111246714-111246714 on Build GRCh38
rs764021932 Chr.X: 111246399-111246399 on Build GRCh38
rs199510760 Chr.X: 111246804-111246804 on Build GRCh38
rs767006015 Chr.X: 111246763-111246763 on Build GRCh38
rs775337373 Chr.X: 111246727-111246727 on Build GRCh38
rs139354933 Chr.X: 111246621-111246621 on Build GRCh38
rs771157688 Chr.X: 111246810-111246810 on Build GRCh38
rs1027464417 Chr.X: 111246435-111246435 on Build GRCh38
rs758530292 Chr.X: 111246365-111246365 on Build GRCh38
rs775183734 Chr.X: 111246811-111246811 on Build GRCh38
rs766693553 Chr.X: 111246777-111246777 on Build GRCh38
rs774548980 Chr.X: 111246526-111246526 on Build GRCh38
rs755562953 Chr.X: 111246779-111246779 on Build GRCh38
rs756062996 Chr.X: 111246546-111246546 on Build GRCh38
rs756900127 Chr.X: 111246636-111246636 on Build GRCh38
rs763646342 Chr.X: 111246565-111246565 on Build GRCh38
rs376205120 Chr.X: 111246527-111246527 on Build GRCh38
rs933505044 Chr.X: 111246364-111246364 on Build GRCh38
rs921083346 Chr.X: 111246598-111246598 on Build GRCh38
rs902628512 Chr.X: 111246384-111246384 on Build GRCh38
rs951413263 Chr.X: 111246436-111246436 on Build GRCh38
rs1000867001 Chr.X: 111246428-111246428 on Build GRCh38
rs201824799 Chr.X: 111246662-111246662 on Build GRCh38
rs1024864899 Chr.X: 111246489-111246489 on Build GRCh38
rs754725862 Chr.X: 111246773-111246773 on Build GRCh38
rs895154714 Chr.X: 111246458-111246458 on Build GRCh38
rs780763063 Chr.X: 111246622-111246622 on Build GRCh38
rs750096166 Chr.X: 111246768-111246768 on Build GRCh38
rs780449744 Chr.X: 111246645-111246645 on Build GRCh38
rs778556558 Chr.X: 111246778-111246778 on Build GRCh38
rs901984639 Chr.X: 111246355-111246355 on Build GRCh38
rs758618487 Chr.X: 111246794-111246794 on Build GRCh38
rs777986875 Chr.X: 111246795-111246795 on Build GRCh38
rs773861656 Chr.X: 111246748-111246748 on Build GRCh38
rs912518566 Chr.X: 111246493-111246493 on Build GRCh38

View Full Product Details