Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 112775542-112776122 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMOT
Entrez Gene ID
154796
Gene Name
angiomotin
Gene Aliases
-
UniGene
Hs.528051
Gene Chromosome Location
Chr.X: 112774877-112840946 on Build GRCh38
Transcripts
NM_001113490.1 NM_133265.2 XM_005262087.1 XM_005262090.1 XM_011530875.2 XM_017029289.1

SNP Detail

SNP ID Chromosome Location
rs976649097 Chr.X: 112775796-112775796 on Build GRCh38
rs369130967 Chr.X: 112775991-112775991 on Build GRCh38
rs1016228322 Chr.X: 112775941-112775941 on Build GRCh38
rs35606486 Chr.X: 112775957-112775958 on Build GRCh38
rs955113638 Chr.X: 112776046-112776047 on Build GRCh38
rs895144156 Chr.X: 112776046-112776046 on Build GRCh38
rs766595376 Chr.X: 112775669-112775669 on Build GRCh38
rs755398190 Chr.X: 112775801-112775801 on Build GRCh38
rs143172417 Chr.X: 112775678-112775678 on Build GRCh38
rs1042912242 Chr.X: 112775888-112775888 on Build GRCh38
rs1004417644 Chr.X: 112775725-112775725 on Build GRCh38
rs754251717 Chr.X: 112775753-112775753 on Build GRCh38
rs763546169 Chr.X: 112776048-112776048 on Build GRCh38
rs753248969 Chr.X: 112776051-112776051 on Build GRCh38
rs989245066 Chr.X: 112775826-112775826 on Build GRCh38
rs886565340 Chr.X: 112775859-112775859 on Build GRCh38
rs765013865 Chr.X: 112775815-112775815 on Build GRCh38
rs138308883 Chr.X: 112776093-112776093 on Build GRCh38
rs779360256 Chr.X: 112775973-112775973 on Build GRCh38
rs1049066603 Chr.X: 112776068-112776068 on Build GRCh38
rs1057350851 Chr.X: 112775838-112775838 on Build GRCh38
rs965241646 Chr.X: 112775767-112775767 on Build GRCh38
rs549116776 Chr.X: 112775957-112775957 on Build GRCh38
rs1015428266 Chr.X: 112775727-112775727 on Build GRCh38
rs758309366 Chr.X: 112775900-112775900 on Build GRCh38
rs1009089047 Chr.X: 112776089-112776091 on Build GRCh38
rs945154455 Chr.X: 112775870-112775870 on Build GRCh38
rs35667754 Chr.X: 112775877-112775878 on Build GRCh38
rs761024403 Chr.X: 112775603-112775603 on Build GRCh38
rs915011511 Chr.X: 112775849-112775849 on Build GRCh38
rs988514948 Chr.X: 112775702-112775702 on Build GRCh38
rs947294686 Chr.X: 112775773-112775773 on Build GRCh38
rs986965906 Chr.X: 112775618-112775618 on Build GRCh38
rs752587391 Chr.X: 112775848-112775848 on Build GRCh38
rs1029215521 Chr.X: 112776023-112776023 on Build GRCh38
rs996405315 Chr.X: 112775960-112775960 on Build GRCh38
rs191533590 Chr.X: 112775672-112775672 on Build GRCh38
rs750656990 Chr.X: 112775673-112775673 on Build GRCh38
rs953739255 Chr.X: 112775817-112775817 on Build GRCh38
rs1055407988 Chr.X: 112776012-112776012 on Build GRCh38

View Full Product Details