Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 155276713-155277016 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLIC2
Entrez Gene ID
1193
Gene Name
chloride intracellular channel 2
Gene Aliases
CLIC2b, MRXS32, XAP121
UniGene
Hs.655445
Gene Chromosome Location
Chr.X: 155276207-155334681 on Build GRCh38
Transcripts
NM_001289.5

SNP Detail

SNP ID Chromosome Location
rs967126353 Chr.X: 155276943-155276943 on Build GRCh38
rs782489111 Chr.X: 155276833-155276833 on Build GRCh38
rs145583465 Chr.X: 155276976-155276976 on Build GRCh38
rs781862120 Chr.X: 155276774-155276774 on Build GRCh38
rs782683622 Chr.X: 155276822-155276822 on Build GRCh38
rs782448069 Chr.X: 155276956-155276956 on Build GRCh38
rs1027717374 Chr.X: 155276809-155276809 on Build GRCh38
rs782282895 Chr.X: 155276966-155276966 on Build GRCh38
rs150105307 Chr.X: 155276768-155276768 on Build GRCh38
rs925674575 Chr.X: 155276988-155276988 on Build GRCh38
rs782451717 Chr.X: 155276763-155276763 on Build GRCh38
rs1018612073 Chr.X: 155276846-155276846 on Build GRCh38
rs782148480 Chr.X: 155276756-155276756 on Build GRCh38
rs782731075 Chr.X: 155276882-155276882 on Build GRCh38
rs1029320072 Chr.X: 155276908-155276908 on Build GRCh38
rs781806541 Chr.X: 155276951-155276951 on Build GRCh38
rs976859390 Chr.X: 155276895-155276895 on Build GRCh38

View Full Product Details