Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 15960089-15960688 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGFBP2
Entrez Gene ID
83888
Gene Name
fibroblast growth factor binding protein 2
Gene Aliases
HBP17RP, KSP37
UniGene
Hs.98785
Gene Chromosome Location
Chr.4: 15960240-15963236 on Build GRCh38
Transcripts
NM_031950.3

SNP Detail

SNP ID Chromosome Location
rs997202338 Chr.4: 15960219-15960219 on Build GRCh38
rs947864569 Chr.4: 15960316-15960316 on Build GRCh38
rs113523111 Chr.4: 15960302-15960302 on Build GRCh38
rs888788040 Chr.4: 15960438-15960439 on Build GRCh38
rs186182647 Chr.4: 15960464-15960464 on Build GRCh38
rs920300956 Chr.4: 15960528-15960528 on Build GRCh38
rs936958446 Chr.4: 15960147-15960147 on Build GRCh38
rs973101541 Chr.4: 15960301-15960301 on Build GRCh38
rs559210802 Chr.4: 15960130-15960130 on Build GRCh38
rs971957606 Chr.4: 15960515-15960515 on Build GRCh38
rs974966895 Chr.4: 15960190-15960190 on Build GRCh38
rs907513856 Chr.4: 15960281-15960281 on Build GRCh38
rs746422322 Chr.4: 15960426-15960426 on Build GRCh38
rs570032344 Chr.4: 15960625-15960625 on Build GRCh38
rs62288687 Chr.4: 15960318-15960318 on Build GRCh38
rs117535467 Chr.4: 15960346-15960346 on Build GRCh38
rs1010894794 Chr.4: 15960348-15960348 on Build GRCh38
rs1028675336 Chr.4: 15960228-15960228 on Build GRCh38
rs1041219863 Chr.4: 15960420-15960420 on Build GRCh38
rs984729826 Chr.4: 15960248-15960248 on Build GRCh38
rs945618449 Chr.4: 15960411-15960411 on Build GRCh38
rs796135073 Chr.4: 15960197-15960197 on Build GRCh38
rs370696719 Chr.4: 15960544-15960544 on Build GRCh38
rs868401060 Chr.4: 15960629-15960629 on Build GRCh38
rs1022335407 Chr.4: 15960371-15960371 on Build GRCh38
rs958146400 Chr.4: 15960627-15960627 on Build GRCh38
rs527692389 Chr.4: 15960241-15960241 on Build GRCh38
rs899905971 Chr.4: 15960384-15960384 on Build GRCh38
rs964149303 Chr.4: 15960165-15960165 on Build GRCh38
rs796362462 Chr.4: 15960589-15960592 on Build GRCh38
rs939565769 Chr.4: 15960337-15960337 on Build GRCh38
rs970520428 Chr.4: 15960615-15960629 on Build GRCh38
rs561034821 Chr.4: 15960183-15960183 on Build GRCh38
rs115402468 Chr.4: 15960436-15960436 on Build GRCh38
rs1014812781 Chr.4: 15960508-15960508 on Build GRCh38
rs1019732428 Chr.4: 15960206-15960206 on Build GRCh38
rs1054044924 Chr.4: 15960400-15960400 on Build GRCh38
rs149995621 Chr.4: 15960593-15960596 on Build GRCh38
rs549996713 Chr.4: 15960510-15960510 on Build GRCh38
rs973029320 Chr.4: 15960518-15960518 on Build GRCh38
rs143322307 Chr.4: 15960428-15960428 on Build GRCh38
rs906705448 Chr.4: 15960328-15960328 on Build GRCh38
rs536280039 Chr.4: 15960576-15960576 on Build GRCh38
rs901628963 Chr.4: 15960208-15960208 on Build GRCh38
rs189469513 Chr.4: 15960640-15960640 on Build GRCh38
rs181407442 Chr.4: 15960647-15960647 on Build GRCh38
rs953131249 Chr.4: 15960233-15960233 on Build GRCh38
rs892643071 Chr.4: 15960151-15960151 on Build GRCh38
rs372840467 Chr.4: 15960511-15960511 on Build GRCh38
rs189427472 Chr.4: 15960291-15960291 on Build GRCh38
rs980888387 Chr.4: 15960617-15960617 on Build GRCh38
rs367616842 Chr.4: 15960325-15960325 on Build GRCh38
rs901324982 Chr.4: 15960423-15960423 on Build GRCh38
rs980554785 Chr.4: 15960565-15960565 on Build GRCh38
rs565108528 Chr.4: 15960150-15960150 on Build GRCh38
rs28544486 Chr.4: 15960251-15960251 on Build GRCh38
rs572631509 Chr.4: 15960140-15960140 on Build GRCh38
rs1010969816 Chr.4: 15960122-15960122 on Build GRCh38
rs987407269 Chr.4: 15960263-15960263 on Build GRCh38
rs961955579 Chr.4: 15960259-15960259 on Build GRCh38
rs989916903 Chr.4: 15960630-15960630 on Build GRCh38
rs928094861 Chr.4: 15960639-15960639 on Build GRCh38
rs74733021 Chr.4: 15960342-15960342 on Build GRCh38
rs961986883 Chr.4: 15960492-15960492 on Build GRCh38
rs756055565 Chr.4: 15960614-15960614 on Build GRCh38
rs181073650 Chr.4: 15960402-15960402 on Build GRCh38
rs996978950 Chr.4: 15960390-15960390 on Build GRCh38
rs550172522 Chr.4: 15960600-15960600 on Build GRCh38
rs539350625 Chr.4: 15960628-15960628 on Build GRCh38

View Full Product Details