Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 68975175-68975745 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EOGT
Entrez Gene ID
285203
Gene Name
EGF domain specific O-linked N-acetylglucosamine transferase
Gene Aliases
AER61, AOS4, C3orf64, EOGT1
UniGene
Hs.518059
Gene Chromosome Location
Chr.3: 68975212-69013961 on Build GRCh38
Transcripts
NM_001278689.1 NM_173654.2 NR_103826.1 XM_005264743.2 XM_011533599.1 XM_011533600.1 XM_011533602.1 XM_011533603.1 XM_017006204.1 XM_017006205.1 XM_017006206.1 XM_017006207.1 XM_017006209.1

SNP Detail

SNP ID Chromosome Location
rs867002211 Chr.3: 68975506-68975506 on Build GRCh38
rs538122260 Chr.3: 68975695-68975695 on Build GRCh38
rs754097448 Chr.3: 68975701-68975703 on Build GRCh38
rs1009060604 Chr.3: 68975354-68975355 on Build GRCh38
rs992620193 Chr.3: 68975484-68975484 on Build GRCh38
rs776244277 Chr.3: 68975445-68975445 on Build GRCh38
rs910169983 Chr.3: 68975548-68975548 on Build GRCh38
rs56388443 Chr.3: 68975714-68975714 on Build GRCh38
rs926666146 Chr.3: 68975273-68975273 on Build GRCh38
rs945893164 Chr.3: 68975302-68975305 on Build GRCh38
rs181222895 Chr.3: 68975349-68975349 on Build GRCh38
rs397706963 Chr.3: 68975542-68975543 on Build GRCh38
rs763882124 Chr.3: 68975353-68975353 on Build GRCh38
rs935913166 Chr.3: 68975529-68975529 on Build GRCh38
rs867184167 Chr.3: 68975269-68975269 on Build GRCh38
rs897590927 Chr.3: 68975579-68975579 on Build GRCh38
rs113721595 Chr.3: 68975591-68975591 on Build GRCh38
rs929062033 Chr.3: 68975580-68975580 on Build GRCh38
rs375834875 Chr.3: 68975499-68975499 on Build GRCh38
rs753790058 Chr.3: 68975450-68975450 on Build GRCh38
rs935945252 Chr.3: 68975332-68975332 on Build GRCh38
rs57738895 Chr.3: 68975536-68975537 on Build GRCh38
rs1021053321 Chr.3: 68975488-68975488 on Build GRCh38
rs765031722 Chr.3: 68975501-68975501 on Build GRCh38
rs189183725 Chr.3: 68975313-68975313 on Build GRCh38
rs569209217 Chr.3: 68975679-68975679 on Build GRCh38
rs386396966 Chr.3: 68975537-68975538 on Build GRCh38
rs185044152 Chr.3: 68975577-68975577 on Build GRCh38
rs992298137 Chr.3: 68975297-68975297 on Build GRCh38
rs1032829260 Chr.3: 68975219-68975219 on Build GRCh38
rs372606238 Chr.3: 68975440-68975440 on Build GRCh38
rs993677892 Chr.3: 68975387-68975387 on Build GRCh38
rs544957565 Chr.3: 68975335-68975335 on Build GRCh38
rs56239520 Chr.3: 68975278-68975278 on Build GRCh38
rs376163851 Chr.3: 68975365-68975365 on Build GRCh38
rs961060747 Chr.3: 68975287-68975287 on Build GRCh38
rs550918461 Chr.3: 68975405-68975405 on Build GRCh38
rs570610266 Chr.3: 68975239-68975244 on Build GRCh38
rs967162217 Chr.3: 68975489-68975489 on Build GRCh38
rs546010428 Chr.3: 68975238-68975238 on Build GRCh38
rs369452310 Chr.3: 68975434-68975434 on Build GRCh38
rs952254603 Chr.3: 68975420-68975420 on Build GRCh38
rs761434479 Chr.3: 68975446-68975446 on Build GRCh38
rs765386517 Chr.3: 68975543-68975543 on Build GRCh38
rs774633965 Chr.3: 68975334-68975334 on Build GRCh38
rs1049243377 Chr.3: 68975584-68975584 on Build GRCh38
rs760980430 Chr.3: 68975543-68975544 on Build GRCh38
rs1021397936 Chr.3: 68975707-68975707 on Build GRCh38
rs546683920 Chr.3: 68975570-68975570 on Build GRCh38
rs896451128 Chr.3: 68975670-68975670 on Build GRCh38
rs1033945500 Chr.3: 68975627-68975627 on Build GRCh38
rs897953826 Chr.3: 68975383-68975383 on Build GRCh38
rs117715435 Chr.3: 68975621-68975621 on Build GRCh38

View Full Product Details