Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 73837072-73837490 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXCL6
Entrez Gene ID
6372
Gene Name
C-X-C motif chemokine ligand 6
Gene Aliases
CKA-3, GCP-2, GCP2, SCYB6
UniGene
Hs.164021
Gene Chromosome Location
Chr.4: 73836556-73838760 on Build GRCh38
Transcripts
NM_002993.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3826187 c.233T>G p.V78G Chr.4: 73837087-73837087 on Build GRCh38
448121 c.281C>T p.P94L Chr.4: 73837241-73837241 on Build GRCh38
3605760 c.314A>T p.K105I Chr.4: 73837274-73837274 on Build GRCh38
6100981 c.271T>G p.C91G Chr.4: 73837231-73837231 on Build GRCh38
3605759 c.280C>T p.P94S Chr.4: 73837240-73837240 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs975429889 Chr.4: 73837241-73837241 on Build GRCh38
rs1043572855 Chr.4: 73837308-73837308 on Build GRCh38
rs776726060 Chr.4: 73837148-73837148 on Build GRCh38
rs750344173 Chr.4: 73837099-73837099 on Build GRCh38
rs576371584 Chr.4: 73837446-73837446 on Build GRCh38
rs191588763 Chr.4: 73837153-73837153 on Build GRCh38
rs766602848 Chr.4: 73837378-73837378 on Build GRCh38
rs200271908 Chr.4: 73837292-73837292 on Build GRCh38
rs756875879 Chr.4: 73837093-73837093 on Build GRCh38
rs778008085 Chr.4: 73837122-73837122 on Build GRCh38
rs945534112 Chr.4: 73837155-73837155 on Build GRCh38
rs753633605 Chr.4: 73837216-73837216 on Build GRCh38
rs770332212 Chr.4: 73837136-73837136 on Build GRCh38
rs768861968 Chr.4: 73837270-73837270 on Build GRCh38
rs543482006 Chr.4: 73837455-73837455 on Build GRCh38
rs1004399887 Chr.4: 73837419-73837419 on Build GRCh38
rs749091972 Chr.4: 73837116-73837116 on Build GRCh38
rs767067704 Chr.4: 73837181-73837181 on Build GRCh38
rs772842842 Chr.4: 73837295-73837295 on Build GRCh38
rs904941373 Chr.4: 73837370-73837370 on Build GRCh38
rs767157662 Chr.4: 73837098-73837098 on Build GRCh38
rs749964064 Chr.4: 73837200-73837200 on Build GRCh38
rs4417936 Chr.4: 73837134-73837134 on Build GRCh38
rs965683481 Chr.4: 73837453-73837453 on Build GRCh38
rs41462544 Chr.4: 73837386-73837386 on Build GRCh38
rs932941033 Chr.4: 73837172-73837172 on Build GRCh38
rs771906311 Chr.4: 73837293-73837293 on Build GRCh38
rs564244632 Chr.4: 73837092-73837093 on Build GRCh38
rs781192657 Chr.4: 73837246-73837246 on Build GRCh38
rs372015132 Chr.4: 73837248-73837248 on Build GRCh38
rs139087582 Chr.4: 73837164-73837164 on Build GRCh38
rs935186545 Chr.4: 73837313-73837313 on Build GRCh38
rs770330032 Chr.4: 73837258-73837258 on Build GRCh38
rs377169766 Chr.4: 73837285-73837285 on Build GRCh38
rs1001466738 Chr.4: 73837358-73837358 on Build GRCh38
rs1049693115 Chr.4: 73837212-73837212 on Build GRCh38
rs372953502 Chr.4: 73837239-73837239 on Build GRCh38
rs1013184396 Chr.4: 73837331-73837331 on Build GRCh38
rs775911783 Chr.4: 73837161-73837161 on Build GRCh38
rs761587299 Chr.4: 73837177-73837177 on Build GRCh38
rs201102235 Chr.4: 73837178-73837178 on Build GRCh38
rs757458903 Chr.4: 73837339-73837339 on Build GRCh38
rs201300540 Chr.4: 73837159-73837159 on Build GRCh38
rs368465900 Chr.4: 73837176-73837176 on Build GRCh38
rs896421963 Chr.4: 73837324-73837324 on Build GRCh38
rs770954191 Chr.4: 73837147-73837147 on Build GRCh38
rs925068282 Chr.4: 73837311-73837311 on Build GRCh38
rs779837611 Chr.4: 73837114-73837114 on Build GRCh38
rs766314228 Chr.4: 73837319-73837319 on Build GRCh38
rs753672610 Chr.4: 73837323-73837323 on Build GRCh38
rs146538128 Chr.4: 73837263-73837263 on Build GRCh38
rs759553133 Chr.4: 73837327-73837327 on Build GRCh38
rs564291905 Chr.4: 73837408-73837408 on Build GRCh38
rs537103202 Chr.4: 73837354-73837354 on Build GRCh38
rs780367243 Chr.4: 73837283-73837284 on Build GRCh38
rs983401379 Chr.4: 73837180-73837180 on Build GRCh38
rs914034603 Chr.4: 73837144-73837144 on Build GRCh38
rs111731742 Chr.4: 73837466-73837466 on Build GRCh38
rs370243514 Chr.4: 73837107-73837107 on Build GRCh38
rs763099546 Chr.4: 73837228-73837228 on Build GRCh38
rs763772236 Chr.4: 73837166-73837166 on Build GRCh38
rs951599305 Chr.4: 73837418-73837418 on Build GRCh38
rs140549348 Chr.4: 73837234-73837234 on Build GRCh38
rs760139934 Chr.4: 73837298-73837298 on Build GRCh38
rs995618006 Chr.4: 73837123-73837123 on Build GRCh38
rs766290365 Chr.4: 73837205-73837205 on Build GRCh38
rs543056372 Chr.4: 73837330-73837330 on Build GRCh38
rs760645932 Chr.4: 73837201-73837201 on Build GRCh38
rs959697397 Chr.4: 73837377-73837377 on Build GRCh38
rs572163238 Chr.4: 73837366-73837366 on Build GRCh38
rs552755043 Chr.4: 73837284-73837284 on Build GRCh38
rs750712335 Chr.4: 73837097-73837098 on Build GRCh38
rs774096701 Chr.4: 73837168-73837168 on Build GRCh38
rs912964650 Chr.4: 73837414-73837414 on Build GRCh38
rs758637436 Chr.4: 73837117-73837117 on Build GRCh38
rs369562400 Chr.4: 73837328-73837328 on Build GRCh38
rs758718337 Chr.4: 73837210-73837233 on Build GRCh38
rs145625563 Chr.4: 73837242-73837242 on Build GRCh38
rs749823854 Chr.4: 73837265-73837265 on Build GRCh38
rs781461819 Chr.4: 73837340-73837340 on Build GRCh38
rs921332526 Chr.4: 73837256-73837256 on Build GRCh38

View Full Product Details