Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 75578957-75579401 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDKL2
Entrez Gene ID
8999
Gene Name
cyclin dependent kinase like 2
Gene Aliases
KKIAMRE, P56
UniGene
Hs.591698
Gene Chromosome Location
Chr.4: 75576494-75630537 on Build GRCh38
Transcripts
NM_003948.3 XM_005263335.2 XM_006714406.2 XM_017008809.1 XM_017008810.1 XM_017008811.1

SNP Detail

SNP ID Chromosome Location
rs932527030 Chr.4: 75579112-75579112 on Build GRCh38
rs976637460 Chr.4: 75579376-75579376 on Build GRCh38
rs1018443723 Chr.4: 75579309-75579309 on Build GRCh38
rs752891263 Chr.4: 75579250-75579250 on Build GRCh38
rs935652810 Chr.4: 75579138-75579138 on Build GRCh38
rs1015537610 Chr.4: 75579339-75579339 on Build GRCh38
rs1055469016 Chr.4: 75579011-75579011 on Build GRCh38
rs188397356 Chr.4: 75579264-75579264 on Build GRCh38
rs891490773 Chr.4: 75579164-75579164 on Build GRCh38
rs774940262 Chr.4: 75579092-75579092 on Build GRCh38
rs547292692 Chr.4: 75579126-75579126 on Build GRCh38
rs759850404 Chr.4: 75579100-75579100 on Build GRCh38
rs377061373 Chr.4: 75579109-75579109 on Build GRCh38
rs895858834 Chr.4: 75579203-75579203 on Build GRCh38
rs766738476 Chr.4: 75579036-75579036 on Build GRCh38
rs764385317 Chr.4: 75579355-75579355 on Build GRCh38
rs866483087 Chr.4: 75579006-75579006 on Build GRCh38
rs906696400 Chr.4: 75579204-75579204 on Build GRCh38
rs1009809392 Chr.4: 75579170-75579170 on Build GRCh38
rs1029538320 Chr.4: 75579378-75579378 on Build GRCh38
rs191907319 Chr.4: 75579098-75579098 on Build GRCh38
rs1049453002 Chr.4: 75579119-75579119 on Build GRCh38
rs976668696 Chr.4: 75579043-75579043 on Build GRCh38
rs796182882 Chr.4: 75579185-75579185 on Build GRCh38
rs767905863 Chr.4: 75579207-75579207 on Build GRCh38
rs1046199562 Chr.4: 75579224-75579224 on Build GRCh38
rs541988349 Chr.4: 75579380-75579380 on Build GRCh38
rs1006477543 Chr.4: 75579299-75579299 on Build GRCh38
rs984359959 Chr.4: 75579055-75579055 on Build GRCh38
rs893506498 Chr.4: 75579282-75579282 on Build GRCh38
rs1046714185 Chr.4: 75579174-75579174 on Build GRCh38
rs762528140 Chr.4: 75579035-75579035 on Build GRCh38
rs184109059 Chr.4: 75579156-75579156 on Build GRCh38
rs180990754 Chr.4: 75579265-75579265 on Build GRCh38
rs565740545 Chr.4: 75579233-75579233 on Build GRCh38
rs185817309 Chr.4: 75579294-75579294 on Build GRCh38
rs902330381 Chr.4: 75579243-75579243 on Build GRCh38

View Full Product Details