Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 122895247-122895788 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF2
Entrez Gene ID
2247
Gene Name
fibroblast growth factor 2
Gene Aliases
BFGF, FGF-2, FGFB, HBGF-2
UniGene
Hs.284244
Gene Chromosome Location
Chr.4: 122826708-122898235 on Build GRCh38
Transcripts
NM_002006.4 NM_007083.4 NM_198041.2

Gene Symbol
NUDT6
Entrez Gene ID
11162
Gene Name
nudix hydrolase 6
Gene Aliases
ASFGF2, FGF-AS, FGF2AS, GFG-1, GFG1
UniGene
Hs.558459
Gene Chromosome Location
Chr.4: 122892644-122923004 on Build GRCh38
Transcripts
NM_002006.4 NM_007083.4 NM_198041.2

SNP Detail

SNP ID Chromosome Location
rs993793559 Chr.4: 122895660-122895660 on Build GRCh38
rs908473028 Chr.4: 122895597-122895597 on Build GRCh38
rs779852076 Chr.4: 122895655-122895655 on Build GRCh38
rs1026299721 Chr.4: 122895696-122895696 on Build GRCh38
rs77571189 Chr.4: 122895483-122895483 on Build GRCh38
rs45495095 Chr.4: 122895638-122895638 on Build GRCh38
rs1003296875 Chr.4: 122895750-122895750 on Build GRCh38
rs764883431 Chr.4: 122895325-122895325 on Build GRCh38
rs939812850 Chr.4: 122895632-122895632 on Build GRCh38
rs182205477 Chr.4: 122895345-122895345 on Build GRCh38
rs868378144 Chr.4: 122895460-122895460 on Build GRCh38
rs45546532 Chr.4: 122895738-122895740 on Build GRCh38
rs975604868 Chr.4: 122895580-122895580 on Build GRCh38
rs6854081 Chr.4: 122895552-122895552 on Build GRCh38
rs529589754 Chr.4: 122895331-122895333 on Build GRCh38
rs113615485 Chr.4: 122895433-122895433 on Build GRCh38
rs1044876755 Chr.4: 122895727-122895727 on Build GRCh38
rs950013055 Chr.4: 122895499-122895499 on Build GRCh38
rs536775550 Chr.4: 122895524-122895524 on Build GRCh38
rs920939054 Chr.4: 122895588-122895588 on Build GRCh38
rs1003086506 Chr.4: 122895373-122895373 on Build GRCh38
rs112480130 Chr.4: 122895756-122895756 on Build GRCh38
rs958560639 Chr.4: 122895412-122895412 on Build GRCh38
rs45441398 Chr.4: 122895417-122895417 on Build GRCh38
rs1054167739 Chr.4: 122895587-122895587 on Build GRCh38
rs888874282 Chr.4: 122895619-122895619 on Build GRCh38
rs532022111 Chr.4: 122895650-122895650 on Build GRCh38
rs758225723 Chr.4: 122895594-122895594 on Build GRCh38
rs1033728974 Chr.4: 122895754-122895754 on Build GRCh38
rs552192823 Chr.4: 122895751-122895751 on Build GRCh38
rs1022957677 Chr.4: 122895340-122895340 on Build GRCh38
rs377638739 Chr.4: 122895559-122895559 on Build GRCh38
rs914763970 Chr.4: 122895498-122895498 on Build GRCh38
rs1050991146 Chr.4: 122895593-122895593 on Build GRCh38
rs887816963 Chr.4: 122895698-122895698 on Build GRCh38
rs371110764 Chr.4: 122895578-122895578 on Build GRCh38
rs1041765819 Chr.4: 122895546-122895546 on Build GRCh38
rs397897556 Chr.4: 122895741-122895743 on Build GRCh38
rs956177693 Chr.4: 122895461-122895461 on Build GRCh38
rs991343985 Chr.4: 122895477-122895477 on Build GRCh38
rs1006301133 Chr.4: 122895722-122895722 on Build GRCh38
rs187737646 Chr.4: 122895538-122895538 on Build GRCh38
rs935750934 Chr.4: 122895557-122895557 on Build GRCh38
rs1013447164 Chr.4: 122895271-122895271 on Build GRCh38
rs933655099 Chr.4: 122895591-122895591 on Build GRCh38
rs991705966 Chr.4: 122895759-122895759 on Build GRCh38

View Full Product Details