Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 24498077-24498610 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH10
Entrez Gene ID
1008
Gene Name
cadherin 10
Gene Aliases
-
UniGene
Hs.92489
Gene Chromosome Location
Chr.5: 24487100-24644978 on Build GRCh38
Transcripts
NM_001317222.1 NM_001317224.1 NM_006727.4 XM_011513923.2 XM_017008915.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4799706 c.1397A>C p.N466T Chr.5: 24498516-24498516 on Build GRCh38
127589 c.1503C>A p.A501A Chr.5: 24498410-24498410 on Build GRCh38
6297522 c.1416C>A p.R472R Chr.5: 24498497-24498497 on Build GRCh38
183728 c.1468G>T p.A490S Chr.5: 24498445-24498445 on Build GRCh38
6493611 c.1400C>A p.P467H Chr.5: 24498513-24498513 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746518413 Chr.5: 24498221-24498221 on Build GRCh38
rs545836910 Chr.5: 24498234-24498234 on Build GRCh38
rs756646914 Chr.5: 24498359-24498359 on Build GRCh38
rs368343446 Chr.5: 24498529-24498529 on Build GRCh38
rs769547516 Chr.5: 24498456-24498456 on Build GRCh38
rs370513099 Chr.5: 24498371-24498371 on Build GRCh38
rs944626311 Chr.5: 24498290-24498290 on Build GRCh38
rs937254910 Chr.5: 24498258-24498258 on Build GRCh38
rs758481430 Chr.5: 24498386-24498386 on Build GRCh38
rs1054728236 Chr.5: 24498268-24498268 on Build GRCh38
rs371232103 Chr.5: 24498362-24498362 on Build GRCh38
rs3815909 Chr.5: 24498581-24498581 on Build GRCh38
rs147882578 Chr.5: 24498458-24498458 on Build GRCh38
rs199776417 Chr.5: 24498496-24498496 on Build GRCh38
rs201398396 Chr.5: 24498542-24498542 on Build GRCh38
rs200724704 Chr.5: 24498372-24498372 on Build GRCh38
rs568094245 Chr.5: 24498153-24498153 on Build GRCh38
rs200673315 Chr.5: 24498519-24498519 on Build GRCh38
rs909369273 Chr.5: 24498124-24498124 on Build GRCh38
rs145457868 Chr.5: 24498504-24498504 on Build GRCh38
rs543755225 Chr.5: 24498509-24498509 on Build GRCh38
rs74795908 Chr.5: 24498484-24498484 on Build GRCh38
rs1010748136 Chr.5: 24498337-24498337 on Build GRCh38
rs535081979 Chr.5: 24498167-24498167 on Build GRCh38
rs776564516 Chr.5: 24498417-24498417 on Build GRCh38
rs139927244 Chr.5: 24498497-24498497 on Build GRCh38
rs544297913 Chr.5: 24498498-24498498 on Build GRCh38
rs894203122 Chr.5: 24498369-24498369 on Build GRCh38
rs754193703 Chr.5: 24498370-24498370 on Build GRCh38
rs775242331 Chr.5: 24498574-24498574 on Build GRCh38
rs920803400 Chr.5: 24498540-24498540 on Build GRCh38
rs3815910 Chr.5: 24498561-24498561 on Build GRCh38
rs755109257 Chr.5: 24498515-24498515 on Build GRCh38
rs771174296 Chr.5: 24498229-24498229 on Build GRCh38
rs892945215 Chr.5: 24498291-24498291 on Build GRCh38
rs188047281 Chr.5: 24498564-24498564 on Build GRCh38
rs182395109 Chr.5: 24498433-24498433 on Build GRCh38
rs1050374347 Chr.5: 24498194-24498194 on Build GRCh38
rs77909063 Chr.5: 24498473-24498473 on Build GRCh38
rs747125379 Chr.5: 24498405-24498405 on Build GRCh38
rs949689321 Chr.5: 24498177-24498177 on Build GRCh38
rs750821300 Chr.5: 24498501-24498501 on Build GRCh38
rs372945121 Chr.5: 24498188-24498188 on Build GRCh38
rs972902858 Chr.5: 24498567-24498567 on Build GRCh38
rs561943909 Chr.5: 24498449-24498449 on Build GRCh38
rs753275798 Chr.5: 24498112-24498112 on Build GRCh38
rs530911338 Chr.5: 24498265-24498265 on Build GRCh38
rs16893442 Chr.5: 24498506-24498506 on Build GRCh38
rs759109710 Chr.5: 24498562-24498562 on Build GRCh38
rs1041618952 Chr.5: 24498326-24498326 on Build GRCh38
rs746115406 Chr.5: 24498373-24498373 on Build GRCh38
rs561788034 Chr.5: 24498240-24498240 on Build GRCh38
rs367790263 Chr.5: 24498355-24498355 on Build GRCh38
rs749111178 Chr.5: 24498557-24498557 on Build GRCh38
rs762565754 Chr.5: 24498490-24498490 on Build GRCh38
rs557052935 Chr.5: 24498186-24498186 on Build GRCh38
rs971053745 Chr.5: 24498466-24498466 on Build GRCh38
rs79743241 Chr.5: 24498489-24498489 on Build GRCh38
rs541806280 Chr.5: 24498133-24498133 on Build GRCh38
rs777217792 Chr.5: 24498147-24498147 on Build GRCh38
rs747033725 Chr.5: 24498512-24498512 on Build GRCh38
rs544303560 Chr.5: 24498411-24498411 on Build GRCh38
rs766154972 Chr.5: 24498491-24498491 on Build GRCh38
rs780057560 Chr.5: 24498401-24498401 on Build GRCh38
rs764699774 Chr.5: 24498367-24498367 on Build GRCh38
rs201738748 Chr.5: 24498499-24498499 on Build GRCh38
rs997556314 Chr.5: 24498470-24498470 on Build GRCh38
rs777385366 Chr.5: 24498547-24498547 on Build GRCh38
rs575266098 Chr.5: 24498210-24498210 on Build GRCh38
rs555629107 Chr.5: 24498110-24498110 on Build GRCh38

View Full Product Details