Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 14693638-14694223 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OTULIN
Entrez Gene ID
90268
Gene Name
OTU deubiquitinase with linear linkage specificity
Gene Aliases
FAM105B, GUM
UniGene
Hs.406335
Gene Chromosome Location
Chr.5: 14660794-14716552 on Build GRCh38
Transcripts
NM_138348.4 XM_011514151.2 XM_011514154.2 XM_017010015.1

SNP Detail

SNP ID Chromosome Location
rs919578881 Chr.5: 14693981-14693981 on Build GRCh38
rs957639919 Chr.5: 14693921-14693921 on Build GRCh38
rs937027429 Chr.5: 14693773-14693773 on Build GRCh38
rs996210904 Chr.5: 14693863-14693863 on Build GRCh38
rs369873401 Chr.5: 14694103-14694103 on Build GRCh38
rs191995664 Chr.5: 14693765-14693765 on Build GRCh38
rs138425828 Chr.5: 14693802-14693802 on Build GRCh38
rs1010204069 Chr.5: 14694168-14694168 on Build GRCh38
rs916328575 Chr.5: 14693994-14693994 on Build GRCh38
rs184701011 Chr.5: 14693932-14693932 on Build GRCh38
rs978021957 Chr.5: 14693930-14693930 on Build GRCh38
rs184786399 Chr.5: 14693667-14693667 on Build GRCh38
rs996652449 Chr.5: 14694054-14694054 on Build GRCh38
rs1008975247 Chr.5: 14693851-14693851 on Build GRCh38
rs927074391 Chr.5: 14693950-14693950 on Build GRCh38
rs765523394 Chr.5: 14693734-14693734 on Build GRCh38
rs189466699 Chr.5: 14694115-14694115 on Build GRCh38
rs1020553779 Chr.5: 14694177-14694177 on Build GRCh38
rs1038062317 Chr.5: 14693957-14693957 on Build GRCh38
rs1008029448 Chr.5: 14694037-14694037 on Build GRCh38
rs539185420 Chr.5: 14693964-14693964 on Build GRCh38
rs115766363 Chr.5: 14694010-14694010 on Build GRCh38
rs143812069 Chr.5: 14693865-14693865 on Build GRCh38
rs1049193086 Chr.5: 14694016-14694016 on Build GRCh38
rs996190566 Chr.5: 14693704-14693704 on Build GRCh38
rs996117008 Chr.5: 14694153-14694153 on Build GRCh38
rs940925100 Chr.5: 14694051-14694051 on Build GRCh38
rs966578269 Chr.5: 14693907-14693907 on Build GRCh38
rs551462928 Chr.5: 14693691-14693691 on Build GRCh38
rs989325386 Chr.5: 14693841-14693841 on Build GRCh38
rs189585611 Chr.5: 14693737-14693737 on Build GRCh38
rs147059703 Chr.5: 14693701-14693701 on Build GRCh38
rs571272616 Chr.5: 14693876-14693876 on Build GRCh38
rs1019038102 Chr.5: 14693852-14693852 on Build GRCh38
rs16903656 Chr.5: 14693956-14693956 on Build GRCh38
rs758887008 Chr.5: 14693869-14693869 on Build GRCh38
rs981982344 Chr.5: 14693949-14693949 on Build GRCh38
rs376815475 Chr.5: 14693713-14693713 on Build GRCh38
rs467136 Chr.5: 14694117-14694117 on Build GRCh38
rs1050896226 Chr.5: 14694187-14694187 on Build GRCh38
rs779608086 Chr.5: 14693670-14693673 on Build GRCh38
rs965128020 Chr.5: 14693861-14693861 on Build GRCh38
rs536473911 Chr.5: 14694159-14694159 on Build GRCh38
rs994026561 Chr.5: 14693814-14693814 on Build GRCh38
rs781201171 Chr.5: 14694044-14694044 on Build GRCh38
rs140575719 Chr.5: 14693672-14693672 on Build GRCh38
rs971561349 Chr.5: 14693947-14693947 on Build GRCh38
rs971608938 Chr.5: 14694186-14694186 on Build GRCh38
rs1025459619 Chr.5: 14693838-14693838 on Build GRCh38
rs1045507514 Chr.5: 14693743-14693743 on Build GRCh38
rs949817614 Chr.5: 14693736-14693736 on Build GRCh38
rs1040619986 Chr.5: 14694106-14694106 on Build GRCh38
rs891003205 Chr.5: 14694026-14694026 on Build GRCh38
rs929928242 Chr.5: 14693984-14693984 on Build GRCh38
rs1038414142 Chr.5: 14693782-14693782 on Build GRCh38
rs751678650 Chr.5: 14693935-14693935 on Build GRCh38
rs890951200 Chr.5: 14693843-14693844 on Build GRCh38

View Full Product Details