Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 10280292-10280775 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMBL
Entrez Gene ID
134147
Gene Name
carboxymethylenebutenolidase homolog
Gene Aliases
JS-1
UniGene
Hs.192586
Gene Chromosome Location
Chr.5: 10277595-10308056 on Build GRCh38
Transcripts
NM_138809.3 XM_017009034.1 XM_017009035.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4672993 c.689T>C p.I230T Chr.5: 10280502-10280502 on Build GRCh38
1059314 c.650G>T p.R217L Chr.5: 10280541-10280541 on Build GRCh38
3428803 c.616A>C p.T206P Chr.5: 10280575-10280575 on Build GRCh38
4555530 c.658G>A p.E220K Chr.5: 10280533-10280533 on Build GRCh38
4637121 c.664T>C p.C222R Chr.5: 10280527-10280527 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs569668328 Chr.5: 10280438-10280438 on Build GRCh38
rs1055078752 Chr.5: 10280678-10280678 on Build GRCh38
rs778186080 Chr.5: 10280505-10280505 on Build GRCh38
rs368492170 Chr.5: 10280428-10280428 on Build GRCh38
rs776109531 Chr.5: 10280675-10280675 on Build GRCh38
rs1004491389 Chr.5: 10280335-10280335 on Build GRCh38
rs372248081 Chr.5: 10280324-10280324 on Build GRCh38
rs529757867 Chr.5: 10280361-10280361 on Build GRCh38
rs756299218 Chr.5: 10280587-10280587 on Build GRCh38
rs887906255 Chr.5: 10280457-10280457 on Build GRCh38
rs761470815 Chr.5: 10280677-10280677 on Build GRCh38
rs758332620 Chr.5: 10280383-10280383 on Build GRCh38
rs367637642 Chr.5: 10280468-10280468 on Build GRCh38
rs995439229 Chr.5: 10280662-10280662 on Build GRCh38
rs116704059 Chr.5: 10280352-10280352 on Build GRCh38
rs371454569 Chr.5: 10280484-10280484 on Build GRCh38
rs1006766272 Chr.5: 10280580-10280580 on Build GRCh38
rs906148660 Chr.5: 10280476-10280476 on Build GRCh38
rs369632245 Chr.5: 10280411-10280411 on Build GRCh38
rs1010283007 Chr.5: 10280563-10280563 on Build GRCh38
rs770915320 Chr.5: 10280513-10280513 on Build GRCh38
rs779980363 Chr.5: 10280396-10280396 on Build GRCh38
rs556283349 Chr.5: 10280611-10280611 on Build GRCh38
rs766250214 Chr.5: 10280412-10280412 on Build GRCh38
rs995134552 Chr.5: 10280381-10280381 on Build GRCh38
rs751973286 Chr.5: 10280684-10280684 on Build GRCh38
rs144737518 Chr.5: 10280630-10280630 on Build GRCh38
rs759654586 Chr.5: 10280570-10280570 on Build GRCh38
rs753529555 Chr.5: 10280542-10280542 on Build GRCh38
rs775097808 Chr.5: 10280527-10280527 on Build GRCh38
rs775804472 Chr.5: 10280479-10280480 on Build GRCh38
rs865870132 Chr.5: 10280605-10280605 on Build GRCh38
rs144479678 Chr.5: 10280503-10280503 on Build GRCh38
rs1021727547 Chr.5: 10280565-10280565 on Build GRCh38
rs374429023 Chr.5: 10280724-10280724 on Build GRCh38
rs759530277 Chr.5: 10280521-10280521 on Build GRCh38
rs959083153 Chr.5: 10280386-10280386 on Build GRCh38
rs200019273 Chr.5: 10280522-10280522 on Build GRCh38
rs759387749 Chr.5: 10280423-10280423 on Build GRCh38
rs545295782 Chr.5: 10280703-10280703 on Build GRCh38
rs749578896 Chr.5: 10280599-10280599 on Build GRCh38
rs760132238 Chr.5: 10280531-10280531 on Build GRCh38
rs147855838 Chr.5: 10280541-10280541 on Build GRCh38
rs768392633 Chr.5: 10280483-10280483 on Build GRCh38
rs777992764 Chr.5: 10280598-10280598 on Build GRCh38
rs755516670 Chr.5: 10280579-10280579 on Build GRCh38
rs747092109 Chr.5: 10280626-10280627 on Build GRCh38
rs187492563 Chr.5: 10280498-10280498 on Build GRCh38
rs774382743 Chr.5: 10280417-10280417 on Build GRCh38
rs753231011 Chr.5: 10280581-10280581 on Build GRCh38
rs764732998 Chr.5: 10280680-10280680 on Build GRCh38
rs190987855 Chr.5: 10280429-10280429 on Build GRCh38
rs1024720030 Chr.5: 10280710-10280710 on Build GRCh38
rs746211990 Chr.5: 10280425-10280425 on Build GRCh38
rs1013708962 Chr.5: 10280708-10280708 on Build GRCh38
rs141200680 Chr.5: 10280478-10280478 on Build GRCh38
rs749486864 Chr.5: 10280510-10280510 on Build GRCh38
rs868330032 Chr.5: 10280497-10280497 on Build GRCh38
rs182773279 Chr.5: 10280458-10280458 on Build GRCh38
rs772213805 Chr.5: 10280472-10280475 on Build GRCh38
rs374924358 Chr.5: 10280485-10280485 on Build GRCh38
rs755769157 Chr.5: 10280447-10280447 on Build GRCh38
rs567772271 Chr.5: 10280523-10280523 on Build GRCh38
rs371057863 Chr.5: 10280515-10280515 on Build GRCh38
rs977087806 Chr.5: 10280638-10280638 on Build GRCh38
rs751816240 Chr.5: 10280578-10280578 on Build GRCh38
rs775339636 Chr.5: 10280636-10280636 on Build GRCh38
rs550464971 Chr.5: 10280519-10280519 on Build GRCh38
rs115081846 Chr.5: 10280327-10280327 on Build GRCh38
rs149368002 Chr.5: 10280628-10280628 on Build GRCh38
rs761454458 Chr.5: 10280549-10280549 on Build GRCh38
rs780254939 Chr.5: 10280480-10280480 on Build GRCh38
rs202229219 Chr.5: 10280614-10280614 on Build GRCh38
rs951120313 Chr.5: 10280707-10280707 on Build GRCh38
rs760575932 Chr.5: 10280654-10280654 on Build GRCh38
rs768226481 Chr.5: 10280663-10280663 on Build GRCh38
rs114442248 Chr.5: 10280617-10280617 on Build GRCh38

View Full Product Details