Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 142002816-142003396 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNPDA1
Entrez Gene ID
10007
Gene Name
glucosamine-6-phosphate deaminase 1
Gene Aliases
GNP1, GNPDA, GNPI, GPI, HLN
UniGene
Hs.633853
Gene Chromosome Location
Chr.5: 142000669-142013055 on Build GRCh38
Transcripts
NM_005471.4 XM_005268348.1 XM_006714747.2 XM_011537537.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4888241 c.754G>T p.V252F Chr.5: 142003103-142003103 on Build GRCh38
7134707 c.649G>A p.E217K Chr.5: 142003208-142003208 on Build GRCh38
232420 c.643G>A p.A215T Chr.5: 142003214-142003214 on Build GRCh38
7095314 c.756C>A p.V252V Chr.5: 142003101-142003101 on Build GRCh38
6672430 c.752C>G p.T251S Chr.5: 142003105-142003105 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765316813 Chr.5: 142003188-142003188 on Build GRCh38
rs901052348 Chr.5: 142002941-142002941 on Build GRCh38
rs923074717 Chr.5: 142002898-142002898 on Build GRCh38
rs755942554 Chr.5: 142003216-142003216 on Build GRCh38
rs373357292 Chr.5: 142003157-142003157 on Build GRCh38
rs759133934 Chr.5: 142003066-142003066 on Build GRCh38
rs772788027 Chr.5: 142003039-142003039 on Build GRCh38
rs771728983 Chr.5: 142003152-142003152 on Build GRCh38
rs745359428 Chr.5: 142003278-142003278 on Build GRCh38
rs776886777 Chr.5: 142003308-142003308 on Build GRCh38
rs13187511 Chr.5: 142002863-142002863 on Build GRCh38
rs201692541 Chr.5: 142003102-142003102 on Build GRCh38
rs923012726 Chr.5: 142003351-142003351 on Build GRCh38
rs369861520 Chr.5: 142003070-142003070 on Build GRCh38
rs777733328 Chr.5: 142003222-142003222 on Build GRCh38
rs763204571 Chr.5: 142002867-142002867 on Build GRCh38
rs887991555 Chr.5: 142002914-142002914 on Build GRCh38
rs1028233215 Chr.5: 142002971-142002971 on Build GRCh38
rs746717238 Chr.5: 142003295-142003295 on Build GRCh38
rs377758422 Chr.5: 142003136-142003136 on Build GRCh38
rs2232201 Chr.5: 142002911-142002911 on Build GRCh38
rs942295805 Chr.5: 142003006-142003006 on Build GRCh38
rs778430842 Chr.5: 142003275-142003275 on Build GRCh38
rs1013616538 Chr.5: 142003033-142003033 on Build GRCh38
rs910230143 Chr.5: 142002853-142002853 on Build GRCh38
rs777169711 Chr.5: 142003178-142003178 on Build GRCh38
rs770413456 Chr.5: 142003063-142003063 on Build GRCh38
rs188563599 Chr.5: 142003067-142003067 on Build GRCh38
rs749210735 Chr.5: 142003242-142003242 on Build GRCh38
rs762511564 Chr.5: 142003062-142003062 on Build GRCh38
rs529463671 Chr.5: 142002882-142002882 on Build GRCh38
rs769521437 Chr.5: 142002987-142002987 on Build GRCh38
rs539324269 Chr.5: 142003159-142003159 on Build GRCh38
rs781064204 Chr.5: 142003114-142003114 on Build GRCh38
rs758634512 Chr.5: 142003208-142003208 on Build GRCh38
rs754374950 Chr.5: 142003083-142003083 on Build GRCh38
rs766694575 Chr.5: 142003209-142003209 on Build GRCh38
rs763897133 Chr.5: 142003274-142003274 on Build GRCh38
rs768955113 Chr.5: 142003304-142003304 on Build GRCh38
rs252114 Chr.5: 142003019-142003019 on Build GRCh38
rs747969149 Chr.5: 142003118-142003118 on Build GRCh38
rs996728862 Chr.5: 142002963-142002963 on Build GRCh38
rs749233775 Chr.5: 142003137-142003137 on Build GRCh38
rs758193314 Chr.5: 142003060-142003062 on Build GRCh38
rs770426124 Chr.5: 142003145-142003145 on Build GRCh38
rs570703487 Chr.5: 142003098-142003098 on Build GRCh38
rs252115 Chr.5: 142002907-142002907 on Build GRCh38
rs373012038 Chr.5: 142003314-142003314 on Build GRCh38
rs138526991 Chr.5: 142003210-142003210 on Build GRCh38
rs758621448 Chr.5: 142002851-142002851 on Build GRCh38
rs750605488 Chr.5: 142003099-142003099 on Build GRCh38
rs757148099 Chr.5: 142003255-142003255 on Build GRCh38
rs750471467 Chr.5: 142003200-142003200 on Build GRCh38
rs867346180 Chr.5: 142003244-142003244 on Build GRCh38
rs1000845197 Chr.5: 142003017-142003017 on Build GRCh38
rs764574469 Chr.5: 142003079-142003079 on Build GRCh38
rs895119773 Chr.5: 142003022-142003022 on Build GRCh38
rs762245122 Chr.5: 142003182-142003182 on Build GRCh38
rs762824840 Chr.5: 142003151-142003151 on Build GRCh38
rs1040363701 Chr.5: 142003037-142003037 on Build GRCh38
rs777849617 Chr.5: 142003124-142003124 on Build GRCh38
rs998648002 Chr.5: 142003154-142003154 on Build GRCh38
rs770774498 Chr.5: 142003231-142003231 on Build GRCh38
rs757333494 Chr.5: 142003087-142003087 on Build GRCh38
rs756028847 Chr.5: 142003120-142003120 on Build GRCh38
rs771639397 Chr.5: 142003282-142003282 on Build GRCh38
rs370128744 Chr.5: 142003156-142003156 on Build GRCh38
rs990086889 Chr.5: 142003243-142003243 on Build GRCh38
rs747563960 Chr.5: 142002915-142002915 on Build GRCh38
rs201291517 Chr.5: 142003283-142003283 on Build GRCh38
rs761115700 Chr.5: 142003071-142003071 on Build GRCh38

View Full Product Details