Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 169695627-169696185 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOCK2
Entrez Gene ID
1794
Gene Name
dedicator of cytokinesis 2
Gene Aliases
IMD40
UniGene
Hs.586174
Gene Chromosome Location
Chr.5: 169637247-170083382 on Build GRCh38
Transcripts
NM_004946.2 XM_005265830.3 XM_011534448.2 XM_011534449.2 XM_011534450.2 XM_011534451.2 XM_017009189.1 XM_017009190.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6170152 c.845A>T p.D282V Chr.5: 169695804-169695804 on Build GRCh38
736808 c.859G>T p.D287Y Chr.5: 169695818-169695818 on Build GRCh38
4803738 c.919C>A p.L307I Chr.5: 169695878-169695878 on Build GRCh38
7239784 c.866A>G p.N289S Chr.5: 169695825-169695825 on Build GRCh38
2152224 c.885G>T p.L295F Chr.5: 169695844-169695844 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs989534402 Chr.5: 169695706-169695706 on Build GRCh38
rs982512709 Chr.5: 169695727-169695727 on Build GRCh38
rs943060885 Chr.5: 169696084-169696084 on Build GRCh38
rs184844141 Chr.5: 169695675-169695675 on Build GRCh38
rs553926409 Chr.5: 169695795-169695795 on Build GRCh38
rs938248362 Chr.5: 169695758-169695758 on Build GRCh38
rs191896706 Chr.5: 169695673-169695673 on Build GRCh38
rs757457405 Chr.5: 169695931-169695931 on Build GRCh38
rs760578545 Chr.5: 169695977-169695977 on Build GRCh38
rs773822361 Chr.5: 169695954-169695954 on Build GRCh38
rs1003997383 Chr.5: 169695896-169695896 on Build GRCh38
rs1036189918 Chr.5: 169695687-169695687 on Build GRCh38
rs369315365 Chr.5: 169695806-169695806 on Build GRCh38
rs114859669 Chr.5: 169696023-169696023 on Build GRCh38
rs780313466 Chr.5: 169695776-169695776 on Build GRCh38
rs769048920 Chr.5: 169695791-169695791 on Build GRCh38
rs3734099 Chr.5: 169695799-169695799 on Build GRCh38
rs370165974 Chr.5: 169695674-169695688 on Build GRCh38
rs534171116 Chr.5: 169695747-169695747 on Build GRCh38
rs748547934 Chr.5: 169695946-169695946 on Build GRCh38
rs116819880 Chr.5: 169695949-169695949 on Build GRCh38
rs759079362 Chr.5: 169695964-169695964 on Build GRCh38
rs187763893 Chr.5: 169695710-169695710 on Build GRCh38
rs933072406 Chr.5: 169695992-169695992 on Build GRCh38
rs758723530 Chr.5: 169695978-169695978 on Build GRCh38
rs756101922 Chr.5: 169695755-169695755 on Build GRCh38
rs960133085 Chr.5: 169695889-169695889 on Build GRCh38
rs369802560 Chr.5: 169695764-169695764 on Build GRCh38
rs545378671 Chr.5: 169696047-169696047 on Build GRCh38
rs997250499 Chr.5: 169695705-169695705 on Build GRCh38
rs920851073 Chr.5: 169696145-169696145 on Build GRCh38
rs753854377 Chr.5: 169695987-169695987 on Build GRCh38
rs770618519 Chr.5: 169695962-169695962 on Build GRCh38
rs774949014 Chr.5: 169695975-169695975 on Build GRCh38
rs774024970 Chr.5: 169695963-169695963 on Build GRCh38
rs745966277 Chr.5: 169695765-169695765 on Build GRCh38
rs568848978 Chr.5: 169695699-169695699 on Build GRCh38
rs367974453 Chr.5: 169695679-169695693 on Build GRCh38
rs759245831 Chr.5: 169695855-169695855 on Build GRCh38
rs930782009 Chr.5: 169696166-169696166 on Build GRCh38
rs774190772 Chr.5: 169695827-169695827 on Build GRCh38
rs73316279 Chr.5: 169695735-169695735 on Build GRCh38
rs199921409 Chr.5: 169695907-169695907 on Build GRCh38
rs752734350 Chr.5: 169695754-169695754 on Build GRCh38
rs966696190 Chr.5: 169695714-169695714 on Build GRCh38
rs904796082 Chr.5: 169696160-169696160 on Build GRCh38
rs749750176 Chr.5: 169695957-169695957 on Build GRCh38
rs764108527 Chr.5: 169695890-169695890 on Build GRCh38
rs967193590 Chr.5: 169696013-169696013 on Build GRCh38
rs762534189 Chr.5: 169695813-169695813 on Build GRCh38
rs758571784 Chr.5: 169695894-169695894 on Build GRCh38
rs755800591 Chr.5: 169695878-169695878 on Build GRCh38
rs548736930 Chr.5: 169695696-169695696 on Build GRCh38
rs373339019 Chr.5: 169695770-169695770 on Build GRCh38
rs200237259 Chr.5: 169695944-169695944 on Build GRCh38
rs982242221 Chr.5: 169696017-169696017 on Build GRCh38
rs767286381 Chr.5: 169695972-169695972 on Build GRCh38
rs753706915 Chr.5: 169695893-169695893 on Build GRCh38
rs765789550 Chr.5: 169695826-169695826 on Build GRCh38
rs959379646 Chr.5: 169696067-169696067 on Build GRCh38
rs369027782 Chr.5: 169695945-169695945 on Build GRCh38
rs766814667 Chr.5: 169696100-169696100 on Build GRCh38
rs748411987 Chr.5: 169695796-169695796 on Build GRCh38
rs768964322 Chr.5: 169695730-169695730 on Build GRCh38
rs146489287 Chr.5: 169695865-169695865 on Build GRCh38
rs28380064 Chr.5: 169696158-169696158 on Build GRCh38
rs996751575 Chr.5: 169695941-169695941 on Build GRCh38
rs763792282 Chr.5: 169695979-169695979 on Build GRCh38
rs777534188 Chr.5: 169695757-169695757 on Build GRCh38
rs201165630 Chr.5: 169695759-169695759 on Build GRCh38
rs763853547 Chr.5: 169695787-169695787 on Build GRCh38
rs192516538 Chr.5: 169695731-169695731 on Build GRCh38
rs1045353276 Chr.5: 169695671-169695671 on Build GRCh38
rs1051479543 Chr.5: 169696021-169696021 on Build GRCh38

View Full Product Details