Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 16274301-16274822 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMPR
Entrez Gene ID
2766
Gene Name
guanosine monophosphate reductase
Gene Aliases
GMPR 1, GMPR1
UniGene
Hs.484741
Gene Chromosome Location
Chr.6: 16238580-16295549 on Build GRCh38
Transcripts
NM_006877.3 XM_011514508.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
161296 c.547+5G>C p.? Chr.6: 16274501-16274501 on Build GRCh38
3430321 c.496G>T p.E166* Chr.6: 16274445-16274445 on Build GRCh38
1442110 c.487G>T p.E163* Chr.6: 16274436-16274436 on Build GRCh38
483669 c.514G>A p.G172R Chr.6: 16274463-16274463 on Build GRCh38
6574763 c.496G>C p.E166Q Chr.6: 16274445-16274445 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779812547 Chr.6: 16274443-16274443 on Build GRCh38
rs752919227 Chr.6: 16274339-16274339 on Build GRCh38
rs900102739 Chr.6: 16274649-16274649 on Build GRCh38
rs537900640 Chr.6: 16274751-16274751 on Build GRCh38
rs552306886 Chr.6: 16274582-16274582 on Build GRCh38
rs761809814 Chr.6: 16274366-16274366 on Build GRCh38
rs753178854 Chr.6: 16274398-16274398 on Build GRCh38
rs905309149 Chr.6: 16274569-16274569 on Build GRCh38
rs903599888 Chr.6: 16274677-16274677 on Build GRCh38
rs565942882 Chr.6: 16274616-16274616 on Build GRCh38
rs764473763 Chr.6: 16274514-16274514 on Build GRCh38
rs893972825 Chr.6: 16274331-16274331 on Build GRCh38
rs373759283 Chr.6: 16274429-16274429 on Build GRCh38
rs1013848435 Chr.6: 16274764-16274764 on Build GRCh38
rs185870947 Chr.6: 16274733-16274733 on Build GRCh38
rs931403213 Chr.6: 16274572-16274572 on Build GRCh38
rs62387698 Chr.6: 16274369-16274369 on Build GRCh38
rs755635778 Chr.6: 16274519-16274519 on Build GRCh38
rs375025738 Chr.6: 16274465-16274465 on Build GRCh38
rs772917096 Chr.6: 16274800-16274800 on Build GRCh38
rs771167264 Chr.6: 16274486-16274486 on Build GRCh38
rs10484359 Chr.6: 16274395-16274395 on Build GRCh38
rs143907374 Chr.6: 16274726-16274726 on Build GRCh38
rs763755103 Chr.6: 16274452-16274454 on Build GRCh38
rs931627947 Chr.6: 16274656-16274656 on Build GRCh38
rs528396154 Chr.6: 16274462-16274462 on Build GRCh38
rs45513699 Chr.6: 16274679-16274679 on Build GRCh38
rs921745956 Chr.6: 16274696-16274696 on Build GRCh38
rs754236935 Chr.6: 16274378-16274378 on Build GRCh38
rs374928250 Chr.6: 16274526-16274526 on Build GRCh38
rs149938350 Chr.6: 16274497-16274497 on Build GRCh38
rs201792237 Chr.6: 16274376-16274376 on Build GRCh38
rs749984472 Chr.6: 16274515-16274515 on Build GRCh38
rs758391748 Chr.6: 16274542-16274542 on Build GRCh38
rs766141065 Chr.6: 16274524-16274524 on Build GRCh38
rs758823883 Chr.6: 16274415-16274415 on Build GRCh38
rs763187138 Chr.6: 16274559-16274559 on Build GRCh38
rs368811344 Chr.6: 16274529-16274529 on Build GRCh38
rs771712074 Chr.6: 16274566-16274566 on Build GRCh38
rs759848366 Chr.6: 16274392-16274392 on Build GRCh38
rs771882851 Chr.6: 16274467-16274467 on Build GRCh38
rs773382464 Chr.6: 16274472-16274472 on Build GRCh38
rs749116791 Chr.6: 16274458-16274458 on Build GRCh38
rs911604306 Chr.6: 16274749-16274749 on Build GRCh38
rs747055824 Chr.6: 16274473-16274473 on Build GRCh38
rs749927150 Chr.6: 16274424-16274424 on Build GRCh38
rs1000667907 Chr.6: 16274516-16274516 on Build GRCh38
rs1000658204 Chr.6: 16274700-16274700 on Build GRCh38
rs62387699 Chr.6: 16274548-16274548 on Build GRCh38
rs754575430 Chr.6: 16274527-16274527 on Build GRCh38
rs1004177323 Chr.6: 16274663-16274663 on Build GRCh38
rs919970423 Chr.6: 16274570-16274570 on Build GRCh38
rs951389283 Chr.6: 16274407-16274407 on Build GRCh38
rs373547039 Chr.6: 16274372-16274372 on Build GRCh38
rs372847384 Chr.6: 16274533-16274533 on Build GRCh38
rs760937715 Chr.6: 16274375-16274375 on Build GRCh38
rs776779386 Chr.6: 16274488-16274488 on Build GRCh38
rs901531917 Chr.6: 16274434-16274434 on Build GRCh38
rs1010988222 Chr.6: 16274342-16274342 on Build GRCh38
rs992115304 Chr.6: 16274543-16274543 on Build GRCh38
rs907751346 Chr.6: 16274455-16274455 on Build GRCh38
rs9396661 Chr.6: 16274560-16274560 on Build GRCh38
rs774480087 Chr.6: 16274665-16274665 on Build GRCh38
rs45472698 Chr.6: 16274508-16274508 on Build GRCh38
rs765697418 Chr.6: 16274498-16274498 on Build GRCh38
rs368739126 Chr.6: 16274463-16274463 on Build GRCh38
rs1050524144 Chr.6: 16274657-16274657 on Build GRCh38
rs1041327525 Chr.6: 16274602-16274602 on Build GRCh38
rs908753447 Chr.6: 16274577-16274577 on Build GRCh38
rs543959846 Chr.6: 16274552-16274552 on Build GRCh38
rs574860685 Chr.6: 16274352-16274352 on Build GRCh38
rs369298282 Chr.6: 16274575-16274575 on Build GRCh38
rs973264433 Chr.6: 16274681-16274681 on Build GRCh38
rs763561635 Chr.6: 16274509-16274509 on Build GRCh38
rs114417367 Chr.6: 16274423-16274423 on Build GRCh38
rs754760026 Chr.6: 16274461-16274461 on Build GRCh38

View Full Product Details