Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32293483-32294026 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf10
Entrez Gene ID
10665
Gene Name
chromosome 6 open reading frame 10
Gene Aliases
TSBP
UniGene
Hs.567414
Gene Chromosome Location
Chr.6: 32290523-32371912 on Build GRCh38
Transcripts
NM_001286474.1 NM_001286475.1 NM_006781.4 XM_017010182.1 XM_017010183.1 XM_017010184.1 XM_017010185.1 NR_136244.1 NR_136245.1 NR_136246.1

Gene Symbol
LOC101929163
Entrez Gene ID
101929163
Gene Name
uncharacterized LOC101929163
Gene Aliases
-
UniGene
Hs.57692
Gene Chromosome Location
Chr.6: 32255173-32407730 on Build GRCh38
Transcripts
NM_001286474.1 NM_001286475.1 NM_006781.4 XM_017010182.1 XM_017010183.1 XM_017010184.1 XM_017010185.1 NR_136244.1 NR_136245.1 NR_136246.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
342539 c.780G>A p.T260T Chr.6: 32293893-32293893 on Build GRCh38
4003970 c.943A>G p.I315V Chr.6: 32293730-32293730 on Build GRCh38
6616368 c.1190A>G p.Q397R Chr.6: 32293483-32293483 on Build GRCh38
5804302 c.1190A>T p.Q397L Chr.6: 32293483-32293483 on Build GRCh38
1578151 c.1093G>A p.E365K Chr.6: 32293580-32293580 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748076073 Chr.6: 32293694-32293694 on Build GRCh38
rs745483322 Chr.6: 32293523-32293523 on Build GRCh38
rs768324713 Chr.6: 32293612-32293612 on Build GRCh38
rs918064003 Chr.6: 32293977-32293977 on Build GRCh38
rs151122056 Chr.6: 32293734-32293734 on Build GRCh38
rs373631463 Chr.6: 32293576-32293576 on Build GRCh38
rs35862719 Chr.6: 32293635-32293636 on Build GRCh38
rs1030537744 Chr.6: 32293662-32293662 on Build GRCh38
rs775838297 Chr.6: 32293729-32293729 on Build GRCh38
rs759143582 Chr.6: 32293992-32293993 on Build GRCh38
rs889543234 Chr.6: 32293546-32293546 on Build GRCh38
rs928491415 Chr.6: 32293759-32293759 on Build GRCh38
rs373884140 Chr.6: 32293782-32293782 on Build GRCh38
rs749027279 Chr.6: 32293678-32293678 on Build GRCh38
rs866361411 Chr.6: 32293658-32293658 on Build GRCh38
rs878930594 Chr.6: 32293533-32293533 on Build GRCh38
rs34498363 Chr.6: 32293853-32293853 on Build GRCh38
rs139286311 Chr.6: 32293936-32293936 on Build GRCh38
rs1027247399 Chr.6: 32293602-32293602 on Build GRCh38
rs746634526 Chr.6: 32293701-32293701 on Build GRCh38
rs61995678 Chr.6: 32293514-32293514 on Build GRCh38
rs574400346 Chr.6: 32293555-32293555 on Build GRCh38
rs148937489 Chr.6: 32293717-32293717 on Build GRCh38
rs754616115 Chr.6: 32293836-32293836 on Build GRCh38
rs762252379 Chr.6: 32293741-32293741 on Build GRCh38
rs778347240 Chr.6: 32293504-32293504 on Build GRCh38
rs775464273 Chr.6: 32293904-32293904 on Build GRCh38
rs541720018 Chr.6: 32293647-32293647 on Build GRCh38
rs763188201 Chr.6: 32293906-32293906 on Build GRCh38
rs934519480 Chr.6: 32293760-32293760 on Build GRCh38
rs376834294 Chr.6: 32293643-32293643 on Build GRCh38
rs752148513 Chr.6: 32293665-32293665 on Build GRCh38
rs749545467 Chr.6: 32293532-32293533 on Build GRCh38
rs756842414 Chr.6: 32293517-32293517 on Build GRCh38
rs753563883 Chr.6: 32293800-32293800 on Build GRCh38
rs889429962 Chr.6: 32293865-32293865 on Build GRCh38
rs145900218 Chr.6: 32293978-32293978 on Build GRCh38
rs138704487 Chr.6: 32293987-32293987 on Build GRCh38
rs868112596 Chr.6: 32293502-32293502 on Build GRCh38
rs764486432 Chr.6: 32293664-32293664 on Build GRCh38
rs4947338 Chr.6: 32293876-32293876 on Build GRCh38
rs564447553 Chr.6: 32293733-32293733 on Build GRCh38
rs774713461 Chr.6: 32293934-32293934 on Build GRCh38
rs758326356 Chr.6: 32294001-32294001 on Build GRCh38
rs765869085 Chr.6: 32293968-32293968 on Build GRCh38
rs755683262 Chr.6: 32293786-32293786 on Build GRCh38
rs997615836 Chr.6: 32293649-32293649 on Build GRCh38
rs3749966 Chr.6: 32293730-32293730 on Build GRCh38
rs371233809 Chr.6: 32293656-32293656 on Build GRCh38
rs770745594 Chr.6: 32293799-32293800 on Build GRCh38
rs757738275 Chr.6: 32293667-32293667 on Build GRCh38
rs892577560 Chr.6: 32293645-32293645 on Build GRCh38
rs961566088 Chr.6: 32293574-32293574 on Build GRCh38
rs563184862 Chr.6: 32293676-32293676 on Build GRCh38
rs369976372 Chr.6: 32293556-32293556 on Build GRCh38
rs866952150 Chr.6: 32293600-32293600 on Build GRCh38
rs143188052 Chr.6: 32293951-32293951 on Build GRCh38
rs776274937 Chr.6: 32293955-32293956 on Build GRCh38
rs867579325 Chr.6: 32293637-32293637 on Build GRCh38
rs963020609 Chr.6: 32293812-32293812 on Build GRCh38
rs377471914 Chr.6: 32293552-32293552 on Build GRCh38
rs1016218176 Chr.6: 32293808-32293808 on Build GRCh38
rs916556095 Chr.6: 32293695-32293695 on Build GRCh38
rs780791775 Chr.6: 32293521-32293521 on Build GRCh38
rs947275416 Chr.6: 32293829-32293829 on Build GRCh38
rs774120324 Chr.6: 32293685-32293685 on Build GRCh38
rs749934218 Chr.6: 32293785-32293785 on Build GRCh38
rs772947936 Chr.6: 32293646-32293649 on Build GRCh38
rs755699475 Chr.6: 32293671-32293671 on Build GRCh38
rs938397621 Chr.6: 32293879-32293879 on Build GRCh38
rs780634055 Chr.6: 32293668-32293668 on Build GRCh38
rs760645031 Chr.6: 32293611-32293611 on Build GRCh38
rs772065928 Chr.6: 32293863-32293863 on Build GRCh38
rs745928315 Chr.6: 32293885-32293885 on Build GRCh38
rs868769441 Chr.6: 32293706-32293706 on Build GRCh38
rs560505 Chr.6: 32293994-32293994 on Build GRCh38
rs768002126 Chr.6: 32293937-32293937 on Build GRCh38
rs768775788 Chr.6: 32293914-32293914 on Build GRCh38
rs772155625 Chr.6: 32293699-32293699 on Build GRCh38
rs866939556 Chr.6: 32293766-32293766 on Build GRCh38
rs776878975 Chr.6: 32293628-32293628 on Build GRCh38
rs779555362 Chr.6: 32293792-32293792 on Build GRCh38
rs745399589 Chr.6: 32293670-32293670 on Build GRCh38
rs747984647 Chr.6: 32293712-32293712 on Build GRCh38
rs769459897 Chr.6: 32293541-32293541 on Build GRCh38
rs766604573 Chr.6: 32293625-32293625 on Build GRCh38
rs758863964 Chr.6: 32293657-32293657 on Build GRCh38
rs770035027 Chr.6: 32293894-32293894 on Build GRCh38
rs948268069 Chr.6: 32293525-32293525 on Build GRCh38
rs1020223737 Chr.6: 32293561-32293561 on Build GRCh38
rs780930636 Chr.6: 32293681-32293681 on Build GRCh38
rs769092254 Chr.6: 32293612-32293613 on Build GRCh38
rs774401145 Chr.6: 32293608-32293608 on Build GRCh38
rs768542271 Chr.6: 32293683-32293683 on Build GRCh38
rs750981966 Chr.6: 32293941-32293941 on Build GRCh38
rs530604728 Chr.6: 32293716-32293716 on Build GRCh38

View Full Product Details