Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 37020339-37020862 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGD2
Entrez Gene ID
221472
Gene Name
FYVE, RhoGEF and PH domain containing 2
Gene Aliases
ZFYVE4
UniGene
Hs.509664
Gene Chromosome Location
Chr.6: 37005647-37029072 on Build GRCh38
Transcripts
NM_173558.3 XM_017010429.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5788511 c.1201C>G p.R401G Chr.6: 37020619-37020619 on Build GRCh38
4161126 c.1170C>T p.S390S Chr.6: 37020588-37020588 on Build GRCh38
116055 c.1124T>G p.V375G Chr.6: 37020542-37020542 on Build GRCh38
6304226 c.1180C>T p.R394C Chr.6: 37020598-37020598 on Build GRCh38
5531232 c.1223C>T p.S408F Chr.6: 37020729-37020729 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs970822395 Chr.6: 37020405-37020405 on Build GRCh38
rs1009557356 Chr.6: 37020828-37020828 on Build GRCh38
rs199576667 Chr.6: 37020787-37020787 on Build GRCh38
rs749426264 Chr.6: 37020599-37020599 on Build GRCh38
rs186152460 Chr.6: 37020648-37020648 on Build GRCh38
rs762301573 Chr.6: 37020619-37020619 on Build GRCh38
rs911997839 Chr.6: 37020657-37020657 on Build GRCh38
rs944733881 Chr.6: 37020476-37020476 on Build GRCh38
rs778296854 Chr.6: 37020654-37020654 on Build GRCh38
rs758421033 Chr.6: 37020696-37020696 on Build GRCh38
rs56016101 Chr.6: 37020420-37020420 on Build GRCh38
rs779046545 Chr.6: 37020569-37020569 on Build GRCh38
rs774894854 Chr.6: 37020617-37020617 on Build GRCh38
rs749497583 Chr.6: 37020755-37020755 on Build GRCh38
rs746847649 Chr.6: 37020737-37020737 on Build GRCh38
rs894622806 Chr.6: 37020510-37020510 on Build GRCh38
rs982226660 Chr.6: 37020601-37020601 on Build GRCh38
rs767956722 Chr.6: 37020620-37020620 on Build GRCh38
rs532829449 Chr.6: 37020478-37020478 on Build GRCh38
rs533775023 Chr.6: 37020504-37020504 on Build GRCh38
rs754510582 Chr.6: 37020545-37020545 on Build GRCh38
rs753419995 Chr.6: 37020544-37020544 on Build GRCh38
rs777519694 Chr.6: 37020703-37020703 on Build GRCh38
rs375261795 Chr.6: 37020575-37020575 on Build GRCh38
rs1027791929 Chr.6: 37020505-37020505 on Build GRCh38
rs760951784 Chr.6: 37020568-37020568 on Build GRCh38
rs754010886 Chr.6: 37020623-37020624 on Build GRCh38
rs749795960 Chr.6: 37020702-37020702 on Build GRCh38
rs831504 Chr.6: 37020588-37020588 on Build GRCh38
rs768699381 Chr.6: 37020615-37020615 on Build GRCh38
rs775652042 Chr.6: 37020490-37020490 on Build GRCh38
rs762211617 Chr.6: 37020519-37020519 on Build GRCh38
rs368683361 Chr.6: 37020541-37020541 on Build GRCh38
rs113388273 Chr.6: 37020538-37020538 on Build GRCh38
rs775530130 Chr.6: 37020593-37020593 on Build GRCh38
rs890528694 Chr.6: 37020473-37020473 on Build GRCh38
rs752026530 Chr.6: 37020513-37020513 on Build GRCh38
rs1040255077 Chr.6: 37020598-37020598 on Build GRCh38
rs773650260 Chr.6: 37020639-37020639 on Build GRCh38
rs1033536368 Chr.6: 37020483-37020483 on Build GRCh38
rs550335367 Chr.6: 37020416-37020416 on Build GRCh38
rs935235005 Chr.6: 37020422-37020422 on Build GRCh38
rs374612325 Chr.6: 37020814-37020814 on Build GRCh38
rs915045161 Chr.6: 37020433-37020433 on Build GRCh38
rs755422593 Chr.6: 37020743-37020744 on Build GRCh38
rs866997298 Chr.6: 37020708-37020708 on Build GRCh38
rs556294687 Chr.6: 37020709-37020709 on Build GRCh38
rs1032276838 Chr.6: 37020391-37020391 on Build GRCh38
rs752186227 Chr.6: 37020692-37020692 on Build GRCh38
rs989318177 Chr.6: 37020432-37020432 on Build GRCh38
rs770074264 Chr.6: 37020589-37020589 on Build GRCh38
rs796415299 Chr.6: 37020423-37020423 on Build GRCh38
rs370264308 Chr.6: 37020749-37020749 on Build GRCh38
rs932069273 Chr.6: 37020436-37020436 on Build GRCh38
rs765776003 Chr.6: 37020785-37020786 on Build GRCh38
rs764838631 Chr.6: 37020687-37020687 on Build GRCh38
rs200949140 Chr.6: 37020496-37020496 on Build GRCh38
rs1030629765 Chr.6: 37020614-37020614 on Build GRCh38
rs1049323199 Chr.6: 37020752-37020752 on Build GRCh38
rs766361926 Chr.6: 37020587-37020587 on Build GRCh38
rs768789313 Chr.6: 37020758-37020758 on Build GRCh38
rs748087766 Chr.6: 37020576-37020576 on Build GRCh38
rs879099160 Chr.6: 37020800-37020800 on Build GRCh38
rs181848181 Chr.6: 37020531-37020531 on Build GRCh38
rs766096616 Chr.6: 37020646-37020646 on Build GRCh38
rs59228579 Chr.6: 37020484-37020484 on Build GRCh38
rs765902273 Chr.6: 37020542-37020542 on Build GRCh38
rs376711130 Chr.6: 37020655-37020655 on Build GRCh38
rs970578890 Chr.6: 37020457-37020457 on Build GRCh38
rs756970544 Chr.6: 37020738-37020738 on Build GRCh38
rs777712865 Chr.6: 37020584-37020584 on Build GRCh38
rs894716223 Chr.6: 37020754-37020754 on Build GRCh38
rs767852638 Chr.6: 37020520-37020520 on Build GRCh38
rs1050917449 Chr.6: 37020458-37020458 on Build GRCh38
rs145257454 Chr.6: 37020579-37020579 on Build GRCh38
rs764611598 Chr.6: 37020509-37020509 on Build GRCh38
rs944768162 Chr.6: 37020682-37020682 on Build GRCh38

View Full Product Details