Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 57171909-57172464 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAG2
Entrez Gene ID
9532
Gene Name
BCL2 associated athanogene 2
Gene Aliases
BAG-2, dJ417I1.2
UniGene
Hs.745046
Gene Chromosome Location
Chr.6: 57172306-57185215 on Build GRCh38
Transcripts
NM_004282.3 NR_110742.1

Gene Symbol
LOC101927211
Entrez Gene ID
101927211
Gene Name
uncharacterized LOC101927211
Gene Aliases
-
UniGene
Hs.705718
Gene Chromosome Location
Chr.6: 57114911-57172213 on Build GRCh38
Transcripts
NM_004282.3 NR_110742.1

SNP Detail

SNP ID Chromosome Location
rs766719751 Chr.6: 57171960-57171960 on Build GRCh38
rs756090899 Chr.6: 57172071-57172072 on Build GRCh38
rs1026472626 Chr.6: 57171950-57171950 on Build GRCh38
rs62415926 Chr.6: 57172197-57172197 on Build GRCh38
rs909595047 Chr.6: 57171992-57171992 on Build GRCh38
rs182728850 Chr.6: 57172221-57172221 on Build GRCh38
rs1008957328 Chr.6: 57172283-57172283 on Build GRCh38
rs933639193 Chr.6: 57172268-57172268 on Build GRCh38
rs935807610 Chr.6: 57172071-57172071 on Build GRCh38
rs927632175 Chr.6: 57172399-57172399 on Build GRCh38
rs901863408 Chr.6: 57172413-57172413 on Build GRCh38
rs999361087 Chr.6: 57172274-57172274 on Build GRCh38
rs902323135 Chr.6: 57172444-57172444 on Build GRCh38
rs747631144 Chr.6: 57172222-57172222 on Build GRCh38
rs753108464 Chr.6: 57172024-57172024 on Build GRCh38
rs939022380 Chr.6: 57172406-57172406 on Build GRCh38
rs185319011 Chr.6: 57172212-57172212 on Build GRCh38
rs1031631040 Chr.6: 57171982-57171982 on Build GRCh38
rs755644072 Chr.6: 57172224-57172224 on Build GRCh38
rs756605975 Chr.6: 57172189-57172189 on Build GRCh38
rs532204254 Chr.6: 57172319-57172319 on Build GRCh38
rs750794043 Chr.6: 57172149-57172149 on Build GRCh38
rs964886889 Chr.6: 57172009-57172009 on Build GRCh38
rs529981253 Chr.6: 57172397-57172397 on Build GRCh38
rs1055186880 Chr.6: 57172408-57172408 on Build GRCh38
rs924494758 Chr.6: 57172371-57172371 on Build GRCh38
rs920720024 Chr.6: 57172304-57172304 on Build GRCh38
rs973634785 Chr.6: 57172303-57172303 on Build GRCh38
rs977427786 Chr.6: 57171974-57171974 on Build GRCh38
rs917204425 Chr.6: 57172014-57172014 on Build GRCh38
rs889198635 Chr.6: 57172164-57172164 on Build GRCh38
rs144323378 Chr.6: 57172082-57172082 on Build GRCh38
rs1043570868 Chr.6: 57172436-57172436 on Build GRCh38
rs542500739 Chr.6: 57172020-57172020 on Build GRCh38
rs1017715452 Chr.6: 57172293-57172293 on Build GRCh38
rs1026546093 Chr.6: 57172263-57172263 on Build GRCh38
rs879703989 Chr.6: 57172424-57172424 on Build GRCh38
rs1010768459 Chr.6: 57172234-57172234 on Build GRCh38
rs1040453298 Chr.6: 57172410-57172410 on Build GRCh38
rs575042941 Chr.6: 57172006-57172006 on Build GRCh38
rs954684403 Chr.6: 57172280-57172280 on Build GRCh38
rs967017005 Chr.6: 57171963-57171963 on Build GRCh38
rs902125177 Chr.6: 57172227-57172227 on Build GRCh38
rs991588223 Chr.6: 57172313-57172313 on Build GRCh38
rs971011955 Chr.6: 57172326-57172326 on Build GRCh38
rs910725779 Chr.6: 57172008-57172008 on Build GRCh38
rs1006357312 Chr.6: 57172290-57172290 on Build GRCh38
rs965147415 Chr.6: 57172295-57172295 on Build GRCh38
rs983674474 Chr.6: 57172356-57172356 on Build GRCh38
rs564653714 Chr.6: 57172236-57172236 on Build GRCh38
rs547276219 Chr.6: 57172329-57172329 on Build GRCh38
rs190596477 Chr.6: 57172219-57172219 on Build GRCh38
rs929253542 Chr.6: 57172428-57172428 on Build GRCh38
rs752172994 Chr.6: 57172046-57172046 on Build GRCh38
rs755442846 Chr.6: 57172182-57172182 on Build GRCh38
rs958480892 Chr.6: 57172310-57172310 on Build GRCh38
rs935866163 Chr.6: 57172383-57172384 on Build GRCh38
rs939095841 Chr.6: 57172214-57172214 on Build GRCh38
rs546550492 Chr.6: 57172039-57172039 on Build GRCh38
rs565825140 Chr.6: 57172393-57172393 on Build GRCh38
rs758814081 Chr.6: 57171936-57171936 on Build GRCh38
rs989996124 Chr.6: 57172392-57172392 on Build GRCh38
rs764977710 Chr.6: 57172272-57172272 on Build GRCh38
rs916367728 Chr.6: 57172417-57172417 on Build GRCh38
rs879046497 Chr.6: 57172277-57172277 on Build GRCh38
rs987483703 Chr.6: 57172007-57172007 on Build GRCh38

View Full Product Details