Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 134679366-134679964 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPGM
Entrez Gene ID
669
Gene Name
bisphosphoglycerate mutase
Gene Aliases
DPGM
UniGene
Hs.198365
Gene Chromosome Location
Chr.7: 134646779-134679816 on Build GRCh38
Transcripts
NM_001293085.1 NM_001724.4 NM_199186.2 XM_011516527.1

SNP Detail

SNP ID Chromosome Location
rs961852411 Chr.7: 134679577-134679577 on Build GRCh38
rs953451391 Chr.7: 134679533-134679533 on Build GRCh38
rs1031195067 Chr.7: 134679453-134679453 on Build GRCh38
rs911245256 Chr.7: 134679933-134679933 on Build GRCh38
rs1025106905 Chr.7: 134679832-134679832 on Build GRCh38
rs200558459 Chr.7: 134679854-134679862 on Build GRCh38
rs184837736 Chr.7: 134679569-134679569 on Build GRCh38
rs999709640 Chr.7: 134679435-134679435 on Build GRCh38
rs556831554 Chr.7: 134679644-134679644 on Build GRCh38
rs894060069 Chr.7: 134679821-134679821 on Build GRCh38
rs1010855793 Chr.7: 134679482-134679482 on Build GRCh38
rs187225222 Chr.7: 134679853-134679853 on Build GRCh38
rs199546846 Chr.7: 134679709-134679709 on Build GRCh38
rs367962105 Chr.7: 134679583-134679583 on Build GRCh38
rs189346801 Chr.7: 134679637-134679637 on Build GRCh38
rs978323841 Chr.7: 134679872-134679873 on Build GRCh38
rs753439869 Chr.7: 134679895-134679895 on Build GRCh38
rs200757659 Chr.7: 134679712-134679712 on Build GRCh38
rs1048101787 Chr.7: 134679593-134679593 on Build GRCh38
rs200662343 Chr.7: 134679898-134679898 on Build GRCh38
rs889130961 Chr.7: 134679880-134679880 on Build GRCh38
rs1028961870 Chr.7: 134679834-134679834 on Build GRCh38
rs57700085 Chr.7: 134679897-134679900 on Build GRCh38
rs942777046 Chr.7: 134679560-134679560 on Build GRCh38
rs971296206 Chr.7: 134679845-134679845 on Build GRCh38
rs1029449336 Chr.7: 134679522-134679522 on Build GRCh38
rs755705040 Chr.7: 134679824-134679824 on Build GRCh38
rs1025581678 Chr.7: 134679462-134679462 on Build GRCh38
rs915569740 Chr.7: 134679651-134679651 on Build GRCh38
rs978757521 Chr.7: 134679464-134679464 on Build GRCh38
rs373501885 Chr.7: 134679707-134679708 on Build GRCh38
rs989506719 Chr.7: 134679494-134679495 on Build GRCh38
rs10249334 Chr.7: 134679925-134679925 on Build GRCh38
rs34972996 Chr.7: 134679891-134679894 on Build GRCh38
rs924251964 Chr.7: 134679465-134679465 on Build GRCh38
rs935489054 Chr.7: 134679775-134679775 on Build GRCh38
rs1045908006 Chr.7: 134679678-134679678 on Build GRCh38
rs181202926 Chr.7: 134679406-134679406 on Build GRCh38
rs1005879193 Chr.7: 134679403-134679403 on Build GRCh38
rs936653654 Chr.7: 134679489-134679489 on Build GRCh38
rs1038844435 Chr.7: 134679565-134679565 on Build GRCh38
rs545767537 Chr.7: 134679813-134679813 on Build GRCh38
rs3202442 Chr.7: 134679581-134679581 on Build GRCh38
rs180874470 Chr.7: 134679662-134679662 on Build GRCh38
rs987971007 Chr.7: 134679547-134679547 on Build GRCh38
rs767527465 Chr.7: 134679514-134679514 on Build GRCh38
rs11550980 Chr.7: 134679584-134679584 on Build GRCh38
rs1008961896 Chr.7: 134679889-134679889 on Build GRCh38
rs534540271 Chr.7: 134679463-134679463 on Build GRCh38
rs911297431 Chr.7: 134679550-134679550 on Build GRCh38
rs1019432361 Chr.7: 134679895-134679895 on Build GRCh38
rs1011579281 Chr.7: 134679825-134679825 on Build GRCh38
rs1061362 Chr.7: 134679578-134679578 on Build GRCh38
rs959141970 Chr.7: 134679407-134679413 on Build GRCh38
rs753519968 Chr.7: 134679603-134679603 on Build GRCh38

View Full Product Details