Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 27735757-27736343 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC25
Entrez Gene ID
55246
Gene Name
coiled-coil domain containing 25
Gene Aliases
-
UniGene
Hs.445512
Gene Chromosome Location
Chr.8: 27733316-27772656 on Build GRCh38
Transcripts
NM_001304529.1 NM_001304530.1 NM_001304532.1 NM_018246.2 NR_130761.1 NR_130762.1 XM_011544570.2 XM_011544571.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1456415 c.602G>T p.G201V Chr.8: 27736241-27736241 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1033849168 Chr.8: 27736009-27736009 on Build GRCh38
rs1013801005 Chr.8: 27736030-27736030 on Build GRCh38
rs903927255 Chr.8: 27735979-27735979 on Build GRCh38
rs770331873 Chr.8: 27736280-27736280 on Build GRCh38
rs532650614 Chr.8: 27736305-27736305 on Build GRCh38
rs3832544 Chr.8: 27735923-27735924 on Build GRCh38
rs781600337 Chr.8: 27736263-27736263 on Build GRCh38
rs749308198 Chr.8: 27736294-27736294 on Build GRCh38
rs961003846 Chr.8: 27736072-27736072 on Build GRCh38
rs544392457 Chr.8: 27736071-27736071 on Build GRCh38
rs779808495 Chr.8: 27736238-27736238 on Build GRCh38
rs112519209 Chr.8: 27736096-27736096 on Build GRCh38
rs182948247 Chr.8: 27736007-27736007 on Build GRCh38
rs1049575745 Chr.8: 27735868-27735868 on Build GRCh38
rs943882831 Chr.8: 27736223-27736223 on Build GRCh38
rs116616175 Chr.8: 27735974-27735974 on Build GRCh38
rs778258197 Chr.8: 27736283-27736283 on Build GRCh38
rs193049795 Chr.8: 27735799-27735799 on Build GRCh38
rs10081527 Chr.8: 27736133-27736133 on Build GRCh38
rs113447699 Chr.8: 27735787-27735787 on Build GRCh38
rs932425487 Chr.8: 27735829-27735829 on Build GRCh38
rs749370191 Chr.8: 27736194-27736195 on Build GRCh38
rs537236479 Chr.8: 27735784-27735784 on Build GRCh38
rs765329289 Chr.8: 27736181-27736181 on Build GRCh38
rs3735748 Chr.8: 27736026-27736026 on Build GRCh38
rs892443988 Chr.8: 27735941-27735941 on Build GRCh38
rs561280999 Chr.8: 27736130-27736130 on Build GRCh38
rs778552840 Chr.8: 27736248-27736248 on Build GRCh38
rs199790637 Chr.8: 27736214-27736214 on Build GRCh38
rs768590461 Chr.8: 27736177-27736177 on Build GRCh38
rs374012430 Chr.8: 27736265-27736265 on Build GRCh38
rs377290483 Chr.8: 27736258-27736258 on Build GRCh38
rs779685602 Chr.8: 27736085-27736085 on Build GRCh38
rs749882415 Chr.8: 27736207-27736207 on Build GRCh38
rs748520719 Chr.8: 27736274-27736274 on Build GRCh38
rs540646107 Chr.8: 27736139-27736139 on Build GRCh38
rs973690061 Chr.8: 27736092-27736092 on Build GRCh38
rs552664085 Chr.8: 27735889-27735889 on Build GRCh38
rs943932752 Chr.8: 27735885-27735885 on Build GRCh38
rs754825805 Chr.8: 27736197-27736197 on Build GRCh38
rs745542951 Chr.8: 27736195-27736202 on Build GRCh38
rs1009589708 Chr.8: 27735957-27735957 on Build GRCh38
rs890267509 Chr.8: 27736293-27736293 on Build GRCh38
rs757991462 Chr.8: 27736234-27736234 on Build GRCh38
rs3735747 Chr.8: 27736064-27736064 on Build GRCh38
rs536271602 Chr.8: 27735884-27735884 on Build GRCh38
rs753622511 Chr.8: 27736167-27736167 on Build GRCh38
rs112789683 Chr.8: 27736064-27736064 on Build GRCh38
rs757202940 Chr.8: 27736173-27736173 on Build GRCh38
rs751291401 Chr.8: 27736243-27736243 on Build GRCh38
rs748159745 Chr.8: 27736282-27736282 on Build GRCh38

View Full Product Details