Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 21967036-21967599 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDKN2A-AS1
Entrez Gene ID
51198
Gene Name
CDKN2A antisense RNA 1 (head to head)
Gene Aliases
C9orf53, bA149I2.3
UniGene
Hs.721053
Gene Chromosome Location
Chr.9: 21967139-21967754 on Build GRCh38
Transcripts
NR_024274.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6205164 c.156C>T p.V52V Chr.9: 21967355-21967355 on Build GRCh38
6286797 c.53G>T p.R18L Chr.9: 21967252-21967252 on Build GRCh38
7428153 c.186G>A p.P62P Chr.9: 21967385-21967385 on Build GRCh38
4445513 c.207G>T p.W69C Chr.9: 21967406-21967406 on Build GRCh38
5998652 c.180G>T p.W60C Chr.9: 21967379-21967379 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs547171109 Chr.9: 21967501-21967501 on Build GRCh38
rs751799650 Chr.9: 21967440-21967440 on Build GRCh38
rs753388237 Chr.9: 21967443-21967443 on Build GRCh38
rs559224584 Chr.9: 21967319-21967319 on Build GRCh38
rs1013380697 Chr.9: 21967138-21967138 on Build GRCh38
rs148131172 Chr.9: 21967327-21967327 on Build GRCh38
rs767211664 Chr.9: 21967370-21967370 on Build GRCh38
rs11793581 Chr.9: 21967447-21967447 on Build GRCh38
rs962080787 Chr.9: 21967106-21967106 on Build GRCh38
rs1037810784 Chr.9: 21967141-21967145 on Build GRCh38
rs533589918 Chr.9: 21967496-21967496 on Build GRCh38
rs746854733 Chr.9: 21967490-21967499 on Build GRCh38
rs1009534921 Chr.9: 21967561-21967561 on Build GRCh38
rs781782526 Chr.9: 21967224-21967224 on Build GRCh38
rs752469998 Chr.9: 21967164-21967165 on Build GRCh38
rs541465553 Chr.9: 21967332-21967332 on Build GRCh38
rs899283989 Chr.9: 21967459-21967459 on Build GRCh38
rs934550861 Chr.9: 21967517-21967517 on Build GRCh38
rs924432756 Chr.9: 21967500-21967500 on Build GRCh38
rs981379336 Chr.9: 21967486-21967486 on Build GRCh38
rs952228219 Chr.9: 21967439-21967439 on Build GRCh38
rs915789618 Chr.9: 21967160-21967160 on Build GRCh38
rs1017654277 Chr.9: 21967267-21967267 on Build GRCh38
rs938686958 Chr.9: 21967109-21967109 on Build GRCh38
rs768959694 Chr.9: 21967318-21967318 on Build GRCh38
rs1055055398 Chr.9: 21967418-21967418 on Build GRCh38
rs966501996 Chr.9: 21967317-21967317 on Build GRCh38
rs753588488 Chr.9: 21967164-21967164 on Build GRCh38
rs1009123945 Chr.9: 21967356-21967356 on Build GRCh38
rs556906507 Chr.9: 21967241-21967241 on Build GRCh38
rs530545351 Chr.9: 21967412-21967412 on Build GRCh38
rs899316479 Chr.9: 21967145-21967145 on Build GRCh38
rs1009155340 Chr.9: 21967253-21967253 on Build GRCh38
rs772924693 Chr.9: 21967396-21967396 on Build GRCh38
rs144944498 Chr.9: 21967163-21967164 on Build GRCh38
rs75763841 Chr.9: 21967336-21967336 on Build GRCh38
rs746385284 Chr.9: 21967252-21967252 on Build GRCh38
rs753296029 Chr.9: 21967159-21967159 on Build GRCh38
rs372012021 Chr.9: 21967476-21967476 on Build GRCh38
rs964422238 Chr.9: 21967284-21967284 on Build GRCh38
rs914293775 Chr.9: 21967578-21967578 on Build GRCh38
rs201859802 Chr.9: 21967292-21967292 on Build GRCh38
rs386471114 Chr.9: 21967480-21967480 on Build GRCh38
rs762256568 Chr.9: 21967161-21967161 on Build GRCh38
rs990392920 Chr.9: 21967558-21967558 on Build GRCh38
rs115448194 Chr.9: 21967136-21967136 on Build GRCh38
rs915015913 Chr.9: 21967305-21967305 on Build GRCh38
rs778401558 Chr.9: 21967194-21967194 on Build GRCh38
rs758142040 Chr.9: 21967387-21967387 on Build GRCh38
rs920576573 Chr.9: 21967123-21967123 on Build GRCh38
rs550633507 Chr.9: 21967427-21967427 on Build GRCh38
rs75679602 Chr.9: 21967174-21967174 on Build GRCh38
rs758889491 Chr.9: 21967434-21967434 on Build GRCh38
rs749337205 Chr.9: 21967304-21967304 on Build GRCh38
rs972200204 Chr.9: 21967121-21967121 on Build GRCh38
rs576953396 Chr.9: 21967254-21967254 on Build GRCh38
rs777798494 Chr.9: 21967444-21967444 on Build GRCh38
rs747458537 Chr.9: 21967204-21967204 on Build GRCh38
rs370747897 Chr.9: 21967174-21967174 on Build GRCh38
rs766425537 Chr.9: 21967343-21967343 on Build GRCh38
rs945230441 Chr.9: 21967073-21967073 on Build GRCh38
rs1054246310 Chr.9: 21967256-21967256 on Build GRCh38
rs1008988323 Chr.9: 21967094-21967094 on Build GRCh38
rs754426210 Chr.9: 21967471-21967471 on Build GRCh38
rs1019053907 Chr.9: 21967384-21967384 on Build GRCh38
rs555760987 Chr.9: 21967102-21967102 on Build GRCh38
rs953633414 Chr.9: 21967075-21967075 on Build GRCh38
rs762997937 Chr.9: 21967341-21967341 on Build GRCh38
rs761723447 Chr.9: 21967362-21967362 on Build GRCh38
rs991810274 Chr.9: 21967137-21967137 on Build GRCh38
rs1029239623 Chr.9: 21967062-21967062 on Build GRCh38
rs567183809 Chr.9: 21967551-21967551 on Build GRCh38
rs112307232 Chr.9: 21967208-21967208 on Build GRCh38
rs962263132 Chr.9: 21967385-21967385 on Build GRCh38
rs754692641 Chr.9: 21967175-21967175 on Build GRCh38

View Full Product Details