Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 42127715-42127877 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2D6
Entrez Gene ID
1565
Gene Name
cytochrome P450 family 2 subfamily D member 6
Gene Aliases
CPD6, CYP2D, CYP2D7AP, CYP2D7BP, CYP2D7P2, CYP2D8P2, CYP2DL1, CYPIID6, P450-DB1, P450C2D, P450DB1
UniGene
Hs.648256
Gene Chromosome Location
Chr.22: 42125531-42130881 on Build GRCh38
Transcripts
NM_000106.5 NM_001025161.2 XM_011529966.2 XM_011529968.2 XM_011529970.2 XM_011529972.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1641562 c.985C>T p.R329C Chr.22: 42127842-42127842 on Build GRCh38
4104552 c.952C>A p.L318I Chr.22: 42127875-42127875 on Build GRCh38
4104553 c.799C>A p.L267I Chr.22: 42127875-42127875 on Build GRCh38
5596555 c.961A>C p.M321L Chr.22: 42127866-42127866 on Build GRCh38
1641563 c.832C>T p.R278C Chr.22: 42127842-42127842 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746736981 Chr.22: 42127821-42127822 on Build GRCh38
rs267608291 Chr.22: 42127761-42127761 on Build GRCh38
rs148250267 Chr.22: 42127833-42127833 on Build GRCh38
rs113420533 Chr.22: 42127839-42127839 on Build GRCh38
rs777955329 Chr.22: 42127846-42127846 on Build GRCh38
rs150216909 Chr.22: 42127842-42127842 on Build GRCh38
rs28371725 Chr.22: 42127803-42127803 on Build GRCh38
rs767401994 Chr.22: 42127837-42127837 on Build GRCh38
rs201895960 Chr.22: 42127831-42127831 on Build GRCh38
rs369999094 Chr.22: 42127810-42127810 on Build GRCh38
rs550835698 Chr.22: 42127830-42127831 on Build GRCh38
rs28371724 Chr.22: 42127855-42127855 on Build GRCh38
rs372910785 Chr.22: 42127851-42127851 on Build GRCh38
rs749933540 Chr.22: 42127793-42127793 on Build GRCh38
rs539070953 Chr.22: 42127758-42127758 on Build GRCh38
rs192090178 Chr.22: 42127807-42127807 on Build GRCh38
rs72549349 Chr.22: 42127841-42127841 on Build GRCh38
rs373844237 Chr.22: 42127799-42127799 on Build GRCh38
rs780137937 Chr.22: 42127796-42127796 on Build GRCh38
rs747917111 Chr.22: 42127804-42127804 on Build GRCh38
rs369176838 Chr.22: 42127849-42127849 on Build GRCh38
rs140513104 Chr.22: 42127853-42127853 on Build GRCh38
rs911213466 Chr.22: 42127763-42127763 on Build GRCh38
rs746324085 Chr.22: 42127813-42127813 on Build GRCh38
rs754895235 Chr.22: 42127835-42127835 on Build GRCh38
rs760996241 Chr.22: 42127826-42127826 on Build GRCh38
rs765006570 Chr.22: 42127792-42127792 on Build GRCh38
rs187334823 Chr.22: 42127786-42127786 on Build GRCh38
rs953106263 Chr.22: 42127850-42127850 on Build GRCh38
rs1014423327 Chr.22: 42127821-42127821 on Build GRCh38
rs730882170 Chr.22: 42127846-42127864 on Build GRCh38
rs778008354 Chr.22: 42127808-42127808 on Build GRCh38
rs143276168 Chr.22: 42127811-42127811 on Build GRCh38
rs569426051 Chr.22: 42127790-42127790 on Build GRCh38
rs554540676 Chr.22: 42127822-42127822 on Build GRCh38
rs536709258 Chr.22: 42127791-42127791 on Build GRCh38
rs760733463 Chr.22: 42127739-42127739 on Build GRCh38
rs965882094 Chr.22: 42127765-42127765 on Build GRCh38
rs952762122 Chr.22: 42127748-42127748 on Build GRCh38
rs773066646 Chr.22: 42127828-42127828 on Build GRCh38
rs79292917 Chr.22: 42127852-42127852 on Build GRCh38
rs751061716 Chr.22: 42127832-42127832 on Build GRCh38
rs879550940 Chr.22: 42127855-42127856 on Build GRCh38
rs764953443 Chr.22: 42127827-42127827 on Build GRCh38
rs772490933 Chr.22: 42127818-42127818 on Build GRCh38

View Full Product Details