Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 99761943-99762114 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP3A4
Entrez Gene ID
1576
Gene Name
cytochrome P450 family 3 subfamily A member 4
Gene Aliases
CP33, CP34, CYP3A, CYP3A3, CYPIIIA3, CYPIIIA4, HLP, NF-25, P450C3, P450PCN1
UniGene
Hs.728751
Gene Chromosome Location
Chr.7: 99756960-99784188 on Build GRCh38
Transcripts
NM_001202855.2 NM_017460.5 XM_017011780.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6255741 c.1232C>A p.P411H Chr.7: 99762062-99762062 on Build GRCh38
5596559 c.1190C>T p.P397L Chr.7: 99762104-99762104 on Build GRCh38
1093880 c.1236G>T p.E412D Chr.7: 99762058-99762058 on Build GRCh38
2867506 c.1207C>T p.R403C Chr.7: 99762087-99762087 on Build GRCh38
5608220 c.1246C>T p.P416S Chr.7: 99762048-99762048 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768110986 Chr.7: 99762049-99762049 on Build GRCh38
rs935238665 Chr.7: 99762031-99762031 on Build GRCh38
rs1044764678 Chr.7: 99762072-99762072 on Build GRCh38
rs763248134 Chr.7: 99762036-99762036 on Build GRCh38
rs770292659 Chr.7: 99761999-99761999 on Build GRCh38
rs372231982 Chr.7: 99762086-99762086 on Build GRCh38
rs772100275 Chr.7: 99762023-99762023 on Build GRCh38
rs778487895 Chr.7: 99762016-99762016 on Build GRCh38
rs4986909 Chr.7: 99762047-99762047 on Build GRCh38
rs773371203 Chr.7: 99762035-99762035 on Build GRCh38
rs766366164 Chr.7: 99762037-99762037 on Build GRCh38
rs368005287 Chr.7: 99762071-99762071 on Build GRCh38
rs72552797 Chr.7: 99762062-99762062 on Build GRCh38
rs143966082 Chr.7: 99762087-99762087 on Build GRCh38
rs768470122 Chr.7: 99762034-99762034 on Build GRCh38
rs933172203 Chr.7: 99761983-99761983 on Build GRCh38
rs1051580054 Chr.7: 99761984-99761984 on Build GRCh38
rs35723517 Chr.7: 99762032-99762032 on Build GRCh38
rs752108433 Chr.7: 99761998-99761998 on Build GRCh38
rs200710508 Chr.7: 99762088-99762088 on Build GRCh38
rs753292003 Chr.7: 99762060-99762060 on Build GRCh38
rs745786117 Chr.7: 99762020-99762020 on Build GRCh38

View Full Product Details