Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 99766998-99767163 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP3A4
Entrez Gene ID
1576
Gene Name
cytochrome P450 family 3 subfamily A member 4
Gene Aliases
CP33, CP34, CYP3A, CYP3A3, CYPIIIA3, CYPIIIA4, HLP, NF-25, P450C3, P450PCN1
UniGene
Hs.728751
Gene Chromosome Location
Chr.7: 99756960-99784188 on Build GRCh38
Transcripts
NM_001202855.2 NM_017460.5 XM_017011780.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3725379 c.793C>T p.Q265* Chr.7: 99767136-99767136 on Build GRCh38
6639289 c.798G>A p.K266K Chr.7: 99767131-99767131 on Build GRCh38
6271202 c.791C>A p.T264K Chr.7: 99767138-99767138 on Build GRCh38
3721395 c.784G>A p.E262K Chr.7: 99767145-99767145 on Build GRCh38
4139368 c.772G>T p.E258* Chr.7: 99767157-99767157 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777545036 Chr.7: 99767050-99767050 on Build GRCh38
rs760060230 Chr.7: 99767123-99767123 on Build GRCh38
rs771222055 Chr.7: 99767108-99767108 on Build GRCh38
rs749810355 Chr.7: 99767085-99767085 on Build GRCh38
rs775030304 Chr.7: 99767121-99767121 on Build GRCh38
rs763680303 Chr.7: 99767140-99767140 on Build GRCh38
rs894287640 Chr.7: 99767045-99767045 on Build GRCh38
rs778205180 Chr.7: 99767080-99767080 on Build GRCh38
rs774766119 Chr.7: 99767110-99767110 on Build GRCh38
rs371565910 Chr.7: 99767111-99767111 on Build GRCh38
rs577699651 Chr.7: 99767035-99767035 on Build GRCh38
rs936722184 Chr.7: 99767043-99767043 on Build GRCh38
rs527906267 Chr.7: 99767077-99767077 on Build GRCh38
rs41280947 Chr.7: 99767069-99767069 on Build GRCh38
rs58256382 Chr.7: 99767119-99767119 on Build GRCh38
rs12721624 Chr.7: 99767049-99767049 on Build GRCh38
rs751836928 Chr.7: 99767071-99767071 on Build GRCh38
rs556220310 Chr.7: 99767029-99767029 on Build GRCh38
rs1054217767 Chr.7: 99767044-99767044 on Build GRCh38

View Full Product Details