Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 28004082-28004250 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLT3
Entrez Gene ID
2322
Gene Name
fms related tyrosine kinase 3
Gene Aliases
CD135, FLK-2, FLK2, STK1
UniGene
Hs.507590
Gene Chromosome Location
Chr.13: 28003274-28100592 on Build GRCh38
Transcripts
NM_004119.2 NR_130706.1 XM_011535015.2 XM_011535017.2 XM_011535018.2 XM_017020486.1 XM_017020487.1 XM_017020488.1 XM_017020489.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6921163 c.2866C>G p.Q956E Chr.13: 28004168-28004168 on Build GRCh38
6897677 c.2937G>A p.M979I Chr.13: 28004097-28004097 on Build GRCh38
6949394 c.2925C>A p.F975L Chr.13: 28004109-28004109 on Build GRCh38
6926261 c.2896C>T p.P966S Chr.13: 28004138-28004138 on Build GRCh38
7368466 c.2881C>A p.R961S Chr.13: 28004153-28004153 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144397269 Chr.13: 28004116-28004116 on Build GRCh38
rs891244072 Chr.13: 28004193-28004193 on Build GRCh38
rs147467327 Chr.13: 28004144-28004144 on Build GRCh38
rs748134082 Chr.13: 28004161-28004161 on Build GRCh38
rs367817562 Chr.13: 28004170-28004170 on Build GRCh38
rs149473536 Chr.13: 28004153-28004153 on Build GRCh38
rs932783214 Chr.13: 28004183-28004183 on Build GRCh38
rs768405963 Chr.13: 28004223-28004223 on Build GRCh38
rs191501493 Chr.13: 28004117-28004117 on Build GRCh38
rs766953291 Chr.13: 28004141-28004141 on Build GRCh38
rs756072254 Chr.13: 28004175-28004175 on Build GRCh38
rs758631139 Chr.13: 28004168-28004168 on Build GRCh38
rs751604744 Chr.13: 28004172-28004172 on Build GRCh38
rs769921000 Chr.13: 28004194-28004195 on Build GRCh38
rs373674960 Chr.13: 28004152-28004152 on Build GRCh38
rs371259230 Chr.13: 28004135-28004135 on Build GRCh38
rs761305303 Chr.13: 28004140-28004140 on Build GRCh38
rs763135511 Chr.13: 28004108-28004108 on Build GRCh38
rs779912505 Chr.13: 28004180-28004180 on Build GRCh38
rs183293726 Chr.13: 28004145-28004145 on Build GRCh38
rs138003347 Chr.13: 28004146-28004146 on Build GRCh38
rs749089462 Chr.13: 28004210-28004210 on Build GRCh38
rs376024436 Chr.13: 28004189-28004189 on Build GRCh38

View Full Product Details