Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 28015056-28015235 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLT3
Entrez Gene ID
2322
Gene Name
fms related tyrosine kinase 3
Gene Aliases
CD135, FLK-2, FLK2, STK1
UniGene
Hs.507590
Gene Chromosome Location
Chr.13: 28003274-28100592 on Build GRCh38
Transcripts
NM_004119.2 NR_130706.1 XM_011535015.2 XM_011535017.2 XM_011535018.2 XM_017020486.1 XM_017020487.1 XM_017020488.1 XM_017020489.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6920101 c.2677C>A p.P893T Chr.13: 28015233-28015233 on Build GRCh38
29021 c.2654_2753del100 p.G885fs*4 Chr.13: 28015157-28015256 on Build GRCh38
946465 c.2749G>T p.E917* Chr.13: 28015161-28015161 on Build GRCh38
5376583 c.2724G>A p.M908I Chr.13: 28015186-28015186 on Build GRCh38
6665261 c.2678C>T p.P893L Chr.13: 28015232-28015232 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs184067612 Chr.13: 28015089-28015089 on Build GRCh38
rs766718234 Chr.13: 28015093-28015093 on Build GRCh38
rs968225958 Chr.13: 28015090-28015090 on Build GRCh38
rs200909894 Chr.13: 28015188-28015188 on Build GRCh38
rs201504848 Chr.13: 28015185-28015185 on Build GRCh38
rs373997542 Chr.13: 28015156-28015156 on Build GRCh38
rs776003598 Chr.13: 28015127-28015127 on Build GRCh38
rs565608482 Chr.13: 28015080-28015080 on Build GRCh38
rs764027375 Chr.13: 28015211-28015211 on Build GRCh38
rs760751139 Chr.13: 28015177-28015177 on Build GRCh38
rs372671564 Chr.13: 28015142-28015142 on Build GRCh38
rs539980970 Chr.13: 28015199-28015199 on Build GRCh38
rs770221306 Chr.13: 28015116-28015116 on Build GRCh38
rs980046447 Chr.13: 28015102-28015102 on Build GRCh38
rs780604821 Chr.13: 28015115-28015115 on Build GRCh38
rs949765528 Chr.13: 28015149-28015149 on Build GRCh38
rs760485979 Chr.13: 28015111-28015112 on Build GRCh38
rs369237950 Chr.13: 28015150-28015150 on Build GRCh38
rs775314249 Chr.13: 28015106-28015107 on Build GRCh38
rs4073630 Chr.13: 28015130-28015130 on Build GRCh38
rs751881381 Chr.13: 28015120-28015120 on Build GRCh38
rs752745594 Chr.13: 28015198-28015198 on Build GRCh38
rs369244249 Chr.13: 28015137-28015137 on Build GRCh38
rs1032753793 Chr.13: 28015113-28015113 on Build GRCh38
rs773189409 Chr.13: 28015163-28015163 on Build GRCh38

View Full Product Details