Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 69462807-69463353 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AAK1
Entrez Gene ID
22848
Gene Name
AP2 associated kinase 1
Gene Aliases
-
UniGene
Hs.468878
Gene Chromosome Location
Chr.2: 69457995-69643845 on Build GRCh38
Transcripts
NM_014911.3

SNP Detail

SNP ID Chromosome Location
rs1039128081 Chr.2: 69462954-69462954 on Build GRCh38
rs931197227 Chr.2: 69463138-69463138 on Build GRCh38
rs909180160 Chr.2: 69463171-69463171 on Build GRCh38
rs759156956 Chr.2: 69463305-69463305 on Build GRCh38
rs377075795 Chr.2: 69463321-69463323 on Build GRCh38
rs930731569 Chr.2: 69463019-69463019 on Build GRCh38
rs943386571 Chr.2: 69463173-69463173 on Build GRCh38
rs1037352413 Chr.2: 69462905-69462905 on Build GRCh38
rs371295912 Chr.2: 69462937-69462937 on Build GRCh38
rs749793935 Chr.2: 69463035-69463035 on Build GRCh38
rs1021606878 Chr.2: 69463013-69463013 on Build GRCh38
rs924962909 Chr.2: 69462874-69462874 on Build GRCh38
rs990531102 Chr.2: 69462892-69462892 on Build GRCh38
rs564331594 Chr.2: 69463125-69463125 on Build GRCh38
rs1032589309 Chr.2: 69463257-69463257 on Build GRCh38
rs774793050 Chr.2: 69463216-69463216 on Build GRCh38
rs533259899 Chr.2: 69463126-69463126 on Build GRCh38
rs917712254 Chr.2: 69463081-69463081 on Build GRCh38
rs937737829 Chr.2: 69462879-69462879 on Build GRCh38
rs191706328 Chr.2: 69463228-69463228 on Build GRCh38
rs752738252 Chr.2: 69463322-69463322 on Build GRCh38
rs769846421 Chr.2: 69462898-69462898 on Build GRCh38
rs959028057 Chr.2: 69463262-69463262 on Build GRCh38
rs13427243 Chr.2: 69463055-69463055 on Build GRCh38
rs1011937042 Chr.2: 69463275-69463275 on Build GRCh38
rs544648321 Chr.2: 69463230-69463230 on Build GRCh38
rs997734760 Chr.2: 69462993-69462993 on Build GRCh38
rs897091735 Chr.2: 69462908-69462908 on Build GRCh38
rs986058989 Chr.2: 69463213-69463213 on Build GRCh38
rs1031928608 Chr.2: 69463036-69463036 on Build GRCh38
rs929276412 Chr.2: 69462920-69462920 on Build GRCh38
rs776021395 Chr.2: 69462951-69462951 on Build GRCh38
rs1010691738 Chr.2: 69463132-69463132 on Build GRCh38
rs527452754 Chr.2: 69463064-69463064 on Build GRCh38
rs966962091 Chr.2: 69463029-69463029 on Build GRCh38
rs944565436 Chr.2: 69462958-69462958 on Build GRCh38
rs949279779 Chr.2: 69462904-69462904 on Build GRCh38
rs149400104 Chr.2: 69462855-69462855 on Build GRCh38
rs887799044 Chr.2: 69462940-69462940 on Build GRCh38
rs59609954 Chr.2: 69463252-69463252 on Build GRCh38
rs764834923 Chr.2: 69463308-69463308 on Build GRCh38
rs776457139 Chr.2: 69463251-69463251 on Build GRCh38
rs970946876 Chr.2: 69463118-69463118 on Build GRCh38
rs1049064600 Chr.2: 69462939-69462939 on Build GRCh38
rs892193906 Chr.2: 69463129-69463129 on Build GRCh38
rs901950578 Chr.2: 69463226-69463226 on Build GRCh38
rs902001836 Chr.2: 69462965-69462965 on Build GRCh38
rs919319522 Chr.2: 69462985-69462985 on Build GRCh38
rs745978422 Chr.2: 69462844-69462844 on Build GRCh38
rs986491482 Chr.2: 69462945-69462945 on Build GRCh38
rs147521618 Chr.2: 69463065-69463065 on Build GRCh38
rs542351238 Chr.2: 69463046-69463046 on Build GRCh38
rs1048346695 Chr.2: 69463151-69463151 on Build GRCh38
rs979090553 Chr.2: 69462863-69462863 on Build GRCh38
rs902692081 Chr.2: 69463246-69463246 on Build GRCh38

View Full Product Details