Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 6294766-6295261 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOT7
Entrez Gene ID
11332
Gene Name
acyl-CoA thioesterase 7
Gene Aliases
ACH1, ACT, BACH, CTE-II, LACH, LACH1, hBACH
UniGene
Hs.126137
Gene Chromosome Location
Chr.1: 6264272-6393766 on Build GRCh38
Transcripts
NM_007274.3 NM_181864.2 NM_181865.2 NM_181866.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1584659 c.704A>G p.H235R Chr.1: 6294929-6294929 on Build GRCh38
1584660 c.641A>G p.H214R Chr.1: 6294929-6294929 on Build GRCh38
2238604 c.810T>C p.F270F Chr.1: 6294883-6294883 on Build GRCh38
308823 c.761G>T p.R254L Chr.1: 6294932-6294932 on Build GRCh38
1646195 c.742A>G p.I248V Chr.1: 6294891-6294891 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754361531 Chr.1: 6295127-6295127 on Build GRCh38
rs767355127 Chr.1: 6294838-6294838 on Build GRCh38
rs1053860348 Chr.1: 6295188-6295188 on Build GRCh38
rs751746256 Chr.1: 6294998-6294998 on Build GRCh38
rs757573621 Chr.1: 6294906-6294906 on Build GRCh38
rs375152254 Chr.1: 6294834-6294834 on Build GRCh38
rs948370728 Chr.1: 6295153-6295153 on Build GRCh38
rs933556957 Chr.1: 6295044-6295044 on Build GRCh38
rs56235663 Chr.1: 6295006-6295006 on Build GRCh38
rs763302103 Chr.1: 6294991-6294991 on Build GRCh38
rs773999418 Chr.1: 6294831-6294831 on Build GRCh38
rs775307423 Chr.1: 6294820-6294820 on Build GRCh38
rs1047326334 Chr.1: 6295120-6295120 on Build GRCh38
rs925454497 Chr.1: 6294809-6294809 on Build GRCh38
rs35553413 Chr.1: 6295005-6295005 on Build GRCh38
rs929413145 Chr.1: 6295114-6295114 on Build GRCh38
rs193157697 Chr.1: 6295058-6295058 on Build GRCh38
rs1024820835 Chr.1: 6295129-6295129 on Build GRCh38
rs995722072 Chr.1: 6294909-6294909 on Build GRCh38
rs199792713 Chr.1: 6294840-6294840 on Build GRCh38
rs755713548 Chr.1: 6294915-6294915 on Build GRCh38
rs377671061 Chr.1: 6294850-6294850 on Build GRCh38
rs917013415 Chr.1: 6295151-6295151 on Build GRCh38
rs367613034 Chr.1: 6295014-6295014 on Build GRCh38
rs116056234 Chr.1: 6295100-6295100 on Build GRCh38
rs910503389 Chr.1: 6294811-6294811 on Build GRCh38
rs1051840042 Chr.1: 6295228-6295228 on Build GRCh38
rs1001786825 Chr.1: 6295174-6295175 on Build GRCh38
rs764569398 Chr.1: 6294889-6294889 on Build GRCh38
rs771888331 Chr.1: 6295027-6295027 on Build GRCh38
rs781767639 Chr.1: 6294912-6294912 on Build GRCh38
rs914104988 Chr.1: 6294824-6294824 on Build GRCh38
rs201187410 Chr.1: 6294932-6294932 on Build GRCh38
rs768540257 Chr.1: 6294828-6294828 on Build GRCh38
rs376311179 Chr.1: 6295000-6295000 on Build GRCh38
rs563333886 Chr.1: 6294955-6294955 on Build GRCh38
rs867443511 Chr.1: 6294958-6294958 on Build GRCh38
rs774272575 Chr.1: 6294949-6294949 on Build GRCh38
rs752950129 Chr.1: 6294854-6294854 on Build GRCh38
rs372156502 Chr.1: 6295001-6295001 on Build GRCh38
rs1010180540 Chr.1: 6295216-6295216 on Build GRCh38
rs767792777 Chr.1: 6294851-6294851 on Build GRCh38
rs1017532266 Chr.1: 6295103-6295103 on Build GRCh38
rs1030719350 Chr.1: 6295082-6295082 on Build GRCh38
rs114381815 Chr.1: 6295126-6295126 on Build GRCh38
rs747904502 Chr.1: 6294952-6294952 on Build GRCh38
rs868517540 Chr.1: 6295060-6295060 on Build GRCh38
rs374912644 Chr.1: 6295020-6295020 on Build GRCh38
rs148303638 Chr.1: 6294913-6294913 on Build GRCh38
rs759627829 Chr.1: 6294841-6294841 on Build GRCh38
rs758517942 Chr.1: 6294860-6294860 on Build GRCh38
rs370938238 Chr.1: 6294985-6294985 on Build GRCh38
rs1804843 Chr.1: 6294895-6294895 on Build GRCh38
rs886989583 Chr.1: 6294925-6294925 on Build GRCh38
rs926843640 Chr.1: 6295182-6295182 on Build GRCh38
rs987947098 Chr.1: 6295240-6295240 on Build GRCh38
rs772269320 Chr.1: 6294943-6294943 on Build GRCh38
rs768176924 Chr.1: 6294937-6294937 on Build GRCh38
rs910541390 Chr.1: 6295095-6295095 on Build GRCh38
rs1044079363 Chr.1: 6295155-6295155 on Build GRCh38
rs963325935 Chr.1: 6295109-6295109 on Build GRCh38
rs762657749 Chr.1: 6294822-6294822 on Build GRCh38
rs779677519 Chr.1: 6294920-6294920 on Build GRCh38
rs78203431 Chr.1: 6295205-6295205 on Build GRCh38
rs1032484612 Chr.1: 6295012-6295012 on Build GRCh38
rs529601305 Chr.1: 6295125-6295125 on Build GRCh38
rs1037817919 Chr.1: 6294856-6294856 on Build GRCh38
rs371712843 Chr.1: 6295016-6295016 on Build GRCh38
rs386628059 Chr.1: 6295005-6295006 on Build GRCh38
rs954722531 Chr.1: 6295087-6295087 on Build GRCh38
rs769040966 Chr.1: 6294984-6294984 on Build GRCh38
rs905137696 Chr.1: 6295149-6295149 on Build GRCh38
rs376223601 Chr.1: 6295015-6295015 on Build GRCh38
rs760567789 Chr.1: 6294983-6294983 on Build GRCh38
rs764226514 Chr.1: 6294997-6294997 on Build GRCh38
rs773157928 Chr.1: 6294971-6294971 on Build GRCh38
rs750629180 Chr.1: 6294990-6294991 on Build GRCh38
rs933561831 Chr.1: 6294825-6294825 on Build GRCh38
rs377658886 Chr.1: 6295121-6295121 on Build GRCh38
rs936927808 Chr.1: 6295185-6295185 on Build GRCh38

View Full Product Details