Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136673619-136674115 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGPAT2
Entrez Gene ID
10555
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 2
Gene Aliases
1-AGPAT2, BSCL, BSCL1, LPAAB, LPAAT-beta
UniGene
Hs.320151
Gene Chromosome Location
Chr.9: 136673143-136687459 on Build GRCh38
Transcripts
NM_001012727.1 NM_006412.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4655451 c.802G>A p.A268T Chr.9: 136673787-136673787 on Build GRCh38
6244346 c.748C>A p.R250R Chr.9: 136673841-136673841 on Build GRCh38
6051510 c.728C>T p.A243V Chr.9: 136673861-136673861 on Build GRCh38
6394473 c.712G>A p.A238T Chr.9: 136673877-136673877 on Build GRCh38
7119384 c.779C>G p.S260C Chr.9: 136673810-136673810 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1011963916 Chr.9: 136673657-136673657 on Build GRCh38
rs1040117955 Chr.9: 136673682-136673682 on Build GRCh38
rs368090654 Chr.9: 136673840-136673840 on Build GRCh38
rs778853584 Chr.9: 136673856-136673856 on Build GRCh38
rs758501299 Chr.9: 136673860-136673860 on Build GRCh38
rs765479536 Chr.9: 136673918-136673918 on Build GRCh38
rs372565832 Chr.9: 136674023-136674023 on Build GRCh38
rs766362929 Chr.9: 136673883-136673883 on Build GRCh38
rs764844105 Chr.9: 136673779-136673779 on Build GRCh38
rs368352508 Chr.9: 136673721-136673721 on Build GRCh38
rs766391983 Chr.9: 136673845-136673845 on Build GRCh38
rs146973799 Chr.9: 136673827-136673827 on Build GRCh38
rs769602973 Chr.9: 136673707-136673707 on Build GRCh38
rs1009735594 Chr.9: 136673999-136673999 on Build GRCh38
rs777675719 Chr.9: 136673714-136673714 on Build GRCh38
rs781637963 Chr.9: 136673914-136673914 on Build GRCh38
rs104894100 Chr.9: 136673906-136673906 on Build GRCh38
rs373878123 Chr.9: 136673799-136673799 on Build GRCh38
rs368902934 Chr.9: 136673769-136673769 on Build GRCh38
rs764643486 Chr.9: 136673821-136673821 on Build GRCh38
rs759149037 Chr.9: 136673776-136673776 on Build GRCh38
rs752499956 Chr.9: 136673712-136673712 on Build GRCh38
rs1026042809 Chr.9: 136674045-136674045 on Build GRCh38
rs530362355 Chr.9: 136674046-136674046 on Build GRCh38
rs553740204 Chr.9: 136673861-136673861 on Build GRCh38
rs147616533 Chr.9: 136673828-136673828 on Build GRCh38
rs758668652 Chr.9: 136673946-136673947 on Build GRCh38
rs577724972 Chr.9: 136673677-136673677 on Build GRCh38
rs373329785 Chr.9: 136673770-136673770 on Build GRCh38
rs768585710 Chr.9: 136673800-136673800 on Build GRCh38
rs773653710 Chr.9: 136673804-136673804 on Build GRCh38
rs767338891 Chr.9: 136673841-136673841 on Build GRCh38
rs545016569 Chr.9: 136674012-136674012 on Build GRCh38
rs777428971 Chr.9: 136673788-136673788 on Build GRCh38
rs370584114 Chr.9: 136673938-136673938 on Build GRCh38
rs116951119 Chr.9: 136673887-136673887 on Build GRCh38
rs757129159 Chr.9: 136673684-136673684 on Build GRCh38
rs982359721 Chr.9: 136673644-136673644 on Build GRCh38
rs144522710 Chr.9: 136673650-136673650 on Build GRCh38
rs768871491 Chr.9: 136673735-136673735 on Build GRCh38
rs760549983 Chr.9: 136673971-136673971 on Build GRCh38
rs139145747 Chr.9: 136674076-136674077 on Build GRCh38
rs751303704 Chr.9: 136673835-136673835 on Build GRCh38
rs561521361 Chr.9: 136674042-136674042 on Build GRCh38
rs948561406 Chr.9: 136674072-136674072 on Build GRCh38
rs908505806 Chr.9: 136674051-136674051 on Build GRCh38
rs200614462 Chr.9: 136673704-136673704 on Build GRCh38
rs1012838445 Chr.9: 136674017-136674017 on Build GRCh38
rs751384599 Chr.9: 136674069-136674069 on Build GRCh38
rs964300549 Chr.9: 136674055-136674055 on Build GRCh38
rs1035556218 Chr.9: 136673685-136673685 on Build GRCh38
rs372910163 Chr.9: 136673741-136673741 on Build GRCh38
rs547267027 Chr.9: 136674074-136674074 on Build GRCh38
rs1003229436 Chr.9: 136673844-136673844 on Build GRCh38
rs763693103 Chr.9: 136673872-136673872 on Build GRCh38
rs145975461 Chr.9: 136673873-136673873 on Build GRCh38
rs748586893 Chr.9: 136673927-136673927 on Build GRCh38
rs1057084115 Chr.9: 136673996-136673996 on Build GRCh38
rs536777709 Chr.9: 136673891-136673891 on Build GRCh38
rs933915348 Chr.9: 136673701-136673701 on Build GRCh38
rs749380856 Chr.9: 136673731-136673731 on Build GRCh38
rs967659798 Chr.9: 136673672-136673672 on Build GRCh38
rs117434864 Chr.9: 136673848-136673848 on Build GRCh38
rs753404041 Chr.9: 136673706-136673706 on Build GRCh38
rs1028407116 Chr.9: 136674039-136674039 on Build GRCh38
rs777195971 Chr.9: 136673865-136673865 on Build GRCh38
rs544283953 Chr.9: 136674049-136674049 on Build GRCh38
rs572859264 Chr.9: 136673952-136673952 on Build GRCh38
rs938672204 Chr.9: 136673982-136673982 on Build GRCh38
rs375808894 Chr.9: 136673742-136673742 on Build GRCh38
rs550828083 Chr.9: 136674030-136674030 on Build GRCh38
rs781126746 Chr.9: 136673794-136673794 on Build GRCh38
rs914738037 Chr.9: 136673920-136673920 on Build GRCh38
rs200656731 Chr.9: 136673876-136673876 on Build GRCh38
rs200073215 Chr.9: 136673922-136673922 on Build GRCh38
rs367550612 Chr.9: 136673936-136673936 on Build GRCh38
rs557103117 Chr.9: 136673915-136673915 on Build GRCh38
rs898986047 Chr.9: 136674026-136674026 on Build GRCh38
rs372240340 Chr.9: 136673842-136673842 on Build GRCh38
rs749592042 Chr.9: 136673718-136673718 on Build GRCh38
rs142207711 Chr.9: 136673869-136673869 on Build GRCh38
rs1039745793 Chr.9: 136673649-136673649 on Build GRCh38
rs17848858 Chr.9: 136673801-136673801 on Build GRCh38
rs370950858 Chr.9: 136673875-136673875 on Build GRCh38
rs1057517655 Chr.9: 136673913-136673913 on Build GRCh38
rs530977763 Chr.9: 136673807-136673807 on Build GRCh38
rs753350881 Chr.9: 136673771-136673771 on Build GRCh38
rs968556261 Chr.9: 136673809-136673809 on Build GRCh38
rs756733036 Chr.9: 136673703-136673703 on Build GRCh38
rs370809970 Chr.9: 136673791-136673791 on Build GRCh38
rs755464331 Chr.9: 136673705-136673706 on Build GRCh38
rs199964729 Chr.9: 136673932-136673932 on Build GRCh38
rs958545912 Chr.9: 136673849-136673849 on Build GRCh38
rs758100175 Chr.9: 136673713-136673713 on Build GRCh38
rs762542535 Chr.9: 136673870-136673870 on Build GRCh38
rs1022433204 Chr.9: 136674015-136674015 on Build GRCh38
rs773769888 Chr.9: 136673762-136673762 on Build GRCh38
rs760141111 Chr.9: 136673810-136673810 on Build GRCh38
rs773906576 Chr.9: 136673935-136673935 on Build GRCh38
rs528513856 Chr.9: 136673786-136673786 on Build GRCh38
rs761143874 Chr.9: 136673806-136673806 on Build GRCh38
rs142417583 Chr.9: 136673780-136673780 on Build GRCh38
rs151215253 Chr.9: 136673803-136673803 on Build GRCh38
rs1041789611 Chr.9: 136673947-136673947 on Build GRCh38
rs112657922 Chr.9: 136673673-136673673 on Build GRCh38
rs572102781 Chr.9: 136673639-136673639 on Build GRCh38
rs777248850 Chr.9: 136673808-136673808 on Build GRCh38
rs772824364 Chr.9: 136673970-136673970 on Build GRCh38
rs768553437 Chr.9: 136673878-136673878 on Build GRCh38
rs1019828485 Chr.9: 136674054-136674054 on Build GRCh38
rs1011578726 Chr.9: 136673813-136673813 on Build GRCh38
rs779176871 Chr.9: 136673740-136673740 on Build GRCh38
rs1020207560 Chr.9: 136673664-136673664 on Build GRCh38
rs11545228 Chr.9: 136673868-136673868 on Build GRCh38
rs747223219 Chr.9: 136673897-136673897 on Build GRCh38
rs1051343001 Chr.9: 136673774-136673774 on Build GRCh38
rs138232867 Chr.9: 136673886-136673886 on Build GRCh38
rs1057517656 Chr.9: 136673826-136673834 on Build GRCh38
rs200288462 Chr.9: 136673857-136673857 on Build GRCh38
rs576838239 Chr.9: 136673683-136673683 on Build GRCh38
rs919781270 Chr.9: 136674070-136674070 on Build GRCh38
rs751365063 Chr.9: 136673790-136673790 on Build GRCh38
rs141456385 Chr.9: 136673825-136673825 on Build GRCh38
rs975626633 Chr.9: 136674066-136674066 on Build GRCh38
rs894360943 Chr.9: 136674000-136674000 on Build GRCh38
rs757623351 Chr.9: 136673910-136673910 on Build GRCh38
rs139800244 Chr.9: 136674028-136674028 on Build GRCh38
rs201868056 Chr.9: 136673729-136673729 on Build GRCh38
rs763816384 Chr.9: 136673831-136673831 on Build GRCh38

View Full Product Details