Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 427704-428211 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AHRR
Entrez Gene ID
57491
Gene Name
aryl-hydrocarbon receptor repressor
Gene Aliases
AHH, AHHR, bHLHe77
UniGene
Hs.50823
Gene Chromosome Location
Chr.5: 304176-438291 on Build GRCh38
Transcripts
NM_001242412.1 NM_020731.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4512139 c.892C>T p.P298S Chr.5: 427924-427924 on Build GRCh38
373669 c.896C>G p.S299C Chr.5: 427928-427928 on Build GRCh38
3342510 c.815_817delAGA p.K275delK Chr.5: 427847-427849 on Build GRCh38
3342514 c.847C>T p.P283S Chr.5: 427879-427879 on Build GRCh38
3342519 c.848C>T p.P283L Chr.5: 427880-427880 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759100906 Chr.5: 427778-427778 on Build GRCh38
rs12188164 Chr.5: 428121-428121 on Build GRCh38
rs776042722 Chr.5: 427956-427956 on Build GRCh38
rs768799918 Chr.5: 427830-427830 on Build GRCh38
rs963825697 Chr.5: 427842-427842 on Build GRCh38
rs202117783 Chr.5: 427862-427862 on Build GRCh38
rs531824368 Chr.5: 428050-428050 on Build GRCh38
rs773693929 Chr.5: 427855-427855 on Build GRCh38
rs752948258 Chr.5: 427995-427995 on Build GRCh38
rs780325148 Chr.5: 427815-427815 on Build GRCh38
rs758853237 Chr.5: 428028-428028 on Build GRCh38
rs376844014 Chr.5: 427726-427726 on Build GRCh38
rs980887728 Chr.5: 428076-428076 on Build GRCh38
rs779297309 Chr.5: 428023-428023 on Build GRCh38
rs41282623 Chr.5: 427727-427727 on Build GRCh38
rs775291415 Chr.5: 427951-427951 on Build GRCh38
rs763636649 Chr.5: 427908-427908 on Build GRCh38
rs913869524 Chr.5: 428090-428090 on Build GRCh38
rs527453593 Chr.5: 427997-427997 on Build GRCh38
rs370682493 Chr.5: 427809-427809 on Build GRCh38
rs1010599195 Chr.5: 427752-427752 on Build GRCh38
rs762740242 Chr.5: 427987-427987 on Build GRCh38
rs926822427 Chr.5: 428078-428078 on Build GRCh38
rs72717408 Chr.5: 427929-427929 on Build GRCh38
rs769858803 Chr.5: 427884-427884 on Build GRCh38
rs771358248 Chr.5: 427892-427892 on Build GRCh38
rs1042033176 Chr.5: 427949-427949 on Build GRCh38
rs768549270 Chr.5: 427724-427724 on Build GRCh38
rs756164524 Chr.5: 427782-427782 on Build GRCh38
rs926770927 Chr.5: 428073-428074 on Build GRCh38
rs1050036244 Chr.5: 428169-428169 on Build GRCh38
rs779465009 Chr.5: 427935-427935 on Build GRCh38
rs766113974 Chr.5: 427991-427991 on Build GRCh38
rs753746552 Chr.5: 427808-427808 on Build GRCh38
rs764954357 Chr.5: 427738-427738 on Build GRCh38
rs917934395 Chr.5: 428108-428108 on Build GRCh38
rs995740128 Chr.5: 427967-427967 on Build GRCh38
rs753647962 Chr.5: 427873-427873 on Build GRCh38
rs542713317 Chr.5: 427998-427998 on Build GRCh38
rs945360710 Chr.5: 428091-428091 on Build GRCh38
rs748194882 Chr.5: 427843-427843 on Build GRCh38
rs12188678 Chr.5: 428116-428116 on Build GRCh38
rs746104566 Chr.5: 427947-427949 on Build GRCh38
rs760251828 Chr.5: 427866-427866 on Build GRCh38
rs757356238 Chr.5: 428005-428005 on Build GRCh38
rs373499533 Chr.5: 427859-427859 on Build GRCh38
rs566262422 Chr.5: 427868-427868 on Build GRCh38
rs747676922 Chr.5: 428052-428052 on Build GRCh38
rs539120226 Chr.5: 428135-428135 on Build GRCh38
rs199509403 Chr.5: 427930-427930 on Build GRCh38
rs758219910 Chr.5: 427771-427771 on Build GRCh38
rs889600867 Chr.5: 428176-428176 on Build GRCh38
rs763381911 Chr.5: 427856-427856 on Build GRCh38
rs898363581 Chr.5: 427958-427958 on Build GRCh38
rs746290276 Chr.5: 427936-427936 on Build GRCh38
rs773606194 Chr.5: 427730-427730 on Build GRCh38
rs750233101 Chr.5: 427916-427916 on Build GRCh38
rs199811656 Chr.5: 428057-428057 on Build GRCh38
rs548076463 Chr.5: 428133-428133 on Build GRCh38
rs756370466 Chr.5: 427996-427996 on Build GRCh38
rs970993862 Chr.5: 428075-428075 on Build GRCh38
rs912194668 Chr.5: 427879-427879 on Build GRCh38
rs749358870 Chr.5: 427931-427931 on Build GRCh38
rs781205104 Chr.5: 427835-427835 on Build GRCh38
rs1053801266 Chr.5: 428083-428083 on Build GRCh38
rs748282971 Chr.5: 427883-427883 on Build GRCh38
rs774074829 Chr.5: 427860-427860 on Build GRCh38
rs541442479 Chr.5: 427978-427978 on Build GRCh38
rs746323505 Chr.5: 428025-428025 on Build GRCh38
rs757086268 Chr.5: 427876-427876 on Build GRCh38
rs746988770 Chr.5: 427966-427966 on Build GRCh38
rs537097339 Chr.5: 427926-427926 on Build GRCh38
rs372726854 Chr.5: 427954-427954 on Build GRCh38
rs995266107 Chr.5: 428020-428020 on Build GRCh38
rs201569519 Chr.5: 427934-427934 on Build GRCh38
rs765144182 Chr.5: 427986-427986 on Build GRCh38
rs77520050 Chr.5: 427867-427867 on Build GRCh38
rs148618879 Chr.5: 428178-428178 on Build GRCh38
rs892476304 Chr.5: 427920-427920 on Build GRCh38
rs187824311 Chr.5: 428035-428035 on Build GRCh38
rs377503046 Chr.5: 427779-427779 on Build GRCh38
rs373769011 Chr.5: 427903-427903 on Build GRCh38
rs763203978 Chr.5: 427740-427740 on Build GRCh38
rs770334938 Chr.5: 427851-427851 on Build GRCh38
rs772501854 Chr.5: 427950-427950 on Build GRCh38
rs747140664 Chr.5: 427828-427828 on Build GRCh38
rs183151904 Chr.5: 427907-427907 on Build GRCh38
rs554434575 Chr.5: 427864-427864 on Build GRCh38
rs1028281257 Chr.5: 427982-427982 on Build GRCh38
rs768238670 Chr.5: 428049-428049 on Build GRCh38
rs370055582 Chr.5: 427896-427896 on Build GRCh38
rs751106807 Chr.5: 427769-427769 on Build GRCh38
rs778268200 Chr.5: 427885-427885 on Build GRCh38
rs761363306 Chr.5: 427962-427962 on Build GRCh38
rs771912601 Chr.5: 427902-427902 on Build GRCh38
rs367633421 Chr.5: 427893-427893 on Build GRCh38
rs371473301 Chr.5: 427863-427863 on Build GRCh38
rs766111369 Chr.5: 427760-427760 on Build GRCh38
rs749711222 Chr.5: 427886-427886 on Build GRCh38
rs1028645018 Chr.5: 427784-427784 on Build GRCh38
rs752420869 Chr.5: 427748-427748 on Build GRCh38
rs761611426 Chr.5: 427914-427914 on Build GRCh38
rs758176713 Chr.5: 427921-427921 on Build GRCh38
rs373140041 Chr.5: 427725-427725 on Build GRCh38
rs780878156 Chr.5: 427918-427920 on Build GRCh38
rs143087196 Chr.5: 428052-428052 on Build GRCh38
rs533891130 Chr.5: 427880-427880 on Build GRCh38
rs374703194 Chr.5: 427878-427878 on Build GRCh38
rs776651339 Chr.5: 427909-427909 on Build GRCh38
rs112763035 Chr.5: 428081-428081 on Build GRCh38
rs768231215 Chr.5: 427847-427849 on Build GRCh38
rs781369657 Chr.5: 427881-427881 on Build GRCh38
rs201183475 Chr.5: 427955-427955 on Build GRCh38
rs539340416 Chr.5: 427803-427803 on Build GRCh38
rs918156915 Chr.5: 428065-428065 on Build GRCh38
rs1019305893 Chr.5: 428019-428019 on Build GRCh38
rs376456003 Chr.5: 427915-427915 on Build GRCh38
rs747343793 Chr.5: 428048-428048 on Build GRCh38

View Full Product Details