Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 26852440-26852941 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AQP4
Entrez Gene ID
361
Gene Name
aquaporin 4
Gene Aliases
MIWC, WCH4
UniGene
Hs.315369
Gene Chromosome Location
Chr.18: 26852038-26865844 on Build GRCh38
Transcripts
NM_001317384.1 NM_001317387.1 NM_001650.5 NM_004028.4 XM_011525942.2

SNP Detail

SNP ID Chromosome Location
rs537164829 Chr.18: 26852907-26852907 on Build GRCh38
rs892077740 Chr.18: 26852705-26852705 on Build GRCh38
rs547302553 Chr.18: 26852887-26852887 on Build GRCh38
rs916984111 Chr.18: 26852673-26852673 on Build GRCh38
rs949736279 Chr.18: 26852749-26852749 on Build GRCh38
rs988607249 Chr.18: 26852667-26852667 on Build GRCh38
rs749589092 Chr.18: 26852547-26852547 on Build GRCh38
rs1045084391 Chr.18: 26852621-26852621 on Build GRCh38
rs1057390047 Chr.18: 26852697-26852697 on Build GRCh38
rs769022626 Chr.18: 26852536-26852536 on Build GRCh38
rs774506954 Chr.18: 26852647-26852647 on Build GRCh38
rs1006845270 Chr.18: 26852488-26852488 on Build GRCh38
rs904584955 Chr.18: 26852819-26852819 on Build GRCh38
rs909594226 Chr.18: 26852565-26852565 on Build GRCh38
rs572116150 Chr.18: 26852473-26852473 on Build GRCh38
rs543686299 Chr.18: 26852902-26852902 on Build GRCh38
rs983947953 Chr.18: 26852556-26852556 on Build GRCh38
rs117140070 Chr.18: 26852643-26852643 on Build GRCh38
rs946185321 Chr.18: 26852809-26852809 on Build GRCh38
rs539511786 Chr.18: 26852629-26852629 on Build GRCh38
rs761734162 Chr.18: 26852817-26852817 on Build GRCh38
rs1051164329 Chr.18: 26852908-26852908 on Build GRCh38
rs901166917 Chr.18: 26852900-26852900 on Build GRCh38
rs562393403 Chr.18: 26852903-26852903 on Build GRCh38
rs186735087 Chr.18: 26852553-26852553 on Build GRCh38
rs7240333 Chr.18: 26852848-26852848 on Build GRCh38
rs947848025 Chr.18: 26852607-26852607 on Build GRCh38
rs951199296 Chr.18: 26852523-26852523 on Build GRCh38
rs370330703 Chr.18: 26852613-26852613 on Build GRCh38
rs918538482 Chr.18: 26852483-26852483 on Build GRCh38
rs1038958914 Chr.18: 26852896-26852896 on Build GRCh38
rs865869507 Chr.18: 26852519-26852519 on Build GRCh38

View Full Product Details