Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 235175142-235175661 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARID4B
Entrez Gene ID
51742
Gene Name
AT-rich interaction domain 4B
Gene Aliases
BCAA, BRCAA1, RBBP1L1, RBP1L1, SAP180
UniGene
Hs.575782
Gene Chromosome Location
Chr.1: 235166895-235328240 on Build GRCh38
Transcripts
NM_001206794.1 NM_016374.5 NM_031371.3 NR_135074.1 XM_006711781.2 XM_011544212.2 XM_017001468.1 XM_017001469.1 XM_017001470.1 XM_017001472.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1689989 c.3560G>A p.R1187K Chr.1: 235175288-235175288 on Build GRCh38
3485514 c.3664+1G>T p.? Chr.1: 235175183-235175183 on Build GRCh38
905802 c.3491T>C p.I1164T Chr.1: 235175357-235175357 on Build GRCh38
3789667 c.3595G>A p.D1199N Chr.1: 235175253-235175253 on Build GRCh38
3485515 c.3612C>T p.L1204L Chr.1: 235175236-235175236 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775654862 Chr.1: 235175342-235175342 on Build GRCh38
rs542985562 Chr.1: 235175547-235175547 on Build GRCh38
rs202117320 Chr.1: 235175370-235175370 on Build GRCh38
rs770121410 Chr.1: 235175225-235175225 on Build GRCh38
rs140515034 Chr.1: 235175285-235175285 on Build GRCh38
rs1054964680 Chr.1: 235175505-235175505 on Build GRCh38
rs976668498 Chr.1: 235175589-235175589 on Build GRCh38
rs202186650 Chr.1: 235175202-235175202 on Build GRCh38
rs899275376 Chr.1: 235175579-235175579 on Build GRCh38
rs761103715 Chr.1: 235175345-235175345 on Build GRCh38
rs966306081 Chr.1: 235175587-235175587 on Build GRCh38
rs750111741 Chr.1: 235175405-235175405 on Build GRCh38
rs776617258 Chr.1: 235175217-235175217 on Build GRCh38
rs755970591 Chr.1: 235175444-235175444 on Build GRCh38
rs756841820 Chr.1: 235175252-235175252 on Build GRCh38
rs537264192 Chr.1: 235175236-235175236 on Build GRCh38
rs780142086 Chr.1: 235175200-235175200 on Build GRCh38
rs770445291 Chr.1: 235175527-235175527 on Build GRCh38
rs778639149 Chr.1: 235175260-235175260 on Build GRCh38
rs371919109 Chr.1: 235175357-235175357 on Build GRCh38
rs754952973 Chr.1: 235175428-235175428 on Build GRCh38
rs781660068 Chr.1: 235175461-235175461 on Build GRCh38
rs769948123 Chr.1: 235175320-235175329 on Build GRCh38
rs771898573 Chr.1: 235175440-235175442 on Build GRCh38
rs758011877 Chr.1: 235175272-235175272 on Build GRCh38
rs749832844 Chr.1: 235175267-235175267 on Build GRCh38
rs1012249117 Chr.1: 235175420-235175421 on Build GRCh38
rs778764861 Chr.1: 235175358-235175358 on Build GRCh38
rs774467532 Chr.1: 235175327-235175327 on Build GRCh38
rs752094055 Chr.1: 235175230-235175230 on Build GRCh38
rs916434246 Chr.1: 235175510-235175510 on Build GRCh38
rs181722934 Chr.1: 235175446-235175446 on Build GRCh38
rs769852275 Chr.1: 235175292-235175292 on Build GRCh38
rs771023913 Chr.1: 235175323-235175323 on Build GRCh38
rs768067220 Chr.1: 235175335-235175335 on Build GRCh38
rs955143667 Chr.1: 235175213-235175213 on Build GRCh38
rs376544379 Chr.1: 235175371-235175371 on Build GRCh38
rs766807801 Chr.1: 235175228-235175228 on Build GRCh38
rs773154620 Chr.1: 235175226-235175226 on Build GRCh38
rs868028840 Chr.1: 235175493-235175493 on Build GRCh38
rs1034159570 Chr.1: 235175368-235175368 on Build GRCh38
rs201386389 Chr.1: 235175219-235175219 on Build GRCh38
rs781204590 Chr.1: 235175430-235175430 on Build GRCh38
rs773489125 Chr.1: 235175293-235175293 on Build GRCh38
rs761909965 Chr.1: 235175222-235175222 on Build GRCh38
rs1031693642 Chr.1: 235175585-235175585 on Build GRCh38
rs771844191 Chr.1: 235175176-235175176 on Build GRCh38
rs747833226 Chr.1: 235175437-235175437 on Build GRCh38
rs757315417 Chr.1: 235175192-235175192 on Build GRCh38
rs748033184 Chr.1: 235175290-235175290 on Build GRCh38
rs763108617 Chr.1: 235175227-235175227 on Build GRCh38
rs145519146 Chr.1: 235175332-235175332 on Build GRCh38
rs367684226 Chr.1: 235175173-235175173 on Build GRCh38
rs759322980 Chr.1: 235175417-235175417 on Build GRCh38
rs773481316 Chr.1: 235175389-235175391 on Build GRCh38
rs148402228 Chr.1: 235175303-235175303 on Build GRCh38
rs751180880 Chr.1: 235175424-235175424 on Build GRCh38
rs756987619 Chr.1: 235175170-235175170 on Build GRCh38
rs948793855 Chr.1: 235175529-235175529 on Build GRCh38
rs375954375 Chr.1: 235175450-235175450 on Build GRCh38
rs746920622 Chr.1: 235175284-235175284 on Build GRCh38
rs765955952 Chr.1: 235175421-235175421 on Build GRCh38
rs978060684 Chr.1: 235175577-235175577 on Build GRCh38
rs753168624 Chr.1: 235175249-235175249 on Build GRCh38
rs147900250 Chr.1: 235175282-235175282 on Build GRCh38
rs200039195 Chr.1: 235175210-235175210 on Build GRCh38
rs370264947 Chr.1: 235175174-235175174 on Build GRCh38
rs1037775671 Chr.1: 235175576-235175576 on Build GRCh38
rs758171415 Chr.1: 235175411-235175411 on Build GRCh38

View Full Product Details