Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 102685917-102686427 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL3
Entrez Gene ID
403
Gene Name
ADP ribosylation factor like GTPase 3
Gene Aliases
ARFL3
UniGene
Hs.182215
Gene Chromosome Location
Chr.10: 102673727-102714433 on Build GRCh38
Transcripts
NM_004311.3 XM_017016260.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
234413 c.388T>C p.L130L Chr.10: 102685929-102685929 on Build GRCh38
5168333 c.395C>T p.T132I Chr.10: 102685922-102685922 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs912573546 Chr.10: 102686269-102686269 on Build GRCh38
rs901848719 Chr.10: 102686367-102686367 on Build GRCh38
rs985399042 Chr.10: 102686046-102686046 on Build GRCh38
rs1025067541 Chr.10: 102686125-102686125 on Build GRCh38
rs763060317 Chr.10: 102686004-102686004 on Build GRCh38
rs904911418 Chr.10: 102686147-102686147 on Build GRCh38
rs752095202 Chr.10: 102685950-102685950 on Build GRCh38
rs1052667934 Chr.10: 102686070-102686070 on Build GRCh38
rs767890413 Chr.10: 102686045-102686045 on Build GRCh38
rs767616572 Chr.10: 102686324-102686324 on Build GRCh38
rs939063692 Chr.10: 102686194-102686194 on Build GRCh38
rs1056212668 Chr.10: 102686200-102686200 on Build GRCh38
rs745708839 Chr.10: 102685995-102685995 on Build GRCh38
rs551366324 Chr.10: 102685964-102685964 on Build GRCh38
rs762104856 Chr.10: 102686323-102686323 on Build GRCh38
rs762457890 Chr.10: 102686043-102686043 on Build GRCh38
rs776111003 Chr.10: 102686159-102686159 on Build GRCh38
rs954618910 Chr.10: 102686209-102686209 on Build GRCh38
rs976174264 Chr.10: 102686031-102686031 on Build GRCh38
rs770692966 Chr.10: 102685990-102685990 on Build GRCh38
rs892127507 Chr.10: 102686091-102686091 on Build GRCh38
rs7086474 Chr.10: 102686273-102686273 on Build GRCh38
rs963235693 Chr.10: 102686187-102686187 on Build GRCh38
rs903747789 Chr.10: 102686237-102686237 on Build GRCh38
rs7086592 Chr.10: 102686300-102686300 on Build GRCh38
rs953615938 Chr.10: 102686386-102686386 on Build GRCh38
rs141933852 Chr.10: 102686206-102686206 on Build GRCh38
rs757913078 Chr.10: 102685951-102685951 on Build GRCh38
rs1029071463 Chr.10: 102686382-102686382 on Build GRCh38
rs770491993 Chr.10: 102685993-102685993 on Build GRCh38
rs936877694 Chr.10: 102686314-102686314 on Build GRCh38
rs186305929 Chr.10: 102686401-102686401 on Build GRCh38
rs201396334 Chr.10: 102686401-102686401 on Build GRCh38
rs966091093 Chr.10: 102686018-102686018 on Build GRCh38
rs754154319 Chr.10: 102686231-102686231 on Build GRCh38
rs755402499 Chr.10: 102686002-102686003 on Build GRCh38
rs756470719 Chr.10: 102686191-102686191 on Build GRCh38
rs1013210172 Chr.10: 102686098-102686098 on Build GRCh38
rs538533205 Chr.10: 102685999-102685999 on Build GRCh38
rs543297446 Chr.10: 102686347-102686347 on Build GRCh38
rs183437903 Chr.10: 102686392-102686392 on Build GRCh38
rs925448943 Chr.10: 102686312-102686312 on Build GRCh38
rs568652625 Chr.10: 102686102-102686102 on Build GRCh38
rs762594294 Chr.10: 102686071-102686072 on Build GRCh38
rs1052403289 Chr.10: 102686351-102686351 on Build GRCh38
rs879229077 Chr.10: 102686072-102686072 on Build GRCh38
rs540283820 Chr.10: 102686391-102686391 on Build GRCh38
rs940964789 Chr.10: 102686140-102686140 on Build GRCh38
rs769860527 Chr.10: 102685998-102685998 on Build GRCh38
rs768923624 Chr.10: 102686032-102686032 on Build GRCh38
rs553943689 Chr.10: 102686173-102686173 on Build GRCh38
rs150441720 Chr.10: 102686222-102686222 on Build GRCh38
rs936914628 Chr.10: 102686067-102686067 on Build GRCh38
rs894920395 Chr.10: 102686205-102686205 on Build GRCh38
rs536080152 Chr.10: 102686158-102686158 on Build GRCh38
rs755184485 Chr.10: 102686385-102686385 on Build GRCh38
rs776696557 Chr.10: 102685945-102685945 on Build GRCh38
rs750580765 Chr.10: 102686368-102686368 on Build GRCh38
rs571007258 Chr.10: 102685979-102685979 on Build GRCh38
rs10708938 Chr.10: 102686392-102686392 on Build GRCh38
rs556934829 Chr.10: 102686072-102686072 on Build GRCh38
rs796677877 Chr.10: 102686400-102686400 on Build GRCh38
rs774984256 Chr.10: 102686040-102686040 on Build GRCh38
rs3961463 Chr.10: 102686111-102686111 on Build GRCh38

View Full Product Details