Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 151801124-151801650 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL5A
Entrez Gene ID
26225
Gene Name
ADP ribosylation factor like GTPase 5A
Gene Aliases
ARFLP5, ARL5
UniGene
Hs.470233
Gene Chromosome Location
Chr.2: 151800966-151828495 on Build GRCh38
Transcripts
NM_001037174.1 NM_012097.3 NM_177985.2

SNP Detail

SNP ID Chromosome Location
rs929791386 Chr.2: 151801477-151801477 on Build GRCh38
rs983937571 Chr.2: 151801478-151801478 on Build GRCh38
rs56312355 Chr.2: 151801404-151801404 on Build GRCh38
rs888155643 Chr.2: 151801176-151801176 on Build GRCh38
rs538743826 Chr.2: 151801218-151801218 on Build GRCh38
rs909627150 Chr.2: 151801495-151801495 on Build GRCh38
rs986148208 Chr.2: 151801330-151801330 on Build GRCh38
rs903399806 Chr.2: 151801561-151801561 on Build GRCh38
rs147841628 Chr.2: 151801301-151801301 on Build GRCh38
rs76387552 Chr.2: 151801600-151801600 on Build GRCh38
rs556949739 Chr.2: 151801297-151801297 on Build GRCh38
rs776106978 Chr.2: 151801373-151801373 on Build GRCh38
rs965317760 Chr.2: 151801379-151801379 on Build GRCh38
rs542671589 Chr.2: 151801308-151801308 on Build GRCh38
rs1000774209 Chr.2: 151801587-151801587 on Build GRCh38
rs1042352117 Chr.2: 151801537-151801537 on Build GRCh38
rs918466942 Chr.2: 151801470-151801470 on Build GRCh38
rs889217900 Chr.2: 151801606-151801606 on Build GRCh38
rs191859855 Chr.2: 151801516-151801516 on Build GRCh38
rs1055350676 Chr.2: 151801213-151801213 on Build GRCh38
rs1001165399 Chr.2: 151801207-151801211 on Build GRCh38
rs953448233 Chr.2: 151801326-151801326 on Build GRCh38
rs950340638 Chr.2: 151801534-151801534 on Build GRCh38
rs186973767 Chr.2: 151801158-151801158 on Build GRCh38
rs559385003 Chr.2: 151801312-151801312 on Build GRCh38
rs368915073 Chr.2: 151801584-151801585 on Build GRCh38
rs577726663 Chr.2: 151801279-151801279 on Build GRCh38
rs557573287 Chr.2: 151801270-151801270 on Build GRCh38
rs895465130 Chr.2: 151801528-151801528 on Build GRCh38
rs530400351 Chr.2: 151801624-151801624 on Build GRCh38
rs924986621 Chr.2: 151801448-151801448 on Build GRCh38
rs573012857 Chr.2: 151801322-151801322 on Build GRCh38
rs551272122 Chr.2: 151801567-151801567 on Build GRCh38
rs565478221 Chr.2: 151801508-151801508 on Build GRCh38
rs1054883 Chr.2: 151801335-151801335 on Build GRCh38
rs1014444257 Chr.2: 151801266-151801266 on Build GRCh38
rs537357477 Chr.2: 151801284-151801284 on Build GRCh38
rs758464194 Chr.2: 151801469-151801469 on Build GRCh38
rs555621793 Chr.2: 151801506-151801506 on Build GRCh38
rs752673074 Chr.2: 151801429-151801429 on Build GRCh38

View Full Product Details