Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 52548990-52549523 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARPP19
Entrez Gene ID
10776
Gene Name
cAMP regulated phosphoprotein 19
Gene Aliases
ARPP-16, ARPP-19, ARPP16, ENSAL
UniGene
Hs.512908
Gene Chromosome Location
Chr.15: 52547045-52569446 on Build GRCh38
Transcripts
NM_001306191.1 NM_001306195.1 NM_001306196.1 NM_006628.5 XM_017021868.1 XM_017021869.1 XM_017021870.1 XM_017021871.1

SNP Detail

SNP ID Chromosome Location
rs765275800 Chr.15: 52549302-52549302 on Build GRCh38
rs192719247 Chr.15: 52549440-52549440 on Build GRCh38
rs754867442 Chr.15: 52549138-52549138 on Build GRCh38
rs937821127 Chr.15: 52549122-52549122 on Build GRCh38
rs183117887 Chr.15: 52549484-52549484 on Build GRCh38
rs141579132 Chr.15: 52549441-52549441 on Build GRCh38
rs150341705 Chr.15: 52549070-52549070 on Build GRCh38
rs1026522571 Chr.15: 52549077-52549077 on Build GRCh38
rs933295397 Chr.15: 52549468-52549468 on Build GRCh38
rs532801906 Chr.15: 52549158-52549158 on Build GRCh38
rs879259910 Chr.15: 52549358-52549359 on Build GRCh38
rs978381594 Chr.15: 52549453-52549453 on Build GRCh38
rs889310046 Chr.15: 52549269-52549269 on Build GRCh38
rs989377637 Chr.15: 52549144-52549144 on Build GRCh38
rs558478628 Chr.15: 52549013-52549013 on Build GRCh38
rs971715311 Chr.15: 52549066-52549066 on Build GRCh38
rs915143467 Chr.15: 52549154-52549154 on Build GRCh38
rs982234651 Chr.15: 52549087-52549087 on Build GRCh38
rs766101409 Chr.15: 52549014-52549014 on Build GRCh38
rs149053555 Chr.15: 52549362-52549362 on Build GRCh38
rs1041719610 Chr.15: 52549231-52549231 on Build GRCh38
rs970768679 Chr.15: 52549365-52549365 on Build GRCh38
rs947427895 Chr.15: 52549155-52549155 on Build GRCh38
rs143097628 Chr.15: 52549364-52549364 on Build GRCh38
rs527375020 Chr.15: 52549464-52549464 on Build GRCh38
rs1007713585 Chr.15: 52549302-52549303 on Build GRCh38
rs761027336 Chr.15: 52549344-52549344 on Build GRCh38
rs201098733 Chr.15: 52549369-52549369 on Build GRCh38
rs933430123 Chr.15: 52549257-52549257 on Build GRCh38
rs924569462 Chr.15: 52549245-52549245 on Build GRCh38
rs576436292 Chr.15: 52549286-52549286 on Build GRCh38
rs570347433 Chr.15: 52549015-52549015 on Build GRCh38
rs559660370 Chr.15: 52549426-52549426 on Build GRCh38
rs910627724 Chr.15: 52549490-52549490 on Build GRCh38
rs182633249 Chr.15: 52549068-52549068 on Build GRCh38
rs539359341 Chr.15: 52549065-52549065 on Build GRCh38
rs1003310775 Chr.15: 52549368-52549368 on Build GRCh38
rs11539780 Chr.15: 52549425-52549425 on Build GRCh38
rs138047547 Chr.15: 52549259-52549259 on Build GRCh38
rs367720509 Chr.15: 52549148-52549148 on Build GRCh38
rs62023330 Chr.15: 52549104-52549104 on Build GRCh38
rs1033826367 Chr.15: 52549385-52549385 on Build GRCh38
rs545598933 Chr.15: 52549253-52549253 on Build GRCh38
rs924649334 Chr.15: 52549465-52549465 on Build GRCh38
rs187768407 Chr.15: 52549354-52549354 on Build GRCh38
rs966603486 Chr.15: 52549448-52549448 on Build GRCh38
rs970505096 Chr.15: 52549080-52549080 on Build GRCh38

View Full Product Details