Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 238450251-238450742 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB1
Entrez Gene ID
51665
Gene Name
ankyrin repeat and SOCS box containing 1
Gene Aliases
ASB-1
UniGene
Hs.516788
Gene Chromosome Location
Chr.2: 238426741-238452250 on Build GRCh38
Transcripts
NM_001040445.1 XM_005246080.1

SNP Detail

SNP ID Chromosome Location
rs150435665 Chr.2: 238450423-238450423 on Build GRCh38
rs1028329779 Chr.2: 238450581-238450581 on Build GRCh38
rs767633507 Chr.2: 238450563-238450563 on Build GRCh38
rs956309326 Chr.2: 238450645-238450645 on Build GRCh38
rs953293107 Chr.2: 238450342-238450342 on Build GRCh38
rs1015865460 Chr.2: 238450301-238450301 on Build GRCh38
rs987809636 Chr.2: 238450387-238450387 on Build GRCh38
rs376783758 Chr.2: 238450591-238450591 on Build GRCh38
rs535721605 Chr.2: 238450670-238450670 on Build GRCh38
rs962902532 Chr.2: 238450521-238450521 on Build GRCh38
rs1014662234 Chr.2: 238450286-238450286 on Build GRCh38
rs1043458787 Chr.2: 238450466-238450466 on Build GRCh38
rs528675136 Chr.2: 238450628-238450628 on Build GRCh38
rs1028853440 Chr.2: 238450336-238450336 on Build GRCh38
rs926334920 Chr.2: 238450680-238450680 on Build GRCh38
rs187715429 Chr.2: 238450460-238450460 on Build GRCh38
rs994651799 Chr.2: 238450303-238450303 on Build GRCh38
rs60171769 Chr.2: 238450572-238450572 on Build GRCh38
rs761851449 Chr.2: 238450537-238450537 on Build GRCh38
rs978200692 Chr.2: 238450427-238450427 on Build GRCh38
rs564469919 Chr.2: 238450625-238450625 on Build GRCh38
rs530960523 Chr.2: 238450505-238450505 on Build GRCh38
rs888947337 Chr.2: 238450636-238450636 on Build GRCh38
rs960624603 Chr.2: 238450288-238450288 on Build GRCh38
rs912792278 Chr.2: 238450428-238450428 on Build GRCh38
rs962812519 Chr.2: 238450325-238450325 on Build GRCh38
rs906194038 Chr.2: 238450270-238450270 on Build GRCh38
rs574746298 Chr.2: 238450424-238450424 on Build GRCh38
rs914759885 Chr.2: 238450446-238450446 on Build GRCh38
rs1047095182 Chr.2: 238450458-238450458 on Build GRCh38
rs980280932 Chr.2: 238450678-238450678 on Build GRCh38
rs775783475 Chr.2: 238450481-238450481 on Build GRCh38
rs774860192 Chr.2: 238450508-238450508 on Build GRCh38
rs924047324 Chr.2: 238450433-238450433 on Build GRCh38
rs771734949 Chr.2: 238450404-238450404 on Build GRCh38
rs1034902956 Chr.2: 238450281-238450281 on Build GRCh38
rs551753235 Chr.2: 238450515-238450515 on Build GRCh38
rs937765810 Chr.2: 238450681-238450681 on Build GRCh38
rs974596782 Chr.2: 238450356-238450356 on Build GRCh38
rs896383036 Chr.2: 238450507-238450507 on Build GRCh38
rs948292150 Chr.2: 238450456-238450456 on Build GRCh38
rs1001843103 Chr.2: 238450489-238450489 on Build GRCh38
rs549762286 Chr.2: 238450659-238450659 on Build GRCh38
rs986745742 Chr.2: 238450632-238450632 on Build GRCh38
rs201416508 Chr.2: 238450580-238450580 on Build GRCh38
rs530009338 Chr.2: 238450349-238450349 on Build GRCh38
rs978118503 Chr.2: 238450658-238450658 on Build GRCh38
rs746687118 Chr.2: 238450432-238450432 on Build GRCh38
rs546837028 Chr.2: 238450649-238450649 on Build GRCh38
rs552615855 Chr.2: 238450590-238450590 on Build GRCh38
rs1004024907 Chr.2: 238450300-238450300 on Build GRCh38
rs898947467 Chr.2: 238450547-238450547 on Build GRCh38
rs370518727 Chr.2: 238450676-238450676 on Build GRCh38
rs563637982 Chr.2: 238450490-238450490 on Build GRCh38
rs914833421 Chr.2: 238450655-238450655 on Build GRCh38
rs934931672 Chr.2: 238450438-238450438 on Build GRCh38
rs540917610 Chr.2: 238450408-238450408 on Build GRCh38
rs1015416992 Chr.2: 238450516-238450516 on Build GRCh38
rs763378085 Chr.2: 238450484-238450484 on Build GRCh38
rs1056127188 Chr.2: 238450721-238450721 on Build GRCh38
rs750645915 Chr.2: 238450640-238450640 on Build GRCh38
rs116379867 Chr.2: 238450439-238450439 on Build GRCh38

View Full Product Details