Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 151049189-151049683 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASIC3
Entrez Gene ID
9311
Gene Name
acid sensing ion channel subunit 3
Gene Aliases
ACCN3, DRASIC, SLNAC1, TNaC1
UniGene
Hs.647113
Gene Chromosome Location
Chr.7: 151048292-151052756 on Build GRCh38
Transcripts
NM_004769.3 NM_020321.3 NM_020322.3 NR_046401.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1622562 c.380G>A p.R127H Chr.7: 151049265-151049265 on Build GRCh38
3381858 c.452C>T p.A151V Chr.7: 151049337-151049337 on Build GRCh38
452627 c.392G>A p.R131Q Chr.7: 151049277-151049277 on Build GRCh38
1449260 c.493C>T p.R165C Chr.7: 151049378-151049378 on Build GRCh38
6456468 c.309G>A p.T103T Chr.7: 151049194-151049194 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111784048 Chr.7: 151049252-151049252 on Build GRCh38
rs989131530 Chr.7: 151049575-151049575 on Build GRCh38
rs765427661 Chr.7: 151049277-151049277 on Build GRCh38
rs767906753 Chr.7: 151049294-151049294 on Build GRCh38
rs771834367 Chr.7: 151049268-151049268 on Build GRCh38
rs757766302 Chr.7: 151049386-151049387 on Build GRCh38
rs533530797 Chr.7: 151049522-151049522 on Build GRCh38
rs570186016 Chr.7: 151049656-151049656 on Build GRCh38
rs200963043 Chr.7: 151049326-151049326 on Build GRCh38
rs187345635 Chr.7: 151049267-151049267 on Build GRCh38
rs560534800 Chr.7: 151049266-151049266 on Build GRCh38
rs768911847 Chr.7: 151049468-151049468 on Build GRCh38
rs770645833 Chr.7: 151049448-151049448 on Build GRCh38
rs752207683 Chr.7: 151049215-151049219 on Build GRCh38
rs143756561 Chr.7: 151049325-151049325 on Build GRCh38
rs766867558 Chr.7: 151049385-151049385 on Build GRCh38
rs749222515 Chr.7: 151049222-151049222 on Build GRCh38
rs372587470 Chr.7: 151049216-151049216 on Build GRCh38
rs201385813 Chr.7: 151049334-151049334 on Build GRCh38
rs147943737 Chr.7: 151049273-151049273 on Build GRCh38
rs746978622 Chr.7: 151049440-151049440 on Build GRCh38
rs752000897 Chr.7: 151049333-151049334 on Build GRCh38
rs202138681 Chr.7: 151049288-151049288 on Build GRCh38
rs142151024 Chr.7: 151049221-151049221 on Build GRCh38
rs375363889 Chr.7: 151049429-151049429 on Build GRCh38
rs778287976 Chr.7: 151049279-151049279 on Build GRCh38
rs1002894418 Chr.7: 151049259-151049259 on Build GRCh38
rs568372040 Chr.7: 151049432-151049432 on Build GRCh38
rs1026428517 Chr.7: 151049623-151049623 on Build GRCh38
rs141998060 Chr.7: 151049264-151049264 on Build GRCh38
rs757767256 Chr.7: 151049293-151049293 on Build GRCh38
rs752199449 Chr.7: 151049422-151049422 on Build GRCh38
rs755778227 Chr.7: 151049256-151049256 on Build GRCh38
rs1031216898 Chr.7: 151049564-151049564 on Build GRCh38
rs199876394 Chr.7: 151049254-151049254 on Build GRCh38
rs770779744 Chr.7: 151049349-151049349 on Build GRCh38
rs975668582 Chr.7: 151049376-151049376 on Build GRCh38
rs558909266 Chr.7: 151049233-151049233 on Build GRCh38
rs762969627 Chr.7: 151049289-151049289 on Build GRCh38
rs747950910 Chr.7: 151049379-151049379 on Build GRCh38
rs888505439 Chr.7: 151049473-151049473 on Build GRCh38
rs145775749 Chr.7: 151049265-151049265 on Build GRCh38
rs894731992 Chr.7: 151049497-151049497 on Build GRCh38
rs553368532 Chr.7: 151049655-151049655 on Build GRCh38
rs952005306 Chr.7: 151049626-151049626 on Build GRCh38
rs201529251 Chr.7: 151049255-151049255 on Build GRCh38
rs772479203 Chr.7: 151049413-151049413 on Build GRCh38
rs965568603 Chr.7: 151049559-151049559 on Build GRCh38
rs62001896 Chr.7: 151049251-151049251 on Build GRCh38
rs753571682 Chr.7: 151049312-151049312 on Build GRCh38
rs778553914 Chr.7: 151049421-151049421 on Build GRCh38
rs768612148 Chr.7: 151049303-151049303 on Build GRCh38
rs777603969 Chr.7: 151049339-151049339 on Build GRCh38
rs749495490 Chr.7: 151049464-151049464 on Build GRCh38
rs1018424838 Chr.7: 151049507-151049507 on Build GRCh38
rs116500016 Chr.7: 151049242-151049242 on Build GRCh38
rs545075879 Chr.7: 151049426-151049426 on Build GRCh38
rs761664922 Chr.7: 151049230-151049230 on Build GRCh38
rs760901541 Chr.7: 151049384-151049384 on Build GRCh38
rs918472570 Chr.7: 151049218-151049218 on Build GRCh38
rs994148129 Chr.7: 151049437-151049437 on Build GRCh38
rs969150900 Chr.7: 151049637-151049637 on Build GRCh38
rs753477836 Chr.7: 151049391-151049391 on Build GRCh38
rs151199388 Chr.7: 151049224-151049224 on Build GRCh38
rs767242495 Chr.7: 151049231-151049231 on Build GRCh38
rs781030978 Chr.7: 151049463-151049463 on Build GRCh38
rs952057471 Chr.7: 151049337-151049337 on Build GRCh38
rs998884006 Chr.7: 151049563-151049563 on Build GRCh38
rs956583284 Chr.7: 151049566-151049566 on Build GRCh38
rs754483769 Chr.7: 151049313-151049313 on Build GRCh38
rs192297262 Chr.7: 151049378-151049378 on Build GRCh38
rs370369500 Chr.7: 151049243-151049243 on Build GRCh38
rs754745451 Chr.7: 151049419-151049419 on Build GRCh38
rs984076395 Chr.7: 151049227-151049227 on Build GRCh38
rs141877615 Chr.7: 151049290-151049290 on Build GRCh38
rs915013478 Chr.7: 151049580-151049580 on Build GRCh38
rs377323790 Chr.7: 151049223-151049223 on Build GRCh38
rs779816600 Chr.7: 151049306-151049306 on Build GRCh38
rs774467659 Chr.7: 151049371-151049371 on Build GRCh38
rs1006910422 Chr.7: 151049481-151049481 on Build GRCh38
rs763166166 Chr.7: 151049364-151049364 on Build GRCh38
rs746740072 Chr.7: 151049340-151049340 on Build GRCh38
rs980508040 Chr.7: 151049648-151049648 on Build GRCh38
rs139085042 Chr.7: 151049235-151049235 on Build GRCh38
rs35364592 Chr.7: 151049314-151049314 on Build GRCh38
rs769278125 Chr.7: 151049213-151049213 on Build GRCh38
rs758830431 Chr.7: 151049272-151049272 on Build GRCh38
rs768713899 Chr.7: 151049369-151049369 on Build GRCh38
rs993571397 Chr.7: 151049610-151049610 on Build GRCh38
rs1054612007 Chr.7: 151049475-151049475 on Build GRCh38
rs759793510 Chr.7: 151049276-151049276 on Build GRCh38
rs1046321872 Chr.7: 151049530-151049533 on Build GRCh38
rs1013051748 Chr.7: 151049504-151049504 on Build GRCh38
rs1036129053 Chr.7: 151049260-151049260 on Build GRCh38
rs370728181 Chr.7: 151049352-151049352 on Build GRCh38
rs375107350 Chr.7: 151049305-151049305 on Build GRCh38

View Full Product Details