Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 66088778-66089282 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASL
Entrez Gene ID
435
Gene Name
argininosuccinate lyase
Gene Aliases
ASAL
UniGene
Hs.632015
Gene Chromosome Location
Chr.7: 66075789-66093343 on Build GRCh38
Transcripts
NM_000048.3 NM_001024943.1 NM_001024944.1 NM_001024946.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5692213 c.897G>T p.K299N Chr.7: 66089154-66089154 on Build GRCh38
1091320 c.771C>T p.A257A Chr.7: 66088859-66088859 on Build GRCh38
7150638 c.780C>A p.L260L Chr.7: 66088868-66088868 on Build GRCh38
4877305 c.762C>T p.S254S Chr.7: 66088850-66088850 on Build GRCh38
6605852 c.758T>G p.L253R Chr.7: 66088846-66088846 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs398123128 Chr.7: 66089103-66089103 on Build GRCh38
rs928734370 Chr.7: 66088952-66088952 on Build GRCh38
rs1017897955 Chr.7: 66089035-66089035 on Build GRCh38
rs781331391 Chr.7: 66089180-66089180 on Build GRCh38
rs367601880 Chr.7: 66088966-66088966 on Build GRCh38
rs755245694 Chr.7: 66089174-66089174 on Build GRCh38
rs755700770 Chr.7: 66089054-66089054 on Build GRCh38
rs143359899 Chr.7: 66088868-66088868 on Build GRCh38
rs201894355 Chr.7: 66089058-66089058 on Build GRCh38
rs752333334 Chr.7: 66088852-66088852 on Build GRCh38
rs749306006 Chr.7: 66089249-66089249 on Build GRCh38
rs978132148 Chr.7: 66089242-66089242 on Build GRCh38
rs943625457 Chr.7: 66088872-66088872 on Build GRCh38
rs949611035 Chr.7: 66088937-66088937 on Build GRCh38
rs371927473 Chr.7: 66088829-66088829 on Build GRCh38
rs757231605 Chr.7: 66089060-66089060 on Build GRCh38
rs574132928 Chr.7: 66089008-66089008 on Build GRCh38
rs796145445 Chr.7: 66088915-66088915 on Build GRCh38
rs745601034 Chr.7: 66088916-66088916 on Build GRCh38
rs757313920 Chr.7: 66089253-66089253 on Build GRCh38
rs778854382 Chr.7: 66088899-66088899 on Build GRCh38
rs1051586035 Chr.7: 66089233-66089233 on Build GRCh38
rs369194533 Chr.7: 66089071-66089071 on Build GRCh38
rs758359461 Chr.7: 66089162-66089162 on Build GRCh38
rs183018428 Chr.7: 66088846-66088846 on Build GRCh38
rs771334759 Chr.7: 66089106-66089106 on Build GRCh38
rs776057556 Chr.7: 66089264-66089265 on Build GRCh38
rs756280770 Chr.7: 66089243-66089243 on Build GRCh38
rs770425849 Chr.7: 66089187-66089187 on Build GRCh38
rs909358635 Chr.7: 66088860-66088860 on Build GRCh38
rs924321091 Chr.7: 66088810-66088810 on Build GRCh38
rs760378137 Chr.7: 66089112-66089112 on Build GRCh38
rs894967634 Chr.7: 66088986-66088986 on Build GRCh38
rs747165417 Chr.7: 66088948-66088948 on Build GRCh38
rs869312991 Chr.7: 66088850-66088850 on Build GRCh38
rs375562374 Chr.7: 66089164-66089164 on Build GRCh38
rs868834862 Chr.7: 66089173-66089173 on Build GRCh38
rs767103799 Chr.7: 66088845-66088845 on Build GRCh38
rs993812214 Chr.7: 66089036-66089036 on Build GRCh38
rs201002874 Chr.7: 66089198-66089198 on Build GRCh38
rs936795106 Chr.7: 66089260-66089260 on Build GRCh38
rs370949043 Chr.7: 66089230-66089230 on Build GRCh38
rs544134586 Chr.7: 66088931-66088931 on Build GRCh38
rs772353887 Chr.7: 66088804-66088804 on Build GRCh38
rs373861796 Chr.7: 66089220-66089220 on Build GRCh38
rs761296434 Chr.7: 66089123-66089123 on Build GRCh38
rs769682758 Chr.7: 66088982-66088982 on Build GRCh38
rs3107306 Chr.7: 66088825-66088825 on Build GRCh38
rs375586912 Chr.7: 66088859-66088859 on Build GRCh38
rs890405845 Chr.7: 66089010-66089010 on Build GRCh38
rs774532004 Chr.7: 66089110-66089110 on Build GRCh38
rs746531359 Chr.7: 66089162-66089163 on Build GRCh38
rs28941472 Chr.7: 66089114-66089114 on Build GRCh38
rs897881743 Chr.7: 66088994-66088994 on Build GRCh38
rs757101256 Chr.7: 66088883-66088883 on Build GRCh38
rs1007427655 Chr.7: 66089030-66089031 on Build GRCh38
rs781009876 Chr.7: 66089232-66089232 on Build GRCh38
rs762415761 Chr.7: 66088958-66088958 on Build GRCh38
rs768826627 Chr.7: 66088951-66088951 on Build GRCh38
rs773583579 Chr.7: 66088970-66088970 on Build GRCh38
rs753218926 Chr.7: 66089059-66089059 on Build GRCh38
rs754800257 Chr.7: 66089231-66089231 on Build GRCh38
rs776526632 Chr.7: 66088957-66088957 on Build GRCh38
rs370973627 Chr.7: 66089118-66089118 on Build GRCh38
rs779050726 Chr.7: 66089067-66089067 on Build GRCh38
rs776183045 Chr.7: 66089217-66089217 on Build GRCh38
rs1012151007 Chr.7: 66088988-66088988 on Build GRCh38
rs747275181 Chr.7: 66089088-66089088 on Build GRCh38
rs781340332 Chr.7: 66089093-66089093 on Build GRCh38
rs748398698 Chr.7: 66089183-66089183 on Build GRCh38
rs374908945 Chr.7: 66088836-66088836 on Build GRCh38
rs766725941 Chr.7: 66088973-66088973 on Build GRCh38
rs750431938 Chr.7: 66089147-66089147 on Build GRCh38
rs919377349 Chr.7: 66089227-66089227 on Build GRCh38
rs772062084 Chr.7: 66088936-66088936 on Build GRCh38
rs769307500 Chr.7: 66089227-66089228 on Build GRCh38
rs777617137 Chr.7: 66089079-66089080 on Build GRCh38
rs776178127 Chr.7: 66089195-66089207 on Build GRCh38
rs563726769 Chr.7: 66088974-66088974 on Build GRCh38
rs905197473 Chr.7: 66088976-66088976 on Build GRCh38
rs777809575 Chr.7: 66089189-66089189 on Build GRCh38
rs777232999 Chr.7: 66088824-66088824 on Build GRCh38
rs752388478 Chr.7: 66089050-66089050 on Build GRCh38
rs1030108212 Chr.7: 66089004-66089004 on Build GRCh38
rs890289218 Chr.7: 66089263-66089263 on Build GRCh38
rs765601012 Chr.7: 66088964-66088964 on Build GRCh38
rs745791464 Chr.7: 66089070-66089070 on Build GRCh38
rs754634171 Chr.7: 66088837-66088837 on Build GRCh38
rs759270998 Chr.7: 66089200-66089207 on Build GRCh38
rs372611875 Chr.7: 66089188-66089188 on Build GRCh38
rs759321434 Chr.7: 66088972-66088972 on Build GRCh38
rs906418871 Chr.7: 66089026-66089026 on Build GRCh38
rs368492142 Chr.7: 66089229-66089229 on Build GRCh38
rs984259237 Chr.7: 66089078-66089078 on Build GRCh38
rs869312992 Chr.7: 66089146-66089146 on Build GRCh38
rs375625546 Chr.7: 66089234-66089234 on Build GRCh38
rs369828060 Chr.7: 66088806-66088806 on Build GRCh38
rs866982987 Chr.7: 66089079-66089079 on Build GRCh38
rs764859231 Chr.7: 66089224-66089224 on Build GRCh38
rs370805656 Chr.7: 66089211-66089211 on Build GRCh38
rs375854730 Chr.7: 66088925-66088925 on Build GRCh38
rs751249678 Chr.7: 66089169-66089169 on Build GRCh38
rs1024855006 Chr.7: 66088989-66088989 on Build GRCh38
rs762064987 Chr.7: 66089133-66089133 on Build GRCh38
rs149057077 Chr.7: 66088855-66088855 on Build GRCh38
rs923926957 Chr.7: 66089248-66089248 on Build GRCh38
rs749819433 Chr.7: 66089194-66089194 on Build GRCh38
rs764804216 Chr.7: 66089127-66089127 on Build GRCh38
rs543255240 Chr.7: 66089094-66089094 on Build GRCh38
rs1039301353 Chr.7: 66088879-66088879 on Build GRCh38
rs373749562 Chr.7: 66089086-66089086 on Build GRCh38
rs35348667 Chr.7: 66089064-66089064 on Build GRCh38
rs764072757 Chr.7: 66088808-66088808 on Build GRCh38
rs765660307 Chr.7: 66088835-66088835 on Build GRCh38
rs993315052 Chr.7: 66088998-66088998 on Build GRCh38
rs768722904 Chr.7: 66089090-66089090 on Build GRCh38

View Full Product Details