Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 50814124-50814523 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATF1
Entrez Gene ID
466
Gene Name
activating transcription factor 1
Gene Aliases
EWS-ATF1, FUS/ATF-1, TREB36
UniGene
Hs.648565
Gene Chromosome Location
Chr.12: 50763682-50821162 on Build GRCh38
Transcripts
NM_005171.4 XM_011538386.2 XM_011538387.2 XM_011538388.2 XM_017019331.1 XM_017019332.1 XM_017019333.1 XM_017019334.1 XM_017019335.1 XM_017019336.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5892167 c.544C>T p.R182* Chr.12: 50814312-50814312 on Build GRCh38
7242271 c.663G>A p.M221I Chr.12: 50814431-50814431 on Build GRCh38
940396 c.647G>C p.R216T Chr.12: 50814415-50814415 on Build GRCh38
5801608 c.671+5_671+15delGTAGTAAAATC p.? Chr.12: 50814444-50814454 on Build GRCh38
3812245 c.540G>C p.Q180H Chr.12: 50814308-50814308 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765693263 Chr.12: 50814180-50814180 on Build GRCh38
rs760148333 Chr.12: 50814445-50814445 on Build GRCh38
rs201826197 Chr.12: 50814455-50814455 on Build GRCh38
rs761297560 Chr.12: 50814305-50814305 on Build GRCh38
rs770356640 Chr.12: 50814382-50814382 on Build GRCh38
rs750484770 Chr.12: 50814473-50814473 on Build GRCh38
rs771546607 Chr.12: 50814170-50814170 on Build GRCh38
rs953935464 Chr.12: 50814218-50814218 on Build GRCh38
rs763703570 Chr.12: 50814296-50814296 on Build GRCh38
rs754864097 Chr.12: 50814356-50814356 on Build GRCh38
rs371428536 Chr.12: 50814313-50814313 on Build GRCh38
rs778139346 Chr.12: 50814359-50814359 on Build GRCh38
rs763554829 Chr.12: 50814196-50814196 on Build GRCh38
rs750430771 Chr.12: 50814238-50814238 on Build GRCh38
rs541406693 Chr.12: 50814200-50814200 on Build GRCh38
rs866106630 Chr.12: 50814306-50814306 on Build GRCh38
rs773933246 Chr.12: 50814408-50814408 on Build GRCh38
rs151321417 Chr.12: 50814425-50814425 on Build GRCh38
rs781084925 Chr.12: 50814470-50814470 on Build GRCh38
rs750462212 Chr.12: 50814274-50814274 on Build GRCh38
rs751891910 Chr.12: 50814362-50814362 on Build GRCh38
rs984375597 Chr.12: 50814395-50814395 on Build GRCh38
rs778557758 Chr.12: 50814252-50814252 on Build GRCh38
rs75866814 Chr.12: 50814456-50814456 on Build GRCh38
rs747057496 Chr.12: 50814254-50814254 on Build GRCh38
rs773453169 Chr.12: 50814149-50814149 on Build GRCh38
rs771205092 Chr.12: 50814256-50814256 on Build GRCh38
rs780465497 Chr.12: 50814393-50814393 on Build GRCh38
rs759818498 Chr.12: 50814175-50814175 on Build GRCh38
rs750257536 Chr.12: 50814321-50814321 on Build GRCh38
rs763356871 Chr.12: 50814295-50814295 on Build GRCh38
rs767563838 Chr.12: 50814262-50814262 on Build GRCh38
rs762599221 Chr.12: 50814192-50814192 on Build GRCh38
rs769987495 Chr.12: 50814281-50814281 on Build GRCh38
rs757530629 Chr.12: 50814469-50814469 on Build GRCh38
rs373951609 Chr.12: 50814447-50814447 on Build GRCh38
rs781338514 Chr.12: 50814372-50814372 on Build GRCh38
rs780523874 Chr.12: 50814486-50814486 on Build GRCh38
rs772796649 Chr.12: 50814442-50814442 on Build GRCh38
rs745516430 Chr.12: 50814257-50814258 on Build GRCh38
rs772657784 Chr.12: 50814245-50814248 on Build GRCh38
rs748504510 Chr.12: 50814241-50814244 on Build GRCh38
rs774055278 Chr.12: 50814302-50814302 on Build GRCh38
rs374223169 Chr.12: 50814203-50814203 on Build GRCh38
rs746321131 Chr.12: 50814376-50814376 on Build GRCh38
rs137918714 Chr.12: 50814407-50814407 on Build GRCh38
rs983579164 Chr.12: 50814154-50814154 on Build GRCh38
rs755954198 Chr.12: 50814245-50814245 on Build GRCh38
rs771441165 Chr.12: 50814428-50814428 on Build GRCh38
rs751778473 Chr.12: 50814185-50814185 on Build GRCh38
rs762062238 Chr.12: 50814186-50814186 on Build GRCh38
rs564265317 Chr.12: 50814319-50814319 on Build GRCh38
rs747474459 Chr.12: 50814162-50814162 on Build GRCh38
rs532503300 Chr.12: 50814337-50814337 on Build GRCh38
rs748275166 Chr.12: 50814243-50814243 on Build GRCh38
rs1046566567 Chr.12: 50814226-50814226 on Build GRCh38
rs775954565 Chr.12: 50814285-50814285 on Build GRCh38
rs372346025 Chr.12: 50814483-50814483 on Build GRCh38
rs751767737 Chr.12: 50814458-50814458 on Build GRCh38
rs757355336 Chr.12: 50814363-50814363 on Build GRCh38
rs781775467 Chr.12: 50814191-50814194 on Build GRCh38
rs749648702 Chr.12: 50814402-50814402 on Build GRCh38
rs113509493 Chr.12: 50814164-50814164 on Build GRCh38
rs767437117 Chr.12: 50814223-50814223 on Build GRCh38
rs368181179 Chr.12: 50814454-50814454 on Build GRCh38
rs547316482 Chr.12: 50814250-50814250 on Build GRCh38
rs2230674 Chr.12: 50814339-50814339 on Build GRCh38
rs771558279 Chr.12: 50814347-50814348 on Build GRCh38
rs753668650 Chr.12: 50814343-50814343 on Build GRCh38
rs367769529 Chr.12: 50814241-50814241 on Build GRCh38
rs746401740 Chr.12: 50814266-50814266 on Build GRCh38
rs200931165 Chr.12: 50814246-50814246 on Build GRCh38
rs777239021 Chr.12: 50814173-50814173 on Build GRCh38
rs546249429 Chr.12: 50814242-50814242 on Build GRCh38

View Full Product Details