Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 55368561-55369062 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG14
Entrez Gene ID
22863
Gene Name
autophagy related 14
Gene Aliases
ATG14L, BARKOR, KIAA0831
UniGene
Hs.414809
Gene Chromosome Location
Chr.14: 55366391-55411858 on Build GRCh38
Transcripts
NM_014924.4 XM_011536563.2

SNP Detail

SNP ID Chromosome Location
rs530380041 Chr.14: 55368801-55368801 on Build GRCh38
rs939118064 Chr.14: 55368707-55368707 on Build GRCh38
rs997838589 Chr.14: 55368708-55368708 on Build GRCh38
rs997218761 Chr.14: 55368997-55368997 on Build GRCh38
rs763463747 Chr.14: 55368947-55368947 on Build GRCh38
rs1004641439 Chr.14: 55369025-55369025 on Build GRCh38
rs932583789 Chr.14: 55369037-55369037 on Build GRCh38
rs982975288 Chr.14: 55369002-55369002 on Build GRCh38
rs990436333 Chr.14: 55368646-55368646 on Build GRCh38
rs1043249123 Chr.14: 55368902-55368902 on Build GRCh38
rs963222708 Chr.14: 55369033-55369033 on Build GRCh38
rs187007926 Chr.14: 55368818-55368818 on Build GRCh38
rs938720804 Chr.14: 55368817-55368817 on Build GRCh38
rs957345651 Chr.14: 55368635-55368635 on Build GRCh38
rs1030632481 Chr.14: 55369000-55369000 on Build GRCh38
rs1000494435 Chr.14: 55368649-55368649 on Build GRCh38
rs113372251 Chr.14: 55368646-55368647 on Build GRCh38
rs146198075 Chr.14: 55368951-55368951 on Build GRCh38
rs565010540 Chr.14: 55368589-55368589 on Build GRCh38
rs908300536 Chr.14: 55369005-55369006 on Build GRCh38
rs982026835 Chr.14: 55368686-55368686 on Build GRCh38
rs1001013810 Chr.14: 55368917-55368917 on Build GRCh38
rs776412146 Chr.14: 55368974-55368975 on Build GRCh38
rs770670215 Chr.14: 55368886-55368887 on Build GRCh38
rs962400439 Chr.14: 55369022-55369022 on Build GRCh38
rs540840818 Chr.14: 55369001-55369001 on Build GRCh38
rs1017695104 Chr.14: 55368688-55368688 on Build GRCh38
rs553971541 Chr.14: 55369031-55369031 on Build GRCh38
rs963440459 Chr.14: 55368799-55368799 on Build GRCh38
rs913500188 Chr.14: 55368844-55368844 on Build GRCh38
rs762047695 Chr.14: 55368588-55368588 on Build GRCh38
rs1032111984 Chr.14: 55368634-55368634 on Build GRCh38
rs566233548 Chr.14: 55368770-55368770 on Build GRCh38
rs996635600 Chr.14: 55368586-55368586 on Build GRCh38
rs377647765 Chr.14: 55369027-55369027 on Build GRCh38
rs1053202219 Chr.14: 55368899-55368899 on Build GRCh38
rs997626368 Chr.14: 55368600-55368618 on Build GRCh38
rs909264168 Chr.14: 55368790-55368790 on Build GRCh38
rs1036295033 Chr.14: 55368982-55368983 on Build GRCh38
rs191291953 Chr.14: 55368840-55368840 on Build GRCh38
rs182643161 Chr.14: 55368908-55368908 on Build GRCh38
rs1010438294 Chr.14: 55368618-55368618 on Build GRCh38
rs928792808 Chr.14: 55368691-55368694 on Build GRCh38
rs1050103633 Chr.14: 55368968-55368980 on Build GRCh38
rs946173673 Chr.14: 55368864-55368864 on Build GRCh38
rs370796609 Chr.14: 55368627-55368627 on Build GRCh38
rs900146894 Chr.14: 55368985-55368985 on Build GRCh38
rs927285080 Chr.14: 55368811-55368811 on Build GRCh38
rs980246347 Chr.14: 55368802-55368802 on Build GRCh38
rs903623070 Chr.14: 55368642-55368642 on Build GRCh38
rs941026802 Chr.14: 55368729-55368729 on Build GRCh38
rs998925777 Chr.14: 55368621-55368622 on Build GRCh38
rs970258545 Chr.14: 55368672-55368672 on Build GRCh38
rs964441331 Chr.14: 55368706-55368706 on Build GRCh38
rs182680080 Chr.14: 55368713-55368713 on Build GRCh38
rs950538838 Chr.14: 55368716-55368716 on Build GRCh38
rs1037959866 Chr.14: 55368765-55368765 on Build GRCh38

View Full Product Details