Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 42639790-42640294 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP5L2
Entrez Gene ID
267020
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex subunit G2
Gene Aliases
ATP5K2
UniGene
Hs.664737
Gene Chromosome Location
Chr.22: 42639803-42640601 on Build GRCh38
Transcripts
NM_001165877.1 NM_000398.6 NM_001129819.2 NM_001171660.1 NM_001171661.1 NM_007326.4

Gene Symbol
CYB5R3
Entrez Gene ID
1727
Gene Name
cytochrome b5 reductase 3
Gene Aliases
B5R, DIA1
UniGene
Hs.561064
Gene Chromosome Location
Chr.22: 42617840-42649399 on Build GRCh38
Transcripts
NM_001165877.1 NM_000398.6 NM_001129819.2 NM_001171660.1 NM_001171661.1 NM_007326.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7334494 c.220T>G p.L74V Chr.22: 42640055-42640055 on Build GRCh38
3061958 c.228C>T p.N76N Chr.22: 42640047-42640047 on Build GRCh38
7285452 c.37C>T p.P13S Chr.22: 42640238-42640238 on Build GRCh38
1593071 c.255G>A p.T85T Chr.22: 42640020-42640020 on Build GRCh38
1184106 c.254C>T p.T85M Chr.22: 42640021-42640021 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772255443 Chr.22: 42639975-42639975 on Build GRCh38
rs1000720378 Chr.22: 42640095-42640095 on Build GRCh38
rs59947151 Chr.22: 42639844-42639845 on Build GRCh38
rs75163608 Chr.22: 42639845-42639845 on Build GRCh38
rs373395069 Chr.22: 42640173-42640173 on Build GRCh38
rs879147278 Chr.22: 42639844-42639844 on Build GRCh38
rs897138660 Chr.22: 42639825-42639825 on Build GRCh38
rs12628044 Chr.22: 42639922-42639922 on Build GRCh38
rs755105480 Chr.22: 42640114-42640114 on Build GRCh38
rs769780072 Chr.22: 42640233-42640233 on Build GRCh38
rs762594007 Chr.22: 42640170-42640170 on Build GRCh38
rs781241872 Chr.22: 42640124-42640124 on Build GRCh38
rs11549139 Chr.22: 42640245-42640245 on Build GRCh38
rs754956706 Chr.22: 42639857-42639858 on Build GRCh38
rs766146399 Chr.22: 42640171-42640171 on Build GRCh38
rs768349855 Chr.22: 42640153-42640153 on Build GRCh38
rs748638349 Chr.22: 42640048-42640048 on Build GRCh38
rs573769598 Chr.22: 42640149-42640149 on Build GRCh38
rs553735089 Chr.22: 42639823-42639823 on Build GRCh38
rs6519327 Chr.22: 42640239-42640239 on Build GRCh38
rs747047782 Chr.22: 42640045-42640045 on Build GRCh38
rs757091725 Chr.22: 42640101-42640104 on Build GRCh38
rs770258622 Chr.22: 42640058-42640058 on Build GRCh38
rs751647818 Chr.22: 42640111-42640111 on Build GRCh38
rs768404888 Chr.22: 42640209-42640209 on Build GRCh38
rs781323667 Chr.22: 42639845-42639846 on Build GRCh38
rs776768242 Chr.22: 42640084-42640084 on Build GRCh38
rs755475458 Chr.22: 42640046-42640046 on Build GRCh38
rs767631567 Chr.22: 42640177-42640177 on Build GRCh38
rs776252710 Chr.22: 42640156-42640156 on Build GRCh38
rs145037839 Chr.22: 42639844-42639844 on Build GRCh38
rs761654392 Chr.22: 42640097-42640097 on Build GRCh38
rs927547039 Chr.22: 42640180-42640180 on Build GRCh38
rs746648738 Chr.22: 42640150-42640150 on Build GRCh38
rs543593140 Chr.22: 42640127-42640127 on Build GRCh38
rs376776418 Chr.22: 42640192-42640192 on Build GRCh38
rs778222320 Chr.22: 42640136-42640136 on Build GRCh38
rs747559321 Chr.22: 42639984-42639984 on Build GRCh38
rs753691222 Chr.22: 42640021-42640021 on Build GRCh38
rs747576014 Chr.22: 42640218-42640218 on Build GRCh38
rs757584774 Chr.22: 42639962-42639962 on Build GRCh38
rs754243793 Chr.22: 42640191-42640191 on Build GRCh38
rs577533684 Chr.22: 42640190-42640190 on Build GRCh38
rs759851533 Chr.22: 42640077-42640077 on Build GRCh38
rs773416768 Chr.22: 42640059-42640059 on Build GRCh38
rs774108301 Chr.22: 42640259-42640259 on Build GRCh38
rs750264117 Chr.22: 42639844-42639846 on Build GRCh38
rs752669494 Chr.22: 42640178-42640178 on Build GRCh38
rs752853980 Chr.22: 42640162-42640163 on Build GRCh38
rs549954057 Chr.22: 42640246-42640246 on Build GRCh38
rs1051608113 Chr.22: 42639913-42639913 on Build GRCh38
rs765122495 Chr.22: 42640099-42640099 on Build GRCh38
rs78719899 Chr.22: 42639855-42639857 on Build GRCh38
rs750727528 Chr.22: 42640031-42640031 on Build GRCh38
rs779995614 Chr.22: 42639976-42639976 on Build GRCh38
rs756554188 Chr.22: 42640132-42640132 on Build GRCh38
rs1007096168 Chr.22: 42639948-42639948 on Build GRCh38
rs764929242 Chr.22: 42640026-42640026 on Build GRCh38
rs757063209 Chr.22: 42640024-42640024 on Build GRCh38
rs773077267 Chr.22: 42640236-42640236 on Build GRCh38
rs368882948 Chr.22: 42640146-42640146 on Build GRCh38
rs75699688 Chr.22: 42639857-42639857 on Build GRCh38
rs760710393 Chr.22: 42640017-42640017 on Build GRCh38
rs559514039 Chr.22: 42640174-42640174 on Build GRCh38
rs754250757 Chr.22: 42640012-42640013 on Build GRCh38
rs756676331 Chr.22: 42640189-42640189 on Build GRCh38
rs756087098 Chr.22: 42639952-42639952 on Build GRCh38
rs866590993 Chr.22: 42640183-42640183 on Build GRCh38
rs764084435 Chr.22: 42640020-42640020 on Build GRCh38
rs775172170 Chr.22: 42640073-42640073 on Build GRCh38
rs766921788 Chr.22: 42639931-42639931 on Build GRCh38
rs761862172 Chr.22: 42640157-42640157 on Build GRCh38
rs758672282 Chr.22: 42640103-42640103 on Build GRCh38
rs1053445154 Chr.22: 42640270-42640270 on Build GRCh38
rs752745380 Chr.22: 42639949-42639949 on Build GRCh38
rs767092840 Chr.22: 42640012-42640012 on Build GRCh38
rs774107546 Chr.22: 42640006-42640006 on Build GRCh38
rs766517606 Chr.22: 42640104-42640104 on Build GRCh38
rs760196781 Chr.22: 42640074-42640074 on Build GRCh38
rs766908135 Chr.22: 42640006-42640007 on Build GRCh38
rs752249798 Chr.22: 42639989-42639989 on Build GRCh38
rs761239976 Chr.22: 42640001-42640004 on Build GRCh38
rs796115033 Chr.22: 42639856-42639857 on Build GRCh38
rs188021718 Chr.22: 42640071-42640071 on Build GRCh38
rs773061051 Chr.22: 42640166-42640166 on Build GRCh38
rs779417584 Chr.22: 42640195-42640195 on Build GRCh38
rs764141942 Chr.22: 42640083-42640083 on Build GRCh38
rs145437748 Chr.22: 42639988-42639988 on Build GRCh38
rs750167355 Chr.22: 42640102-42640102 on Build GRCh38
rs980646700 Chr.22: 42640176-42640176 on Build GRCh38
rs565966885 Chr.22: 42639870-42639870 on Build GRCh38
rs752233265 Chr.22: 42640013-42640013 on Build GRCh38
rs780233162 Chr.22: 42640041-42640041 on Build GRCh38
rs796330665 Chr.22: 42639857-42639858 on Build GRCh38
rs780875840 Chr.22: 42640214-42640214 on Build GRCh38
rs148856876 Chr.22: 42640258-42640258 on Build GRCh38
rs779126083 Chr.22: 42639972-42639972 on Build GRCh38
rs758521854 Chr.22: 42640035-42640035 on Build GRCh38
rs777759825 Chr.22: 42639956-42639956 on Build GRCh38
rs770501144 Chr.22: 42639997-42639997 on Build GRCh38
rs564579475 Chr.22: 42640196-42640196 on Build GRCh38
rs375276982 Chr.22: 42640163-42640163 on Build GRCh38
rs745582457 Chr.22: 42640068-42640068 on Build GRCh38
rs781619084 Chr.22: 42640047-42640047 on Build GRCh38

View Full Product Details